Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113651564-113652062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4B
Entrez Gene ID
496
Gene Name
ATPase H+/K+ transporting beta subunit
Gene Aliases
ATP6B
UniGene
Hs.434202
Gene Chromosome Location
Chr.13: 113648804-113658198 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
UniGene
Hs.103501
Gene Chromosome Location
Chr.13: 113645771-113735664 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138044 c.605C>T p.A202V Chr.13: 113651678-113651678 on Build GRCh38
945638 c.581G>A p.G194D Chr.13: 113651702-113651702 on Build GRCh38
945639 c.579C>T p.N193N Chr.13: 113651704-113651704 on Build GRCh38
945637 c.582C>A p.G194G Chr.13: 113651701-113651701 on Build GRCh38
7329444 c.608T>C p.F203S Chr.13: 113651675-113651675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970313197 Chr.13: 113651793-113651793 on Build GRCh38
rs901530068 Chr.13: 113651711-113651711 on Build GRCh38
rs577904798 Chr.13: 113651644-113651644 on Build GRCh38
rs755040637 Chr.13: 113651678-113651678 on Build GRCh38
rs757749709 Chr.13: 113651689-113651689 on Build GRCh38
rs1019050351 Chr.13: 113651977-113651977 on Build GRCh38
rs924253013 Chr.13: 113651798-113651798 on Build GRCh38
rs368745387 Chr.13: 113651704-113651704 on Build GRCh38
rs12427449 Chr.13: 113651817-113651817 on Build GRCh38
rs777500183 Chr.13: 113651770-113651770 on Build GRCh38
rs759709961 Chr.13: 113651735-113651735 on Build GRCh38
rs569166317 Chr.13: 113652008-113652008 on Build GRCh38
rs538945627 Chr.13: 113651651-113651651 on Build GRCh38
rs773058837 Chr.13: 113651662-113651662 on Build GRCh38
rs1006333453 Chr.13: 113652012-113652012 on Build GRCh38
rs995101093 Chr.13: 113651927-113651927 on Build GRCh38
rs556990604 Chr.13: 113651980-113651980 on Build GRCh38
rs889322918 Chr.13: 113651936-113651936 on Build GRCh38
rs774460903 Chr.13: 113651777-113651777 on Build GRCh38
rs915697292 Chr.13: 113651686-113651686 on Build GRCh38
rs767595440 Chr.13: 113651740-113651740 on Build GRCh38
rs779409641 Chr.13: 113651755-113651755 on Build GRCh38
rs747801448 Chr.13: 113651703-113651703 on Build GRCh38
rs541124613 Chr.13: 113651747-113651747 on Build GRCh38
rs766621067 Chr.13: 113651670-113651670 on Build GRCh38
rs778166654 Chr.13: 113651646-113651646 on Build GRCh38
rs764189454 Chr.13: 113651743-113651743 on Build GRCh38
rs1045202815 Chr.13: 113651826-113651826 on Build GRCh38
rs879793671 Chr.13: 113652005-113652005 on Build GRCh38
rs906495254 Chr.13: 113651590-113651590 on Build GRCh38
rs573826285 Chr.13: 113651726-113651726 on Build GRCh38
rs1031339492 Chr.13: 113651993-113651993 on Build GRCh38
rs957319025 Chr.13: 113651810-113651810 on Build GRCh38
rs767873013 Chr.13: 113652004-113652004 on Build GRCh38
rs764007657 Chr.13: 113651679-113651679 on Build GRCh38
rs374655962 Chr.13: 113651658-113651658 on Build GRCh38
rs749106748 Chr.13: 113651771-113651771 on Build GRCh38
rs545381324 Chr.13: 113651773-113651773 on Build GRCh38
rs530436943 Chr.13: 113651816-113651816 on Build GRCh38
rs901474742 Chr.13: 113651990-113651990 on Build GRCh38
rs546679116 Chr.13: 113651905-113651905 on Build GRCh38
rs1023155916 Chr.13: 113651787-113651787 on Build GRCh38
rs555389170 Chr.13: 113652040-113652040 on Build GRCh38
rs369199790 Chr.13: 113651655-113651655 on Build GRCh38
rs962552375 Chr.13: 113651611-113651611 on Build GRCh38
rs367873321 Chr.13: 113652036-113652036 on Build GRCh38
rs376283075 Chr.13: 113651979-113651979 on Build GRCh38
rs897784644 Chr.13: 113651889-113651889 on Build GRCh38
rs751875907 Chr.13: 113651626-113651626 on Build GRCh38
rs762958937 Chr.13: 113651663-113651663 on Build GRCh38
rs985415554 Chr.13: 113651603-113651603 on Build GRCh38
rs546298527 Chr.13: 113651991-113651991 on Build GRCh38
rs544792621 Chr.13: 113651698-113651698 on Build GRCh38
rs990118611 Chr.13: 113652007-113652007 on Build GRCh38
rs557176544 Chr.13: 113651659-113651659 on Build GRCh38
rs201009622 Chr.13: 113651749-113651749 on Build GRCh38
rs536135249 Chr.13: 113652023-113652023 on Build GRCh38
rs372844884 Chr.13: 113651657-113651657 on Build GRCh38
rs768191657 Chr.13: 113651656-113651656 on Build GRCh38
rs928015411 Chr.13: 113651841-113651841 on Build GRCh38
rs754483380 Chr.13: 113651758-113651758 on Build GRCh38
rs374169282 Chr.13: 113651741-113651741 on Build GRCh38
rs769480619 Chr.13: 113651768-113651768 on Build GRCh38
rs775684906 Chr.13: 113651676-113651676 on Build GRCh38
rs372676406 Chr.13: 113651748-113651748 on Build GRCh38
rs756467882 Chr.13: 113651682-113651682 on Build GRCh38
rs763050139 Chr.13: 113651721-113651721 on Build GRCh38
rs543766251 Chr.13: 113651699-113651699 on Build GRCh38
rs957260184 Chr.13: 113652001-113652001 on Build GRCh38
rs533309026 Chr.13: 113651754-113651754 on Build GRCh38
rs1011257015 Chr.13: 113651931-113651931 on Build GRCh38
rs368921019 Chr.13: 113651694-113651694 on Build GRCh38
rs977077584 Chr.13: 113651786-113651786 on Build GRCh38
rs376524272 Chr.13: 113651654-113651654 on Build GRCh38
rs139310316 Chr.13: 113651594-113651594 on Build GRCh38
rs201861369 Chr.13: 113651687-113651687 on Build GRCh38
rs372348110 Chr.13: 113651725-113651725 on Build GRCh38
rs752914990 Chr.13: 113651680-113651680 on Build GRCh38
rs756442964 Chr.13: 113651645-113651645 on Build GRCh38
rs767647116 Chr.13: 113651679-113651679 on Build GRCh38
rs10161594 Chr.13: 113651928-113651928 on Build GRCh38
rs939392253 Chr.13: 113651871-113651871 on Build GRCh38
rs966208731 Chr.13: 113651588-113651588 on Build GRCh38
rs749845867 Chr.13: 113651688-113651688 on Build GRCh38
rs773125482 Chr.13: 113651706-113651706 on Build GRCh38
rs143217818 Chr.13: 113651968-113651968 on Build GRCh38
rs948552947 Chr.13: 113651819-113651819 on Build GRCh38
rs780727607 Chr.13: 113651760-113651760 on Build GRCh38
rs776175770 Chr.13: 113651702-113651702 on Build GRCh38
rs528238846 Chr.13: 113651872-113651872 on Build GRCh38
rs377601482 Chr.13: 113651652-113651652 on Build GRCh38
rs1023650197 Chr.13: 113651589-113651589 on Build GRCh38
rs755236964 Chr.13: 113651627-113651627 on Build GRCh38
rs563979435 Chr.13: 113651795-113651795 on Build GRCh38
rs973577609 Chr.13: 113651614-113651614 on Build GRCh38
rs747810806 Chr.13: 113651763-113651763 on Build GRCh38
rs143404371 Chr.13: 113651593-113651593 on Build GRCh38

View Full Product Details