Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113653185-113653728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4B
Entrez Gene ID
496
Gene Name
ATPase H+/K+ transporting beta subunit
Gene Aliases
ATP6B
UniGene
Hs.434202
Gene Chromosome Location
Chr.13: 113648804-113658198 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
UniGene
Hs.103501
Gene Chromosome Location
Chr.13: 113645771-113735664 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2264468 c.278G>A p.G93D Chr.13: 113653398-113653398 on Build GRCh38
6499649 c.257C>G p.P86R Chr.13: 113653419-113653419 on Build GRCh38
1741974 c.302C>G p.S101C Chr.13: 113653374-113653374 on Build GRCh38
1365627 c.268G>A p.G90R Chr.13: 113653408-113653408 on Build GRCh38
5027814 c.258G>A p.P86P Chr.13: 113653418-113653418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015787833 Chr.13: 113653625-113653625 on Build GRCh38
rs891894613 Chr.13: 113653380-113653380 on Build GRCh38
rs1015189536 Chr.13: 113653300-113653300 on Build GRCh38
rs759355813 Chr.13: 113653318-113653318 on Build GRCh38
rs375087478 Chr.13: 113653242-113653242 on Build GRCh38
rs764130095 Chr.13: 113653463-113653463 on Build GRCh38
rs146718679 Chr.13: 113653399-113653399 on Build GRCh38
rs748629488 Chr.13: 113653290-113653308 on Build GRCh38
rs772594790 Chr.13: 113653666-113653673 on Build GRCh38
rs375894425 Chr.13: 113653333-113653333 on Build GRCh38
rs759069762 Chr.13: 113653405-113653405 on Build GRCh38
rs935640139 Chr.13: 113653529-113653529 on Build GRCh38
rs774231376 Chr.13: 113653661-113653661 on Build GRCh38
rs538508691 Chr.13: 113653688-113653688 on Build GRCh38
rs757305267 Chr.13: 113653347-113653347 on Build GRCh38
rs960221081 Chr.13: 113653417-113653417 on Build GRCh38
rs765585202 Chr.13: 113653419-113653419 on Build GRCh38
rs879014161 Chr.13: 113653303-113653303 on Build GRCh38
rs753869057 Chr.13: 113653345-113653345 on Build GRCh38
rs1005334371 Chr.13: 113653676-113653676 on Build GRCh38
rs559890514 Chr.13: 113653238-113653238 on Build GRCh38
rs781753159 Chr.13: 113653285-113653303 on Build GRCh38
rs531765507 Chr.13: 113653571-113653574 on Build GRCh38
rs781382411 Chr.13: 113653445-113653445 on Build GRCh38
rs767267609 Chr.13: 113653326-113653326 on Build GRCh38
rs747580982 Chr.13: 113653373-113653373 on Build GRCh38
rs200364303 Chr.13: 113653424-113653424 on Build GRCh38
rs755458376 Chr.13: 113653508-113653508 on Build GRCh38
rs780547171 Chr.13: 113653368-113653368 on Build GRCh38
rs755991470 Chr.13: 113653336-113653336 on Build GRCh38
rs776247699 Chr.13: 113653471-113653471 on Build GRCh38
rs983818141 Chr.13: 113653285-113653285 on Build GRCh38
rs1030218628 Chr.13: 113653361-113653361 on Build GRCh38
rs1055780888 Chr.13: 113653629-113653629 on Build GRCh38
rs141052353 Chr.13: 113653393-113653393 on Build GRCh38
rs550730170 Chr.13: 113653515-113653515 on Build GRCh38
rs773371291 Chr.13: 113653438-113653439 on Build GRCh38
rs548662765 Chr.13: 113653550-113653550 on Build GRCh38
rs559217478 Chr.13: 113653666-113653666 on Build GRCh38
rs199662536 Chr.13: 113653280-113653280 on Build GRCh38
rs937164947 Chr.13: 113653243-113653243 on Build GRCh38
rs763782100 Chr.13: 113653409-113653409 on Build GRCh38
rs1001230708 Chr.13: 113653258-113653258 on Build GRCh38
rs1033601306 Chr.13: 113653668-113653668 on Build GRCh38
rs552041599 Chr.13: 113653264-113653264 on Build GRCh38
rs878939603 Chr.13: 113653524-113653524 on Build GRCh38
rs777976903 Chr.13: 113653343-113653343 on Build GRCh38
rs573466734 Chr.13: 113653289-113653289 on Build GRCh38
rs942623406 Chr.13: 113653301-113653301 on Build GRCh38
rs1043068603 Chr.13: 113653394-113653394 on Build GRCh38
rs142872172 Chr.13: 113653367-113653367 on Build GRCh38
rs534360499 Chr.13: 113653635-113653635 on Build GRCh38
rs767385897 Chr.13: 113653406-113653406 on Build GRCh38
rs374221658 Chr.13: 113653461-113653461 on Build GRCh38
rs748578826 Chr.13: 113653446-113653446 on Build GRCh38
rs946193089 Chr.13: 113653545-113653545 on Build GRCh38
rs113555226 Chr.13: 113653231-113653231 on Build GRCh38
rs151052185 Chr.13: 113653378-113653378 on Build GRCh38
rs769399039 Chr.13: 113653274-113653274 on Build GRCh38
rs960190156 Chr.13: 113653262-113653262 on Build GRCh38
rs184063977 Chr.13: 113653284-113653284 on Build GRCh38
rs151211765 Chr.13: 113653524-113653525 on Build GRCh38
rs1038416129 Chr.13: 113653237-113653237 on Build GRCh38
rs922504095 Chr.13: 113653519-113653519 on Build GRCh38
rs770523181 Chr.13: 113653449-113653449 on Build GRCh38
rs913689346 Chr.13: 113653528-113653528 on Build GRCh38
rs1035857908 Chr.13: 113653277-113653277 on Build GRCh38
rs933812229 Chr.13: 113653334-113653334 on Build GRCh38
rs188356364 Chr.13: 113653377-113653377 on Build GRCh38
rs772588960 Chr.13: 113653281-113653281 on Build GRCh38
rs761235905 Chr.13: 113653273-113653273 on Build GRCh38
rs1043528506 Chr.13: 113653546-113653546 on Build GRCh38
rs769251679 Chr.13: 113653280-113653281 on Build GRCh38
rs190216434 Chr.13: 113653673-113653673 on Build GRCh38
rs369809506 Chr.13: 113653408-113653408 on Build GRCh38
rs770018994 Chr.13: 113653289-113653290 on Build GRCh38
rs141704998 Chr.13: 113653456-113653456 on Build GRCh38
rs576793214 Chr.13: 113653308-113653308 on Build GRCh38
rs376655942 Chr.13: 113653428-113653428 on Build GRCh38
rs969008330 Chr.13: 113653239-113653239 on Build GRCh38
rs763243876 Chr.13: 113653475-113653477 on Build GRCh38
rs1014887475 Chr.13: 113653664-113653664 on Build GRCh38
rs370965119 Chr.13: 113653525-113653525 on Build GRCh38
rs891279766 Chr.13: 113653580-113653580 on Build GRCh38
rs372485483 Chr.13: 113653288-113653288 on Build GRCh38
rs896083205 Chr.13: 113653652-113653652 on Build GRCh38
rs538630381 Chr.13: 113653269-113653269 on Build GRCh38
rs949904463 Chr.13: 113653685-113653685 on Build GRCh38
rs530259810 Chr.13: 113653549-113653549 on Build GRCh38
rs558936934 Chr.13: 113653307-113653307 on Build GRCh38
rs369324820 Chr.13: 113653287-113653287 on Build GRCh38
rs147966257 Chr.13: 113653418-113653418 on Build GRCh38
rs573499987 Chr.13: 113653636-113653636 on Build GRCh38
rs760505999 Chr.13: 113653462-113653462 on Build GRCh38
rs771565671 Chr.13: 113653459-113653459 on Build GRCh38
rs527624239 Chr.13: 113653261-113653261 on Build GRCh38
rs749487200 Chr.13: 113653437-113653437 on Build GRCh38
rs551072875 Chr.13: 113653272-113653272 on Build GRCh38
rs1016626255 Chr.13: 113653696-113653696 on Build GRCh38
rs746050736 Chr.13: 113653357-113653357 on Build GRCh38
rs749093021 Chr.13: 113653219-113653241 on Build GRCh38
rs200666442 Chr.13: 113653421-113653421 on Build GRCh38
rs779302172 Chr.13: 113653352-113653352 on Build GRCh38
rs377650427 Chr.13: 113653351-113653351 on Build GRCh38
rs571620063 Chr.13: 113653265-113653265 on Build GRCh38
rs755561448 Chr.13: 113653440-113653440 on Build GRCh38
rs552184407 Chr.13: 113653490-113653490 on Build GRCh38
rs1042021722 Chr.13: 113653516-113653516 on Build GRCh38
rs201906864 Chr.13: 113653379-113653379 on Build GRCh38
rs371178123 Chr.13: 113653458-113653458 on Build GRCh38
rs141298331 Chr.13: 113653673-113653673 on Build GRCh38
rs567075018 Chr.13: 113653585-113653585 on Build GRCh38
rs534506913 Chr.13: 113653306-113653306 on Build GRCh38
rs747318650 Chr.13: 113653271-113653288 on Build GRCh38
rs941371631 Chr.13: 113653220-113653221 on Build GRCh38

View Full Product Details