Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113832213-113832719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4046247 c.1056C>T p.A352A Chr.13: 113832386-113832386 on Build GRCh38
4519512 c.1013G>A p.G338E Chr.13: 113832429-113832429 on Build GRCh38
550774 c.991G>A p.E331K Chr.13: 113832451-113832451 on Build GRCh38
4046248 c.1185C>T p.A395A Chr.13: 113832386-113832386 on Build GRCh38
244661 c.1061G>A p.R354Q Chr.13: 113832381-113832381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755494619 Chr.13: 113832416-113832416 on Build GRCh38
rs374445149 Chr.13: 113832404-113832404 on Build GRCh38
rs879336735 Chr.13: 113832373-113832373 on Build GRCh38
rs758057920 Chr.13: 113832519-113832519 on Build GRCh38
rs959882100 Chr.13: 113832509-113832509 on Build GRCh38
rs779039704 Chr.13: 113832604-113832604 on Build GRCh38
rs776397737 Chr.13: 113832357-113832357 on Build GRCh38
rs1027443738 Chr.13: 113832568-113832568 on Build GRCh38
rs1047289936 Chr.13: 113832592-113832592 on Build GRCh38
rs202194869 Chr.13: 113832331-113832331 on Build GRCh38
rs554977857 Chr.13: 113832546-113832546 on Build GRCh38
rs542438763 Chr.13: 113832672-113832672 on Build GRCh38
rs368353540 Chr.13: 113832470-113832470 on Build GRCh38
rs371080284 Chr.13: 113832440-113832440 on Build GRCh38
rs376044728 Chr.13: 113832288-113832288 on Build GRCh38
rs149144442 Chr.13: 113832692-113832692 on Build GRCh38
rs776653031 Chr.13: 113832284-113832284 on Build GRCh38
rs770495975 Chr.13: 113832261-113832261 on Build GRCh38
rs775850939 Chr.13: 113832569-113832569 on Build GRCh38
rs202125638 Chr.13: 113832482-113832482 on Build GRCh38
rs757506592 Chr.13: 113832507-113832507 on Build GRCh38
rs894015706 Chr.13: 113832522-113832522 on Build GRCh38
rs759596582 Chr.13: 113832494-113832494 on Build GRCh38
rs767962743 Chr.13: 113832390-113832390 on Build GRCh38
rs1039486807 Chr.13: 113832557-113832557 on Build GRCh38
rs550806526 Chr.13: 113832500-113832500 on Build GRCh38
rs567124992 Chr.13: 113832556-113832556 on Build GRCh38
rs753633530 Chr.13: 113832681-113832681 on Build GRCh38
rs201876951 Chr.13: 113832264-113832264 on Build GRCh38
rs144457857 Chr.13: 113832403-113832403 on Build GRCh38
rs753479956 Chr.13: 113832332-113832332 on Build GRCh38
rs747268075 Chr.13: 113832252-113832252 on Build GRCh38
rs747914644 Chr.13: 113832452-113832452 on Build GRCh38
rs149844317 Chr.13: 113832508-113832508 on Build GRCh38
rs200492385 Chr.13: 113832345-113832345 on Build GRCh38
rs776076149 Chr.13: 113832641-113832641 on Build GRCh38
rs760098401 Chr.13: 113832389-113832389 on Build GRCh38
rs767016321 Chr.13: 113832671-113832671 on Build GRCh38
rs773471138 Chr.13: 113832379-113832379 on Build GRCh38
rs540191352 Chr.13: 113832278-113832278 on Build GRCh38
rs558548139 Chr.13: 113832582-113832582 on Build GRCh38
rs200350506 Chr.13: 113832437-113832437 on Build GRCh38
rs141287368 Chr.13: 113832658-113832658 on Build GRCh38
rs1024053144 Chr.13: 113832348-113832348 on Build GRCh38
rs566553692 Chr.13: 113832466-113832466 on Build GRCh38
rs748976929 Chr.13: 113832253-113832253 on Build GRCh38
rs749092015 Chr.13: 113832597-113832597 on Build GRCh38
rs544430070 Chr.13: 113832349-113832349 on Build GRCh38
rs944398250 Chr.13: 113832439-113832439 on Build GRCh38
rs779817827 Chr.13: 113832353-113832353 on Build GRCh38
rs371350136 Chr.13: 113832631-113832631 on Build GRCh38
rs750039866 Chr.13: 113832514-113832514 on Build GRCh38
rs749742211 Chr.13: 113832591-113832591 on Build GRCh38
rs41284480 Chr.13: 113832563-113832563 on Build GRCh38
rs374837053 Chr.13: 113832682-113832682 on Build GRCh38
rs373974683 Chr.13: 113832526-113832526 on Build GRCh38
rs529145541 Chr.13: 113832363-113832363 on Build GRCh38
rs781147555 Chr.13: 113832451-113832451 on Build GRCh38
rs373825026 Chr.13: 113832364-113832364 on Build GRCh38
rs750498523 Chr.13: 113832289-113832289 on Build GRCh38
rs145302934 Chr.13: 113832664-113832664 on Build GRCh38
rs748113385 Chr.13: 113832538-113832538 on Build GRCh38
rs772313363 Chr.13: 113832619-113832619 on Build GRCh38
rs777752250 Chr.13: 113832679-113832679 on Build GRCh38
rs774474465 Chr.13: 113832493-113832493 on Build GRCh38
rs147632674 Chr.13: 113832691-113832691 on Build GRCh38
rs140003424 Chr.13: 113832445-113832445 on Build GRCh38
rs944452622 Chr.13: 113832315-113832315 on Build GRCh38
rs756029174 Chr.13: 113832674-113832674 on Build GRCh38
rs771638195 Chr.13: 113832350-113832350 on Build GRCh38
rs147823988 Chr.13: 113832431-113832431 on Build GRCh38
rs766773434 Chr.13: 113832300-113832300 on Build GRCh38
rs759006256 Chr.13: 113832667-113832667 on Build GRCh38
rs114684411 Chr.13: 113832383-113832383 on Build GRCh38
rs555163572 Chr.13: 113832665-113832665 on Build GRCh38
rs746972667 Chr.13: 113832355-113832355 on Build GRCh38
rs185797569 Chr.13: 113832281-113832281 on Build GRCh38
rs372048005 Chr.13: 113832524-113832524 on Build GRCh38
rs771176163 Chr.13: 113832603-113832603 on Build GRCh38
rs113785834 Chr.13: 113832338-113832338 on Build GRCh38
rs775549947 Chr.13: 113832273-113832273 on Build GRCh38
rs757783247 Chr.13: 113832426-113832426 on Build GRCh38
rs562664965 Chr.13: 113832356-113832356 on Build GRCh38
rs1038992903 Chr.13: 113832650-113832650 on Build GRCh38
rs200744856 Chr.13: 113832643-113832643 on Build GRCh38
rs375897325 Chr.13: 113832659-113832659 on Build GRCh38
rs778576355 Chr.13: 113832263-113832263 on Build GRCh38
rs935732068 Chr.13: 113832645-113832645 on Build GRCh38
rs953248692 Chr.13: 113832572-113832572 on Build GRCh38
rs769599558 Chr.13: 113832465-113832465 on Build GRCh38
rs773808304 Chr.13: 113832684-113832684 on Build GRCh38
rs761084638 Chr.13: 113832505-113832505 on Build GRCh38
rs746206978 Chr.13: 113832539-113832539 on Build GRCh38
rs906798049 Chr.13: 113832690-113832690 on Build GRCh38
rs200979235 Chr.13: 113832327-113832327 on Build GRCh38
rs779159581 Chr.13: 113832694-113832694 on Build GRCh38
rs201261387 Chr.13: 113832530-113832530 on Build GRCh38
rs370199266 Chr.13: 113832400-113832400 on Build GRCh38
rs752128196 Chr.13: 113832326-113832326 on Build GRCh38
rs745915511 Chr.13: 113832614-113832614 on Build GRCh38
rs1005804227 Chr.13: 113832635-113832635 on Build GRCh38
rs1018868205 Chr.13: 113832596-113832596 on Build GRCh38
rs768102646 Chr.13: 113832498-113832498 on Build GRCh38
rs548166528 Chr.13: 113832381-113832381 on Build GRCh38
rs371599789 Chr.13: 113832608-113832608 on Build GRCh38
rs766013191 Chr.13: 113832523-113832523 on Build GRCh38
rs763061345 Chr.13: 113832299-113832299 on Build GRCh38
rs769346633 Chr.13: 113832655-113832655 on Build GRCh38
rs946016412 Chr.13: 113832536-113832536 on Build GRCh38
rs377242538 Chr.13: 113832386-113832386 on Build GRCh38
rs777555815 Chr.13: 113832589-113832589 on Build GRCh38

View Full Product Details