Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18911881-18912380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFFO2
Entrez Gene ID
126917
Gene Name
intermediate filament family orphan 2
Gene Aliases
-
UniGene
Hs.466625
Gene Chromosome Location
Chr.1: 18904280-18956672 on Build GRCh38
Transcripts
NM_001136265.1 XM_011540630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295598 c.1224+6C>A p.? Chr.1: 18911957-18911957 on Build GRCh38
6506982 c.1128C>T p.D376D Chr.1: 18912059-18912059 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs56172147 Chr.1: 18912200-18912200 on Build GRCh38
rs774198059 Chr.1: 18911914-18911914 on Build GRCh38
rs993631162 Chr.1: 18911913-18911913 on Build GRCh38
rs1017825428 Chr.1: 18911991-18911991 on Build GRCh38
rs3828046 Chr.1: 18912317-18912317 on Build GRCh38
rs955061261 Chr.1: 18912329-18912329 on Build GRCh38
rs890584996 Chr.1: 18911929-18911929 on Build GRCh38
rs770609043 Chr.1: 18912127-18912127 on Build GRCh38
rs758784110 Chr.1: 18911986-18911986 on Build GRCh38
rs753943445 Chr.1: 18911978-18911978 on Build GRCh38
rs972407513 Chr.1: 18911990-18911990 on Build GRCh38
rs1047476785 Chr.1: 18911924-18911924 on Build GRCh38
rs974822773 Chr.1: 18912101-18912101 on Build GRCh38
rs1009085389 Chr.1: 18912322-18912322 on Build GRCh38
rs767467659 Chr.1: 18911952-18911952 on Build GRCh38
rs906067649 Chr.1: 18912232-18912232 on Build GRCh38
rs757623590 Chr.1: 18911979-18911979 on Build GRCh38
rs202081563 Chr.1: 18911908-18911908 on Build GRCh38
rs936058313 Chr.1: 18912154-18912154 on Build GRCh38
rs61750842 Chr.1: 18912074-18912074 on Build GRCh38
rs1001318141 Chr.1: 18911951-18911951 on Build GRCh38
rs17492693 Chr.1: 18912035-18912035 on Build GRCh38
rs760814726 Chr.1: 18911966-18911966 on Build GRCh38
rs1016719318 Chr.1: 18912257-18912257 on Build GRCh38
rs1045972765 Chr.1: 18912194-18912194 on Build GRCh38
rs764023535 Chr.1: 18911977-18911977 on Build GRCh38
rs950355762 Chr.1: 18912181-18912181 on Build GRCh38
rs180830770 Chr.1: 18912331-18912331 on Build GRCh38
rs993895011 Chr.1: 18912307-18912307 on Build GRCh38
rs765527973 Chr.1: 18911982-18911982 on Build GRCh38
rs891755674 Chr.1: 18912177-18912177 on Build GRCh38
rs568319398 Chr.1: 18912311-18912311 on Build GRCh38
rs1053547866 Chr.1: 18912167-18912167 on Build GRCh38
rs796130593 Chr.1: 18912319-18912319 on Build GRCh38
rs755663116 Chr.1: 18912059-18912059 on Build GRCh38
rs879025665 Chr.1: 18911909-18911909 on Build GRCh38
rs77352288 Chr.1: 18912277-18912277 on Build GRCh38
rs56042056 Chr.1: 18912310-18912310 on Build GRCh38
rs989260585 Chr.1: 18912081-18912081 on Build GRCh38
rs201056277 Chr.1: 18912310-18912311 on Build GRCh38
rs759294395 Chr.1: 18911936-18911936 on Build GRCh38
rs77793103 Chr.1: 18912146-18912146 on Build GRCh38
rs775253305 Chr.1: 18911965-18911965 on Build GRCh38
rs1001682865 Chr.1: 18912247-18912247 on Build GRCh38
rs955475653 Chr.1: 18912041-18912041 on Build GRCh38
rs768831114 Chr.1: 18911946-18911946 on Build GRCh38
rs780638009 Chr.1: 18912030-18912030 on Build GRCh38
rs200746091 Chr.1: 18912319-18912319 on Build GRCh38
rs986703377 Chr.1: 18912062-18912062 on Build GRCh38
rs928996012 Chr.1: 18912291-18912291 on Build GRCh38
rs937902755 Chr.1: 18912172-18912172 on Build GRCh38
rs200305590 Chr.1: 18911985-18911985 on Build GRCh38
rs758983984 Chr.1: 18912343-18912343 on Build GRCh38
rs964938769 Chr.1: 18912020-18912020 on Build GRCh38
rs144158925 Chr.1: 18912086-18912086 on Build GRCh38
rs537336966 Chr.1: 18912140-18912140 on Build GRCh38

View Full Product Details