Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20663929-20664436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF17
Entrez Gene ID
57576
Gene Name
kinesin family member 17
Gene Aliases
KIF17B, KIF3X, KLP-2, OSM-3
UniGene
Hs.130411
Gene Chromosome Location
Chr.1: 20662294-20718017 on Build GRCh38
Transcripts
NM_001122819.2 NM_001287212.1 NM_020816.3 XM_005245950.3 XM_005245951.3 XM_011541837.1 XM_011541838.1 XM_011541839.2 XM_011541840.2 XM_011541841.2 XM_011541842.1 XM_011541843.1 XM_011541844.2 XM_011541845.2 XM_011541846.2 XM_011541847.2

SNP Detail

SNP ID Chromosome Location
rs148080180 Chr.1: 20664411-20664411 on Build GRCh38
rs554125970 Chr.1: 20663970-20663970 on Build GRCh38
rs549433773 Chr.1: 20664401-20664401 on Build GRCh38
rs1014600069 Chr.1: 20664079-20664079 on Build GRCh38
rs1057244565 Chr.1: 20664190-20664190 on Build GRCh38
rs573881589 Chr.1: 20664156-20664156 on Build GRCh38
rs886631905 Chr.1: 20664004-20664004 on Build GRCh38
rs1044879112 Chr.1: 20664210-20664210 on Build GRCh38
rs576097795 Chr.1: 20664091-20664091 on Build GRCh38
rs962092209 Chr.1: 20664082-20664082 on Build GRCh38
rs893177238 Chr.1: 20664392-20664392 on Build GRCh38
rs117696159 Chr.1: 20664101-20664101 on Build GRCh38
rs746601433 Chr.1: 20664150-20664150 on Build GRCh38
rs112389271 Chr.1: 20664105-20664105 on Build GRCh38
rs981236471 Chr.1: 20664198-20664198 on Build GRCh38
rs145698899 Chr.1: 20664297-20664297 on Build GRCh38
rs780502488 Chr.1: 20664387-20664387 on Build GRCh38
rs1056700498 Chr.1: 20663972-20663972 on Build GRCh38
rs908292759 Chr.1: 20664312-20664312 on Build GRCh38
rs928537285 Chr.1: 20664222-20664222 on Build GRCh38
rs779058763 Chr.1: 20664088-20664088 on Build GRCh38
rs902064451 Chr.1: 20664056-20664056 on Build GRCh38
rs938744406 Chr.1: 20664139-20664139 on Build GRCh38
rs924676998 Chr.1: 20664134-20664134 on Build GRCh38
rs958586462 Chr.1: 20664223-20664223 on Build GRCh38
rs530951155 Chr.1: 20664329-20664329 on Build GRCh38
rs559640591 Chr.1: 20664250-20664250 on Build GRCh38
rs10799659 Chr.1: 20664412-20664412 on Build GRCh38
rs535032914 Chr.1: 20664108-20664108 on Build GRCh38
rs1051099162 Chr.1: 20664391-20664391 on Build GRCh38
rs955933228 Chr.1: 20664320-20664320 on Build GRCh38
rs967240568 Chr.1: 20664187-20664187 on Build GRCh38
rs533400943 Chr.1: 20664278-20664278 on Build GRCh38
rs887661085 Chr.1: 20664211-20664211 on Build GRCh38
rs947865578 Chr.1: 20664201-20664201 on Build GRCh38
rs1030904226 Chr.1: 20664093-20664093 on Build GRCh38
rs930955509 Chr.1: 20664277-20664277 on Build GRCh38
rs1020787821 Chr.1: 20664125-20664125 on Build GRCh38
rs947628817 Chr.1: 20664119-20664119 on Build GRCh38
rs951980250 Chr.1: 20664047-20664047 on Build GRCh38
rs572344458 Chr.1: 20663981-20663981 on Build GRCh38
rs562959066 Chr.1: 20664196-20664196 on Build GRCh38
rs977819220 Chr.1: 20664122-20664122 on Build GRCh38
rs140575156 Chr.1: 20664074-20664074 on Build GRCh38
rs996472122 Chr.1: 20664390-20664390 on Build GRCh38
rs986266119 Chr.1: 20664405-20664405 on Build GRCh38
rs545487856 Chr.1: 20664020-20664020 on Build GRCh38
rs1012103833 Chr.1: 20664012-20664012 on Build GRCh38
rs1005004669 Chr.1: 20664010-20664010 on Build GRCh38
rs563689480 Chr.1: 20664328-20664328 on Build GRCh38
rs777534920 Chr.1: 20664115-20664115 on Build GRCh38
rs1047237773 Chr.1: 20663965-20663965 on Build GRCh38
rs902118131 Chr.1: 20664373-20664373 on Build GRCh38
rs919630375 Chr.1: 20664272-20664272 on Build GRCh38

View Full Product Details