Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20682571-20683075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF17
Entrez Gene ID
57576
Gene Name
kinesin family member 17
Gene Aliases
KIF17B, KIF3X, KLP-2, OSM-3
UniGene
Hs.130411
Gene Chromosome Location
Chr.1: 20662294-20718017 on Build GRCh38
Transcripts
NM_001122819.2 NM_001287212.1 NM_020816.3 XM_005245950.3 XM_005245951.3 XM_011541837.1 XM_011541838.1 XM_011541839.2 XM_011541840.2 XM_011541841.2 XM_011541842.1 XM_011541843.1 XM_011541844.2 XM_011541845.2 XM_011541846.2 XM_011541847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6694850 c.2363C>A p.S788* Chr.1: 20682753-20682753 on Build GRCh38
6123878 c.2384G>T p.W795L Chr.1: 20682732-20682732 on Build GRCh38
5726102 c.2411T>C p.I804T Chr.1: 20682705-20682705 on Build GRCh38
300969 c.2283G>T p.K761N Chr.1: 20682833-20682833 on Build GRCh38
3667454 c.2379G>A p.G793G Chr.1: 20682737-20682737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369190162 Chr.1: 20682933-20682933 on Build GRCh38
rs539685340 Chr.1: 20682966-20682966 on Build GRCh38
rs200844482 Chr.1: 20682739-20682739 on Build GRCh38
rs115933091 Chr.1: 20683035-20683035 on Build GRCh38
rs116005658 Chr.1: 20682740-20682740 on Build GRCh38
rs748236046 Chr.1: 20682730-20682730 on Build GRCh38
rs549777277 Chr.1: 20682874-20682874 on Build GRCh38
rs766248541 Chr.1: 20682714-20682714 on Build GRCh38
rs923679040 Chr.1: 20683023-20683023 on Build GRCh38
rs531505555 Chr.1: 20682862-20682862 on Build GRCh38
rs761978631 Chr.1: 20682925-20682925 on Build GRCh38
rs967723113 Chr.1: 20682996-20682996 on Build GRCh38
rs776340258 Chr.1: 20682699-20682699 on Build GRCh38
rs765285634 Chr.1: 20682927-20682927 on Build GRCh38
rs112091782 Chr.1: 20682641-20682641 on Build GRCh38
rs146864100 Chr.1: 20682865-20682865 on Build GRCh38
rs747794960 Chr.1: 20682876-20682876 on Build GRCh38
rs750913451 Chr.1: 20682928-20682928 on Build GRCh38
rs999794935 Chr.1: 20682860-20682860 on Build GRCh38
rs761442869 Chr.1: 20682759-20682759 on Build GRCh38
rs369573433 Chr.1: 20682919-20682919 on Build GRCh38
rs372516027 Chr.1: 20682935-20682935 on Build GRCh38
rs762986160 Chr.1: 20682771-20682771 on Build GRCh38
rs756546397 Chr.1: 20682736-20682736 on Build GRCh38
rs768594043 Chr.1: 20682640-20682640 on Build GRCh38
rs764147289 Chr.1: 20682854-20682854 on Build GRCh38
rs373972661 Chr.1: 20682797-20682797 on Build GRCh38
rs775324323 Chr.1: 20682916-20682916 on Build GRCh38
rs374049433 Chr.1: 20682810-20682810 on Build GRCh38
rs775026398 Chr.1: 20682693-20682693 on Build GRCh38
rs368058864 Chr.1: 20682752-20682752 on Build GRCh38
rs759439067 Chr.1: 20682901-20682901 on Build GRCh38
rs749604072 Chr.1: 20682737-20682737 on Build GRCh38
rs985786933 Chr.1: 20682856-20682856 on Build GRCh38
rs370565385 Chr.1: 20682753-20682753 on Build GRCh38
rs748017247 Chr.1: 20683034-20683034 on Build GRCh38
rs139870251 Chr.1: 20682801-20682801 on Build GRCh38
rs116783305 Chr.1: 20682761-20682761 on Build GRCh38
rs375213890 Chr.1: 20682932-20682932 on Build GRCh38
rs386629315 Chr.1: 20682839-20682840 on Build GRCh38
rs199907647 Chr.1: 20682889-20682889 on Build GRCh38
rs768677101 Chr.1: 20682760-20682760 on Build GRCh38
rs972563859 Chr.1: 20682838-20682838 on Build GRCh38
rs781018860 Chr.1: 20682806-20682806 on Build GRCh38
rs760925611 Chr.1: 20682853-20682853 on Build GRCh38
rs1050038782 Chr.1: 20682807-20682807 on Build GRCh38
rs114545284 Chr.1: 20682691-20682691 on Build GRCh38
rs1022414146 Chr.1: 20682632-20682632 on Build GRCh38
rs555878262 Chr.1: 20682630-20682630 on Build GRCh38
rs116460177 Chr.1: 20683016-20683016 on Build GRCh38
rs151177164 Chr.1: 20682787-20682787 on Build GRCh38
rs995945498 Chr.1: 20682596-20682596 on Build GRCh38
rs952145988 Chr.1: 20682910-20682910 on Build GRCh38
rs578074180 Chr.1: 20682839-20682839 on Build GRCh38
rs114242123 Chr.1: 20682738-20682738 on Build GRCh38
rs115257742 Chr.1: 20682696-20682696 on Build GRCh38
rs769625439 Chr.1: 20682878-20682878 on Build GRCh38
rs200888075 Chr.1: 20682791-20682791 on Build GRCh38
rs547112275 Chr.1: 20682890-20682890 on Build GRCh38
rs1018639912 Chr.1: 20682972-20682972 on Build GRCh38
rs560938587 Chr.1: 20682827-20682827 on Build GRCh38
rs768156873 Chr.1: 20682802-20682802 on Build GRCh38
rs923710375 Chr.1: 20682877-20682877 on Build GRCh38
rs781316871 Chr.1: 20682893-20682893 on Build GRCh38
rs770910500 Chr.1: 20682832-20682832 on Build GRCh38
rs145619281 Chr.1: 20682881-20682881 on Build GRCh38
rs369600051 Chr.1: 20682635-20682635 on Build GRCh38
rs116203233 Chr.1: 20682701-20682701 on Build GRCh38
rs952198469 Chr.1: 20682634-20682634 on Build GRCh38
rs748646545 Chr.1: 20682655-20682655 on Build GRCh38
rs372429850 Chr.1: 20682892-20682892 on Build GRCh38
rs770339243 Chr.1: 20682665-20682665 on Build GRCh38
rs773946520 Chr.1: 20682833-20682833 on Build GRCh38
rs376783254 Chr.1: 20682811-20682811 on Build GRCh38
rs539628418 Chr.1: 20683017-20683017 on Build GRCh38
rs373433546 Chr.1: 20682921-20682921 on Build GRCh38
rs201236609 Chr.1: 20682638-20682638 on Build GRCh38
rs139817126 Chr.1: 20682718-20682718 on Build GRCh38
rs931893852 Chr.1: 20682948-20682948 on Build GRCh38
rs868810286 Chr.1: 20683015-20683015 on Build GRCh38
rs773263008 Chr.1: 20682762-20682762 on Build GRCh38
rs762784041 Chr.1: 20682829-20682829 on Build GRCh38
rs202095343 Chr.1: 20682620-20682620 on Build GRCh38
rs773507745 Chr.1: 20682683-20682683 on Build GRCh38
rs772406660 Chr.1: 20682938-20682938 on Build GRCh38
rs779789884 Chr.1: 20682629-20682630 on Build GRCh38
rs200472696 Chr.1: 20682735-20682735 on Build GRCh38
rs760831442 Chr.1: 20682917-20682917 on Build GRCh38
rs1012274334 Chr.1: 20682600-20682600 on Build GRCh38
rs549469387 Chr.1: 20682840-20682840 on Build GRCh38
rs765744507 Chr.1: 20682599-20682599 on Build GRCh38
rs150132550 Chr.1: 20682712-20682712 on Build GRCh38
rs771821302 Chr.1: 20682903-20682903 on Build GRCh38
rs148629128 Chr.1: 20682786-20682786 on Build GRCh38
rs753967263 Chr.1: 20682857-20682857 on Build GRCh38
rs767729742 Chr.1: 20682790-20682790 on Build GRCh38
rs147429225 Chr.1: 20682713-20682713 on Build GRCh38
rs1029415265 Chr.1: 20682939-20682939 on Build GRCh38
rs776193819 Chr.1: 20682595-20682595 on Build GRCh38
rs145153675 Chr.1: 20682690-20682690 on Build GRCh38
rs576701111 Chr.1: 20683026-20683026 on Build GRCh38
rs779016326 Chr.1: 20682785-20682785 on Build GRCh38
rs985367933 Chr.1: 20682967-20682967 on Build GRCh38
rs116367430 Chr.1: 20682793-20682793 on Build GRCh38
rs779090340 Chr.1: 20682799-20682799 on Build GRCh38
rs377150584 Chr.1: 20682906-20682906 on Build GRCh38
rs757638344 Chr.1: 20682859-20682859 on Build GRCh38
rs77555030 Chr.1: 20682596-20682596 on Build GRCh38
rs777878864 Chr.1: 20682796-20682796 on Build GRCh38
rs892767230 Chr.1: 20682972-20682972 on Build GRCh38
rs555989120 Chr.1: 20683044-20683044 on Build GRCh38
rs191271690 Chr.1: 20682719-20682719 on Build GRCh38
rs999725624 Chr.1: 20682769-20682769 on Build GRCh38
rs558038200 Chr.1: 20682708-20682708 on Build GRCh38
rs186978787 Chr.1: 20682808-20682808 on Build GRCh38
rs535180343 Chr.1: 20682642-20682642 on Build GRCh38
rs766080010 Chr.1: 20682784-20682784 on Build GRCh38
rs781115691 Chr.1: 20682729-20682729 on Build GRCh38

View Full Product Details