Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20684720-20685209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF17
Entrez Gene ID
57576
Gene Name
kinesin family member 17
Gene Aliases
KIF17B, KIF3X, KLP-2, OSM-3
UniGene
Hs.130411
Gene Chromosome Location
Chr.1: 20662294-20718017 on Build GRCh38
Transcripts
NM_001122819.2 NM_001287212.1 NM_020816.3 XM_005245950.3 XM_005245951.3 XM_011541837.1 XM_011541838.1 XM_011541839.2 XM_011541840.2 XM_011541841.2 XM_011541842.1 XM_011541843.1 XM_011541844.2 XM_011541845.2 XM_011541846.2 XM_011541847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377038 c.2164G>A p.G722S Chr.1: 20684876-20684876 on Build GRCh38
903371 c.2058G>A p.V686V Chr.1: 20684982-20684982 on Build GRCh38
903370 c.2075G>A p.G692D Chr.1: 20684965-20684965 on Build GRCh38
1338566 c.2186C>T p.T729I Chr.1: 20684854-20684854 on Build GRCh38
6387429 c.2213A>G p.Q738R Chr.1: 20684827-20684827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905251582 Chr.1: 20685066-20685066 on Build GRCh38
rs750049097 Chr.1: 20685045-20685045 on Build GRCh38
rs765357529 Chr.1: 20684828-20684828 on Build GRCh38
rs115463317 Chr.1: 20685174-20685174 on Build GRCh38
rs1040575820 Chr.1: 20685011-20685011 on Build GRCh38
rs779989905 Chr.1: 20684887-20684887 on Build GRCh38
rs61744456 Chr.1: 20685186-20685186 on Build GRCh38
rs753059794 Chr.1: 20684940-20684940 on Build GRCh38
rs774976007 Chr.1: 20684916-20684916 on Build GRCh38
rs886260380 Chr.1: 20684840-20684840 on Build GRCh38
rs141025234 Chr.1: 20685175-20685175 on Build GRCh38
rs3215776 Chr.1: 20684783-20684784 on Build GRCh38
rs202130525 Chr.1: 20684773-20684773 on Build GRCh38
rs1011287142 Chr.1: 20685041-20685041 on Build GRCh38
rs61778497 Chr.1: 20684784-20684784 on Build GRCh38
rs879010222 Chr.1: 20684924-20684924 on Build GRCh38
rs143758204 Chr.1: 20684908-20684908 on Build GRCh38
rs773357079 Chr.1: 20684909-20684909 on Build GRCh38
rs551278299 Chr.1: 20685040-20685040 on Build GRCh38
rs937549224 Chr.1: 20684969-20684969 on Build GRCh38
rs570421559 Chr.1: 20685130-20685130 on Build GRCh38
rs755465154 Chr.1: 20685115-20685115 on Build GRCh38
rs931342523 Chr.1: 20684994-20684994 on Build GRCh38
rs139742596 Chr.1: 20684947-20684947 on Build GRCh38
rs149255241 Chr.1: 20684855-20684855 on Build GRCh38
rs558760 Chr.1: 20685017-20685017 on Build GRCh38
rs756769150 Chr.1: 20684777-20684777 on Build GRCh38
rs771236748 Chr.1: 20685104-20685104 on Build GRCh38
rs931828884 Chr.1: 20685131-20685131 on Build GRCh38
rs74486081 Chr.1: 20685038-20685038 on Build GRCh38
rs569249224 Chr.1: 20684810-20684810 on Build GRCh38
rs138799650 Chr.1: 20684780-20684781 on Build GRCh38
rs763493826 Chr.1: 20684838-20684838 on Build GRCh38
rs761983478 Chr.1: 20684816-20684816 on Build GRCh38
rs376410495 Chr.1: 20684774-20684774 on Build GRCh38
rs944098772 Chr.1: 20685106-20685106 on Build GRCh38
rs763152449 Chr.1: 20684914-20684914 on Build GRCh38
rs771336204 Chr.1: 20685098-20685098 on Build GRCh38
rs1024320692 Chr.1: 20684752-20684752 on Build GRCh38
rs1010451008 Chr.1: 20684742-20684742 on Build GRCh38
rs188255115 Chr.1: 20685026-20685026 on Build GRCh38
rs12028811 Chr.1: 20684853-20684853 on Build GRCh38
rs774638949 Chr.1: 20685020-20685020 on Build GRCh38
rs2296224 Chr.1: 20685059-20685059 on Build GRCh38
rs572966268 Chr.1: 20685014-20685014 on Build GRCh38
rs764831126 Chr.1: 20684944-20684944 on Build GRCh38
rs1055989695 Chr.1: 20684970-20684970 on Build GRCh38
rs532054254 Chr.1: 20685013-20685013 on Build GRCh38
rs1036243043 Chr.1: 20685060-20685060 on Build GRCh38
rs372590207 Chr.1: 20685135-20685136 on Build GRCh38
rs373137026 Chr.1: 20684932-20684932 on Build GRCh38
rs946219421 Chr.1: 20685152-20685152 on Build GRCh38
rs748888738 Chr.1: 20684902-20684902 on Build GRCh38
rs768818031 Chr.1: 20684809-20684809 on Build GRCh38
rs963158639 Chr.1: 20685015-20685015 on Build GRCh38
rs1056167046 Chr.1: 20684797-20684797 on Build GRCh38
rs749515375 Chr.1: 20685095-20685095 on Build GRCh38
rs541953044 Chr.1: 20684941-20684941 on Build GRCh38
rs200435612 Chr.1: 20684880-20684880 on Build GRCh38
rs768471031 Chr.1: 20684897-20684897 on Build GRCh38
rs760160254 Chr.1: 20684918-20684918 on Build GRCh38
rs745389399 Chr.1: 20684879-20684879 on Build GRCh38
rs778161378 Chr.1: 20685006-20685006 on Build GRCh38
rs140931562 Chr.1: 20685003-20685003 on Build GRCh38
rs560975253 Chr.1: 20684999-20684999 on Build GRCh38
rs920202411 Chr.1: 20685077-20685077 on Build GRCh38
rs147992129 Chr.1: 20685129-20685130 on Build GRCh38
rs777749707 Chr.1: 20685089-20685089 on Build GRCh38
rs754931904 Chr.1: 20684996-20684996 on Build GRCh38
rs745799936 Chr.1: 20684783-20684783 on Build GRCh38
rs748696484 Chr.1: 20684776-20684776 on Build GRCh38
rs550732147 Chr.1: 20684802-20684802 on Build GRCh38
rs989888717 Chr.1: 20684905-20684905 on Build GRCh38
rs61750850 Chr.1: 20684862-20684862 on Build GRCh38
rs957040956 Chr.1: 20684890-20684890 on Build GRCh38
rs548165825 Chr.1: 20685129-20685129 on Build GRCh38
rs13375609 Chr.1: 20684837-20684837 on Build GRCh38
rs775201329 Chr.1: 20684794-20684794 on Build GRCh38
rs747000949 Chr.1: 20684895-20684895 on Build GRCh38
rs766994045 Chr.1: 20684844-20684844 on Build GRCh38
rs1052163915 Chr.1: 20685123-20685123 on Build GRCh38
rs1045918442 Chr.1: 20685173-20685173 on Build GRCh38
rs921670829 Chr.1: 20684746-20684746 on Build GRCh38
rs566583367 Chr.1: 20684846-20684846 on Build GRCh38
rs751087135 Chr.1: 20685056-20685056 on Build GRCh38
rs113457722 Chr.1: 20685075-20685075 on Build GRCh38
rs752485809 Chr.1: 20685072-20685072 on Build GRCh38
rs565624281 Chr.1: 20684779-20684779 on Build GRCh38
rs766940874 Chr.1: 20684915-20684915 on Build GRCh38
rs982778347 Chr.1: 20685100-20685100 on Build GRCh38
rs781019353 Chr.1: 20685067-20685067 on Build GRCh38
rs775884962 Chr.1: 20685030-20685030 on Build GRCh38
rs747504880 Chr.1: 20685103-20685103 on Build GRCh38
rs748392634 Chr.1: 20684907-20684907 on Build GRCh38
rs779582888 Chr.1: 20684980-20684980 on Build GRCh38
rs542213034 Chr.1: 20684981-20684981 on Build GRCh38
rs770015366 Chr.1: 20685004-20685004 on Build GRCh38
rs369388105 Chr.1: 20684964-20684964 on Build GRCh38
rs765864414 Chr.1: 20685051-20685051 on Build GRCh38
rs760073819 Chr.1: 20685025-20685025 on Build GRCh38
rs957071474 Chr.1: 20684753-20684753 on Build GRCh38
rs1055036974 Chr.1: 20684935-20684935 on Build GRCh38
rs112270148 Chr.1: 20684845-20684845 on Build GRCh38
rs878935663 Chr.1: 20685061-20685061 on Build GRCh38

View Full Product Details