Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20687531-20688024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF17
Entrez Gene ID
57576
Gene Name
kinesin family member 17
Gene Aliases
KIF17B, KIF3X, KLP-2, OSM-3
UniGene
Hs.130411
Gene Chromosome Location
Chr.1: 20662294-20718017 on Build GRCh38
Transcripts
NM_001122819.2 NM_001287212.1 NM_020816.3 XM_005245950.3 XM_005245951.3 XM_011541837.1 XM_011541838.1 XM_011541839.2 XM_011541840.2 XM_011541841.2 XM_011541842.1 XM_011541843.1 XM_011541844.2 XM_011541845.2 XM_011541846.2 XM_011541847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
321231 c.1387G>T p.V463F Chr.1: 20687939-20687939 on Build GRCh38
131050 c.1396G>T p.V466L Chr.1: 20687930-20687930 on Build GRCh38
6694848 c.1705G>A p.E569K Chr.1: 20687621-20687621 on Build GRCh38
6123876 c.1643C>G p.S548C Chr.1: 20687683-20687683 on Build GRCh38
365574 c.1460C>A p.P487Q Chr.1: 20687866-20687866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561922364 Chr.1: 20687595-20687595 on Build GRCh38
rs770052336 Chr.1: 20687745-20687745 on Build GRCh38
rs765880074 Chr.1: 20687661-20687661 on Build GRCh38
rs756582673 Chr.1: 20687902-20687902 on Build GRCh38
rs775659554 Chr.1: 20687642-20687642 on Build GRCh38
rs547092044 Chr.1: 20687694-20687694 on Build GRCh38
rs74956681 Chr.1: 20687822-20687822 on Build GRCh38
rs116376356 Chr.1: 20687582-20687582 on Build GRCh38
rs754694005 Chr.1: 20687897-20687897 on Build GRCh38
rs771849889 Chr.1: 20687706-20687706 on Build GRCh38
rs766885886 Chr.1: 20687675-20687675 on Build GRCh38
rs534971002 Chr.1: 20687866-20687866 on Build GRCh38
rs781103880 Chr.1: 20687901-20687901 on Build GRCh38
rs763493675 Chr.1: 20687853-20687853 on Build GRCh38
rs138561064 Chr.1: 20687757-20687757 on Build GRCh38
rs370414135 Chr.1: 20687608-20687608 on Build GRCh38
rs896454556 Chr.1: 20687558-20687558 on Build GRCh38
rs758293573 Chr.1: 20687887-20687887 on Build GRCh38
rs142146939 Chr.1: 20687691-20687691 on Build GRCh38
rs35006590 Chr.1: 20687969-20687969 on Build GRCh38
rs987566615 Chr.1: 20687635-20687635 on Build GRCh38
rs750347823 Chr.1: 20687799-20687799 on Build GRCh38
rs774735815 Chr.1: 20687865-20687865 on Build GRCh38
rs767729557 Chr.1: 20687562-20687562 on Build GRCh38
rs775880941 Chr.1: 20687971-20687971 on Build GRCh38
rs753338404 Chr.1: 20687775-20687775 on Build GRCh38
rs759729829 Chr.1: 20687947-20687948 on Build GRCh38
rs371293452 Chr.1: 20687921-20687921 on Build GRCh38
rs370470841 Chr.1: 20687973-20687973 on Build GRCh38
rs772233726 Chr.1: 20687636-20687636 on Build GRCh38
rs936575779 Chr.1: 20687941-20687941 on Build GRCh38
rs746813758 Chr.1: 20687810-20687810 on Build GRCh38
rs1046127328 Chr.1: 20687979-20687979 on Build GRCh38
rs528825687 Chr.1: 20687663-20687663 on Build GRCh38
rs772658029 Chr.1: 20687990-20687990 on Build GRCh38
rs201867180 Chr.1: 20687650-20687650 on Build GRCh38
rs41265077 Chr.1: 20687805-20687805 on Build GRCh38
rs540680062 Chr.1: 20687817-20687817 on Build GRCh38
rs200910771 Chr.1: 20687842-20687842 on Build GRCh38
rs947124230 Chr.1: 20687722-20687722 on Build GRCh38
rs764899515 Chr.1: 20687985-20687985 on Build GRCh38
rs750824605 Chr.1: 20687583-20687583 on Build GRCh38
rs753401086 Chr.1: 20687868-20687868 on Build GRCh38
rs754366304 Chr.1: 20687578-20687578 on Build GRCh38
rs200416385 Chr.1: 20687893-20687893 on Build GRCh38
rs368357803 Chr.1: 20687826-20687826 on Build GRCh38
rs761824033 Chr.1: 20687744-20687744 on Build GRCh38
rs986188668 Chr.1: 20687704-20687704 on Build GRCh38
rs751561906 Chr.1: 20687894-20687894 on Build GRCh38
rs754641038 Chr.1: 20687589-20687589 on Build GRCh38
rs139858579 Chr.1: 20687571-20687571 on Build GRCh38
rs768767723 Chr.1: 20687733-20687733 on Build GRCh38
rs772310290 Chr.1: 20687960-20687960 on Build GRCh38
rs776735699 Chr.1: 20687739-20687739 on Build GRCh38
rs200073708 Chr.1: 20687876-20687876 on Build GRCh38
rs745693652 Chr.1: 20687632-20687632 on Build GRCh38
rs953097695 Chr.1: 20687697-20687697 on Build GRCh38
rs749297675 Chr.1: 20687629-20687630 on Build GRCh38
rs752385893 Chr.1: 20687676-20687676 on Build GRCh38
rs771343541 Chr.1: 20687854-20687854 on Build GRCh38
rs773829153 Chr.1: 20687746-20687746 on Build GRCh38
rs760973084 Chr.1: 20687971-20687972 on Build GRCh38
rs747197585 Chr.1: 20687711-20687711 on Build GRCh38
rs367672816 Chr.1: 20687693-20687693 on Build GRCh38
rs34589742 Chr.1: 20687664-20687665 on Build GRCh38
rs756421987 Chr.1: 20687904-20687904 on Build GRCh38
rs762317411 Chr.1: 20687658-20687658 on Build GRCh38
rs949078938 Chr.1: 20687976-20687976 on Build GRCh38
rs578044364 Chr.1: 20687958-20687958 on Build GRCh38
rs369340351 Chr.1: 20687916-20687916 on Build GRCh38
rs756737847 Chr.1: 20687788-20687788 on Build GRCh38
rs770993075 Chr.1: 20687698-20687698 on Build GRCh38
rs760306635 Chr.1: 20687761-20687761 on Build GRCh38
rs372978316 Chr.1: 20687796-20687796 on Build GRCh38
rs749233289 Chr.1: 20687617-20687617 on Build GRCh38
rs191996405 Chr.1: 20687858-20687858 on Build GRCh38
rs770866526 Chr.1: 20687629-20687629 on Build GRCh38
rs200690761 Chr.1: 20687605-20687605 on Build GRCh38
rs761387455 Chr.1: 20687983-20687983 on Build GRCh38
rs775845659 Chr.1: 20687963-20687963 on Build GRCh38
rs149437507 Chr.1: 20687570-20687570 on Build GRCh38
rs769986016 Chr.1: 20687843-20687843 on Build GRCh38
rs553516160 Chr.1: 20687939-20687939 on Build GRCh38
rs545727010 Chr.1: 20687959-20687959 on Build GRCh38
rs75925314 Chr.1: 20687760-20687760 on Build GRCh38
rs755778084 Chr.1: 20687681-20687681 on Build GRCh38
rs773479871 Chr.1: 20687849-20687849 on Build GRCh38
rs373585154 Chr.1: 20687643-20687643 on Build GRCh38
rs777211794 Chr.1: 20687613-20687613 on Build GRCh38
rs763714504 Chr.1: 20687703-20687703 on Build GRCh38
rs115506787 Chr.1: 20687772-20687772 on Build GRCh38
rs752440383 Chr.1: 20687649-20687649 on Build GRCh38
rs747808678 Chr.1: 20687603-20687603 on Build GRCh38
rs140327262 Chr.1: 20687698-20687698 on Build GRCh38
rs115228567 Chr.1: 20687806-20687806 on Build GRCh38
rs764670643 Chr.1: 20687879-20687879 on Build GRCh38
rs147249030 Chr.1: 20687881-20687881 on Build GRCh38
rs1030234102 Chr.1: 20687930-20687930 on Build GRCh38
rs762663001 Chr.1: 20687669-20687669 on Build GRCh38
rs1039642164 Chr.1: 20687666-20687666 on Build GRCh38
rs369477297 Chr.1: 20687565-20687565 on Build GRCh38
rs757763220 Chr.1: 20687579-20687579 on Build GRCh38
rs759694974 Chr.1: 20687556-20687556 on Build GRCh38
rs115071410 Chr.1: 20687709-20687709 on Build GRCh38
rs768877673 Chr.1: 20687818-20687818 on Build GRCh38
rs764606354 Chr.1: 20687975-20687976 on Build GRCh38
rs867454014 Chr.1: 20687884-20687884 on Build GRCh38
rs576076610 Chr.1: 20687957-20687958 on Build GRCh38
rs774469446 Chr.1: 20687818-20687818 on Build GRCh38

View Full Product Details