Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40736197-40736710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223748 c.1378G>A p.D460N Chr.15: 40736576-40736576 on Build GRCh38
6767030 c.1373A>G p.Q458R Chr.15: 40736581-40736581 on Build GRCh38
1478083 c.1371C>T p.I457I Chr.15: 40736583-40736583 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187596143 Chr.15: 40736309-40736309 on Build GRCh38
rs760403872 Chr.15: 40736547-40736548 on Build GRCh38
rs144634421 Chr.15: 40736307-40736308 on Build GRCh38
rs983119241 Chr.15: 40736483-40736483 on Build GRCh38
rs748441704 Chr.15: 40736616-40736616 on Build GRCh38
rs572379647 Chr.15: 40736681-40736681 on Build GRCh38
rs938405755 Chr.15: 40736226-40736226 on Build GRCh38
rs1043986 Chr.15: 40736507-40736507 on Build GRCh38
rs754721788 Chr.15: 40736609-40736609 on Build GRCh38
rs1014062801 Chr.15: 40736223-40736223 on Build GRCh38
rs775310669 Chr.15: 40736523-40736524 on Build GRCh38
rs201041208 Chr.15: 40736536-40736536 on Build GRCh38
rs766545697 Chr.15: 40736644-40736661 on Build GRCh38
rs1022280971 Chr.15: 40736274-40736274 on Build GRCh38
rs979378134 Chr.15: 40736665-40736665 on Build GRCh38
rs569803789 Chr.15: 40736508-40736508 on Build GRCh38
rs547635953 Chr.15: 40736407-40736407 on Build GRCh38
rs749323676 Chr.15: 40736622-40736622 on Build GRCh38
rs368450150 Chr.15: 40736495-40736495 on Build GRCh38
rs767183788 Chr.15: 40736499-40736500 on Build GRCh38
rs993758330 Chr.15: 40736619-40736619 on Build GRCh38
rs758251233 Chr.15: 40736505-40736505 on Build GRCh38
rs774937138 Chr.15: 40736633-40736633 on Build GRCh38
rs372357790 Chr.15: 40736496-40736496 on Build GRCh38
rs570589991 Chr.15: 40736409-40736409 on Build GRCh38
rs910737926 Chr.15: 40736346-40736346 on Build GRCh38
rs929888158 Chr.15: 40736445-40736445 on Build GRCh38
rs199738362 Chr.15: 40736635-40736635 on Build GRCh38
rs879690822 Chr.15: 40736549-40736549 on Build GRCh38
rs768871035 Chr.15: 40736511-40736511 on Build GRCh38
rs928595935 Chr.15: 40736676-40736676 on Build GRCh38
rs915595419 Chr.15: 40736686-40736686 on Build GRCh38
rs1022019995 Chr.15: 40736412-40736413 on Build GRCh38
rs1055478920 Chr.15: 40736235-40736235 on Build GRCh38
rs757324308 Chr.15: 40736590-40736591 on Build GRCh38
rs761343703 Chr.15: 40736565-40736565 on Build GRCh38
rs748256673 Chr.15: 40736260-40736260 on Build GRCh38
rs144065093 Chr.15: 40736556-40736556 on Build GRCh38
rs1016683004 Chr.15: 40736534-40736534 on Build GRCh38
rs1044595665 Chr.15: 40736490-40736490 on Build GRCh38
rs879078635 Chr.15: 40736651-40736651 on Build GRCh38
rs115463166 Chr.15: 40736339-40736339 on Build GRCh38
rs545954630 Chr.15: 40736281-40736281 on Build GRCh38
rs763849510 Chr.15: 40736562-40736562 on Build GRCh38
rs369312512 Chr.15: 40736618-40736618 on Build GRCh38
rs926019306 Chr.15: 40736670-40736678 on Build GRCh38
rs373551332 Chr.15: 40736652-40736652 on Build GRCh38
rs370466451 Chr.15: 40736548-40736548 on Build GRCh38
rs372202647 Chr.15: 40736310-40736311 on Build GRCh38
rs748708581 Chr.15: 40736648-40736648 on Build GRCh38
rs766888053 Chr.15: 40736491-40736491 on Build GRCh38
rs781336679 Chr.15: 40736515-40736515 on Build GRCh38
rs938353150 Chr.15: 40736488-40736488 on Build GRCh38
rs964888613 Chr.15: 40736661-40736661 on Build GRCh38
rs547869415 Chr.15: 40736338-40736338 on Build GRCh38
rs894609766 Chr.15: 40736247-40736247 on Build GRCh38
rs769876369 Chr.15: 40736524-40736524 on Build GRCh38
rs751369428 Chr.15: 40736608-40736608 on Build GRCh38
rs1055467530 Chr.15: 40736489-40736489 on Build GRCh38
rs756565040 Chr.15: 40736261-40736261 on Build GRCh38
rs375316849 Chr.15: 40736521-40736521 on Build GRCh38
rs982681935 Chr.15: 40736673-40736673 on Build GRCh38
rs928270968 Chr.15: 40736486-40736486 on Build GRCh38
rs951298565 Chr.15: 40736670-40736670 on Build GRCh38
rs973965102 Chr.15: 40736390-40736390 on Build GRCh38
rs773132446 Chr.15: 40736530-40736530 on Build GRCh38
rs199724125 Chr.15: 40736489-40736490 on Build GRCh38
rs192555861 Chr.15: 40736684-40736684 on Build GRCh38
rs764854114 Chr.15: 40736573-40736573 on Build GRCh38
rs11856571 Chr.15: 40736494-40736494 on Build GRCh38
rs1030302470 Chr.15: 40736333-40736333 on Build GRCh38
rs527611866 Chr.15: 40736350-40736350 on Build GRCh38
rs536065749 Chr.15: 40736298-40736299 on Build GRCh38
rs779900252 Chr.15: 40736600-40736600 on Build GRCh38
rs763256768 Chr.15: 40736490-40736490 on Build GRCh38
rs919822200 Chr.15: 40736437-40736437 on Build GRCh38
rs753717352 Chr.15: 40736589-40736590 on Build GRCh38
rs202196709 Chr.15: 40736620-40736620 on Build GRCh38
rs562547269 Chr.15: 40736305-40736305 on Build GRCh38
rs1011786758 Chr.15: 40736257-40736257 on Build GRCh38
rs771565260 Chr.15: 40736623-40736623 on Build GRCh38
rs963862133 Chr.15: 40736379-40736379 on Build GRCh38
rs535514649 Chr.15: 40736574-40736574 on Build GRCh38
rs753479088 Chr.15: 40736564-40736564 on Build GRCh38
rs1018301008 Chr.15: 40736657-40736657 on Build GRCh38
rs978855880 Chr.15: 40736400-40736400 on Build GRCh38
rs758400366 Chr.15: 40736598-40736598 on Build GRCh38
rs999530319 Chr.15: 40736307-40736307 on Build GRCh38
rs764028784 Chr.15: 40736572-40736590 on Build GRCh38
rs751949477 Chr.15: 40736492-40736492 on Build GRCh38

View Full Product Details