Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40929615-40930113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL4
Entrez Gene ID
54567
Gene Name
delta like canonical Notch ligand 4
Gene Aliases
AOS6, hdelta2
UniGene
Hs.511076
Gene Chromosome Location
Chr.15: 40929333-40939060 on Build GRCh38
Transcripts
NM_019074.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6037747 c.225C>A p.T75T Chr.15: 40930005-40930005 on Build GRCh38
5447140 c.66+2T>C p.? Chr.15: 40929736-40929736 on Build GRCh38
3377714 c.219C>A p.P73P Chr.15: 40929999-40929999 on Build GRCh38
4590812 c.298C>G p.R100G Chr.15: 40930078-40930078 on Build GRCh38
4592097 c.311A>C p.Q104P Chr.15: 40930091-40930091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774844925 Chr.15: 40929937-40929937 on Build GRCh38
rs774055759 Chr.15: 40929757-40929757 on Build GRCh38
rs761616606 Chr.15: 40929957-40929957 on Build GRCh38
rs761902036 Chr.15: 40930071-40930072 on Build GRCh38
rs1025548919 Chr.15: 40930026-40930026 on Build GRCh38
rs897147422 Chr.15: 40929646-40929646 on Build GRCh38
rs776556363 Chr.15: 40930053-40930053 on Build GRCh38
rs927013257 Chr.15: 40929689-40929689 on Build GRCh38
rs778736053 Chr.15: 40929927-40929927 on Build GRCh38
rs571968428 Chr.15: 40929931-40929931 on Build GRCh38
rs199931075 Chr.15: 40929985-40929985 on Build GRCh38
rs893270432 Chr.15: 40929716-40929716 on Build GRCh38
rs766491570 Chr.15: 40930078-40930078 on Build GRCh38
rs370892158 Chr.15: 40929723-40929723 on Build GRCh38
rs754069384 Chr.15: 40929673-40929673 on Build GRCh38
rs775498713 Chr.15: 40929778-40929778 on Build GRCh38
rs763278905 Chr.15: 40930077-40930077 on Build GRCh38
rs768446870 Chr.15: 40929943-40929943 on Build GRCh38
rs748023227 Chr.15: 40930055-40930055 on Build GRCh38
rs202204117 Chr.15: 40929707-40929707 on Build GRCh38
rs372446567 Chr.15: 40929662-40929662 on Build GRCh38
rs778222660 Chr.15: 40930016-40930016 on Build GRCh38
rs774586523 Chr.15: 40930087-40930087 on Build GRCh38
rs767470734 Chr.15: 40929649-40929649 on Build GRCh38
rs752526172 Chr.15: 40929652-40929652 on Build GRCh38
rs769078168 Chr.15: 40929730-40929730 on Build GRCh38
rs757456948 Chr.15: 40929677-40929677 on Build GRCh38
rs370744960 Chr.15: 40929848-40929848 on Build GRCh38
rs763726702 Chr.15: 40929659-40929659 on Build GRCh38
rs765421830 Chr.15: 40929833-40929833 on Build GRCh38
rs937051098 Chr.15: 40929694-40929694 on Build GRCh38
rs751566523 Chr.15: 40929998-40929998 on Build GRCh38
rs779940325 Chr.15: 40929858-40929858 on Build GRCh38
rs776704947 Chr.15: 40929813-40929813 on Build GRCh38
rs917452596 Chr.15: 40929849-40929849 on Build GRCh38
rs750605020 Chr.15: 40929839-40929839 on Build GRCh38
rs759136630 Chr.15: 40929769-40929769 on Build GRCh38
rs768104229 Chr.15: 40930048-40930048 on Build GRCh38
rs772988101 Chr.15: 40930076-40930076 on Build GRCh38
rs750469977 Chr.15: 40929686-40929686 on Build GRCh38
rs3212277 Chr.15: 40929827-40929827 on Build GRCh38
rs772922527 Chr.15: 40929640-40929640 on Build GRCh38
rs374705119 Chr.15: 40929990-40929990 on Build GRCh38
rs769634656 Chr.15: 40930056-40930056 on Build GRCh38
rs759589482 Chr.15: 40929645-40929645 on Build GRCh38
rs115657051 Chr.15: 40929789-40929789 on Build GRCh38
rs949695484 Chr.15: 40929644-40929644 on Build GRCh38
rs889210508 Chr.15: 40929866-40929866 on Build GRCh38
rs779030792 Chr.15: 40929680-40929680 on Build GRCh38
rs766048930 Chr.15: 40929643-40929643 on Build GRCh38
rs746476747 Chr.15: 40930038-40930038 on Build GRCh38
rs770679832 Chr.15: 40929749-40929749 on Build GRCh38
rs755559750 Chr.15: 40930011-40930011 on Build GRCh38
rs760469031 Chr.15: 40929784-40929784 on Build GRCh38
rs776456937 Chr.15: 40929954-40929954 on Build GRCh38
rs770238390 Chr.15: 40929913-40929913 on Build GRCh38
rs753212508 Chr.15: 40930015-40930015 on Build GRCh38
rs367900130 Chr.15: 40929936-40929936 on Build GRCh38
rs760730986 Chr.15: 40929942-40929942 on Build GRCh38
rs374868641 Chr.15: 40929904-40929904 on Build GRCh38
rs763165309 Chr.15: 40929898-40929898 on Build GRCh38
rs200039900 Chr.15: 40929637-40929637 on Build GRCh38
rs368545705 Chr.15: 40930014-40930014 on Build GRCh38
rs1006227049 Chr.15: 40929874-40929874 on Build GRCh38
rs973096955 Chr.15: 40929989-40929989 on Build GRCh38
rs997452593 Chr.15: 40929797-40929797 on Build GRCh38
rs749019585 Chr.15: 40929741-40929741 on Build GRCh38
rs757981768 Chr.15: 40930030-40930030 on Build GRCh38
rs779655567 Chr.15: 40930035-40930035 on Build GRCh38
rs753625901 Chr.15: 40929809-40929809 on Build GRCh38
rs781400930 Chr.15: 40929731-40929731 on Build GRCh38
rs781721194 Chr.15: 40929901-40929901 on Build GRCh38
rs994110782 Chr.15: 40929665-40929665 on Build GRCh38
rs752167071 Chr.15: 40929879-40929879 on Build GRCh38
rs534328045 Chr.15: 40929980-40929980 on Build GRCh38
rs764006498 Chr.15: 40929806-40929806 on Build GRCh38
rs1010908427 Chr.15: 40929771-40929771 on Build GRCh38
rs765796727 Chr.15: 40929762-40929762 on Build GRCh38
rs372984064 Chr.15: 40930020-40930020 on Build GRCh38
rs760547707 Chr.15: 40929656-40929656 on Build GRCh38
rs780643868 Chr.15: 40929721-40929721 on Build GRCh38
rs748504505 Chr.15: 40929910-40929910 on Build GRCh38
rs901869248 Chr.15: 40929791-40929791 on Build GRCh38

View Full Product Details