Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40936138-40936631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL4
Entrez Gene ID
54567
Gene Name
delta like canonical Notch ligand 4
Gene Aliases
AOS6, hdelta2
UniGene
Hs.511076
Gene Chromosome Location
Chr.15: 40929333-40939060 on Build GRCh38
Transcripts
NM_019074.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464462 c.1335C>T p.A445A Chr.15: 40936322-40936322 on Build GRCh38
5383243 c.1381G>A p.G461R Chr.15: 40936368-40936368 on Build GRCh38
5383241 c.1341C>A p.N447K Chr.15: 40936328-40936328 on Build GRCh38
94134 c.1412C>T p.S471F Chr.15: 40936399-40936399 on Build GRCh38
6871449 c.1288C>A p.P430T Chr.15: 40936275-40936275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766236166 Chr.15: 40936403-40936403 on Build GRCh38
rs374162819 Chr.15: 40936308-40936308 on Build GRCh38
rs770793521 Chr.15: 40936517-40936517 on Build GRCh38
rs367841822 Chr.15: 40936419-40936419 on Build GRCh38
rs373338394 Chr.15: 40936257-40936257 on Build GRCh38
rs779764235 Chr.15: 40936223-40936224 on Build GRCh38
rs758847335 Chr.15: 40936561-40936561 on Build GRCh38
rs535125961 Chr.15: 40936258-40936258 on Build GRCh38
rs760234335 Chr.15: 40936285-40936285 on Build GRCh38
rs1025354705 Chr.15: 40936428-40936428 on Build GRCh38
rs776252375 Chr.15: 40936290-40936290 on Build GRCh38
rs774301166 Chr.15: 40936534-40936534 on Build GRCh38
rs753874647 Chr.15: 40936189-40936189 on Build GRCh38
rs758218516 Chr.15: 40936211-40936211 on Build GRCh38
rs761840661 Chr.15: 40936190-40936190 on Build GRCh38
rs370339149 Chr.15: 40936273-40936273 on Build GRCh38
rs990283280 Chr.15: 40936538-40936538 on Build GRCh38
rs771130564 Chr.15: 40936267-40936267 on Build GRCh38
rs760886659 Chr.15: 40936547-40936547 on Build GRCh38
rs549126795 Chr.15: 40936586-40936586 on Build GRCh38
rs752793079 Chr.15: 40936334-40936334 on Build GRCh38
rs762820678 Chr.15: 40936314-40936314 on Build GRCh38
rs747692838 Chr.15: 40936374-40936374 on Build GRCh38
rs942527179 Chr.15: 40936400-40936400 on Build GRCh38
rs780996581 Chr.15: 40936479-40936479 on Build GRCh38
rs751673521 Chr.15: 40936218-40936218 on Build GRCh38
rs368013738 Chr.15: 40936340-40936340 on Build GRCh38
rs371847465 Chr.15: 40936341-40936341 on Build GRCh38
rs545940366 Chr.15: 40936459-40936459 on Build GRCh38
rs750346321 Chr.15: 40936313-40936313 on Build GRCh38
rs903572679 Chr.15: 40936498-40936498 on Build GRCh38
rs762794387 Chr.15: 40936392-40936392 on Build GRCh38
rs376378103 Chr.15: 40936306-40936306 on Build GRCh38
rs750876689 Chr.15: 40936560-40936560 on Build GRCh38
rs758675187 Chr.15: 40936253-40936253 on Build GRCh38
rs749294716 Chr.15: 40936501-40936501 on Build GRCh38
rs759324100 Chr.15: 40936416-40936416 on Build GRCh38
rs974074895 Chr.15: 40936381-40936381 on Build GRCh38
rs777516760 Chr.15: 40936581-40936581 on Build GRCh38
rs369813246 Chr.15: 40936553-40936553 on Build GRCh38
rs757313247 Chr.15: 40936558-40936558 on Build GRCh38
rs755422423 Chr.15: 40936335-40936335 on Build GRCh38
rs775751614 Chr.15: 40936545-40936545 on Build GRCh38
rs890965449 Chr.15: 40936466-40936466 on Build GRCh38
rs747842030 Chr.15: 40936569-40936569 on Build GRCh38
rs779386236 Chr.15: 40936444-40936444 on Build GRCh38
rs980190964 Chr.15: 40936353-40936353 on Build GRCh38
rs377278209 Chr.15: 40936484-40936484 on Build GRCh38
rs757677186 Chr.15: 40936431-40936431 on Build GRCh38
rs750221835 Chr.15: 40936208-40936208 on Build GRCh38
rs773340803 Chr.15: 40936391-40936391 on Build GRCh38
rs376863553 Chr.15: 40936590-40936590 on Build GRCh38
rs796065345 Chr.15: 40936352-40936352 on Build GRCh38
rs374587964 Chr.15: 40936286-40936286 on Build GRCh38
rs751873412 Chr.15: 40936323-40936323 on Build GRCh38
rs575946294 Chr.15: 40936233-40936233 on Build GRCh38
rs780355579 Chr.15: 40936213-40936213 on Build GRCh38
rs778724796 Chr.15: 40936611-40936611 on Build GRCh38
rs768173236 Chr.15: 40936371-40936371 on Build GRCh38
rs751697404 Chr.15: 40936316-40936316 on Build GRCh38
rs769363130 Chr.15: 40936375-40936375 on Build GRCh38
rs770405453 Chr.15: 40936603-40936603 on Build GRCh38
rs779144185 Chr.15: 40936354-40936354 on Build GRCh38
rs781379364 Chr.15: 40936324-40936324 on Build GRCh38
rs765165782 Chr.15: 40936193-40936193 on Build GRCh38
rs372429686 Chr.15: 40936604-40936604 on Build GRCh38
rs184386888 Chr.15: 40936493-40936493 on Build GRCh38
rs767238730 Chr.15: 40936544-40936544 on Build GRCh38
rs550859837 Chr.15: 40936559-40936559 on Build GRCh38
rs778252260 Chr.15: 40936337-40936337 on Build GRCh38
rs780561241 Chr.15: 40936562-40936562 on Build GRCh38
rs371550881 Chr.15: 40936445-40936445 on Build GRCh38
rs540754224 Chr.15: 40936418-40936418 on Build GRCh38
rs962595544 Chr.15: 40936182-40936182 on Build GRCh38
rs889650035 Chr.15: 40936386-40936386 on Build GRCh38
rs942495528 Chr.15: 40936482-40936482 on Build GRCh38
rs572028305 Chr.15: 40936405-40936405 on Build GRCh38
rs764149583 Chr.15: 40936420-40936420 on Build GRCh38
rs370387440 Chr.15: 40936246-40936246 on Build GRCh38
rs376300590 Chr.15: 40936262-40936262 on Build GRCh38
rs750653634 Chr.15: 40936231-40936231 on Build GRCh38
rs544067517 Chr.15: 40936580-40936580 on Build GRCh38
rs374717233 Chr.15: 40936373-40936373 on Build GRCh38
rs201981567 Chr.15: 40936539-40936539 on Build GRCh38
rs1002658065 Chr.15: 40936478-40936478 on Build GRCh38
rs746718541 Chr.15: 40936359-40936359 on Build GRCh38
rs755141768 Chr.15: 40936220-40936220 on Build GRCh38
rs189323864 Chr.15: 40936548-40936548 on Build GRCh38
rs868764278 Chr.15: 40936464-40936464 on Build GRCh38
rs754439712 Chr.15: 40936430-40936430 on Build GRCh38
rs370793294 Chr.15: 40936307-40936307 on Build GRCh38

View Full Product Details