Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22586394-22586891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
UniGene
Hs.283613
Gene Chromosome Location
Chr.1: 22563508-22603594 on Build GRCh38
Transcripts
NM_001006943.1 XM_011540969.1 XM_011540970.1 XM_011540972.1 XM_011540973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751179 c.979+1G>A p.? Chr.1: 22586636-22586636 on Build GRCh38
3484891 c.852C>T p.A284A Chr.1: 22586508-22586508 on Build GRCh38
4572705 c.833T>G p.L278R Chr.1: 22586489-22586489 on Build GRCh38
6507057 c.888C>A p.H296Q Chr.1: 22586544-22586544 on Build GRCh38
4029298 c.897C>T p.S299S Chr.1: 22586553-22586553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755402558 Chr.1: 22586531-22586531 on Build GRCh38
rs79274528 Chr.1: 22586538-22586538 on Build GRCh38
rs768613874 Chr.1: 22586663-22586663 on Build GRCh38
rs367579366 Chr.1: 22586461-22586461 on Build GRCh38
rs759442934 Chr.1: 22586457-22586457 on Build GRCh38
rs550629807 Chr.1: 22586746-22586746 on Build GRCh38
rs753076516 Chr.1: 22586649-22586649 on Build GRCh38
rs756394651 Chr.1: 22586650-22586650 on Build GRCh38
rs1004281727 Chr.1: 22586862-22586862 on Build GRCh38
rs746880859 Chr.1: 22586566-22586566 on Build GRCh38
rs914998190 Chr.1: 22586741-22586741 on Build GRCh38
rs766504458 Chr.1: 22586619-22586619 on Build GRCh38
rs564697923 Chr.1: 22586684-22586684 on Build GRCh38
rs920964917 Chr.1: 22586706-22586706 on Build GRCh38
rs752059573 Chr.1: 22586530-22586530 on Build GRCh38
rs372203085 Chr.1: 22586553-22586553 on Build GRCh38
rs112043215 Chr.1: 22586801-22586801 on Build GRCh38
rs1002198723 Chr.1: 22586782-22586782 on Build GRCh38
rs938959891 Chr.1: 22586799-22586799 on Build GRCh38
rs929755154 Chr.1: 22586635-22586635 on Build GRCh38
rs386366467 Chr.1: 22586787-22586788 on Build GRCh38
rs1048846359 Chr.1: 22586840-22586840 on Build GRCh38
rs141028919 Chr.1: 22586554-22586554 on Build GRCh38
rs768504842 Chr.1: 22586590-22586590 on Build GRCh38
rs924763457 Chr.1: 22586786-22586786 on Build GRCh38
rs111410895 Chr.1: 22586740-22586740 on Build GRCh38
rs368255315 Chr.1: 22586519-22586519 on Build GRCh38
rs771968021 Chr.1: 22586459-22586459 on Build GRCh38
rs965314491 Chr.1: 22586699-22586699 on Build GRCh38
rs769097532 Chr.1: 22586437-22586437 on Build GRCh38
rs56656925 Chr.1: 22586618-22586618 on Build GRCh38
rs140379033 Chr.1: 22586523-22586523 on Build GRCh38
rs371407590 Chr.1: 22586463-22586463 on Build GRCh38
rs200249624 Chr.1: 22586464-22586464 on Build GRCh38
rs920922969 Chr.1: 22586522-22586522 on Build GRCh38
rs60727050 Chr.1: 22586788-22586789 on Build GRCh38
rs75174037 Chr.1: 22586783-22586784 on Build GRCh38
rs568070513 Chr.1: 22586780-22586781 on Build GRCh38
rs934218867 Chr.1: 22586745-22586745 on Build GRCh38
rs773901645 Chr.1: 22586456-22586456 on Build GRCh38
rs976685686 Chr.1: 22586702-22586702 on Build GRCh38
rs369018351 Chr.1: 22586565-22586565 on Build GRCh38
rs147152756 Chr.1: 22586501-22586501 on Build GRCh38
rs757907231 Chr.1: 22586661-22586661 on Build GRCh38
rs951250083 Chr.1: 22586854-22586854 on Build GRCh38
rs903455856 Chr.1: 22586561-22586561 on Build GRCh38
rs534782124 Chr.1: 22586858-22586858 on Build GRCh38
rs756914105 Chr.1: 22586547-22586547 on Build GRCh38
rs748907782 Chr.1: 22586444-22586444 on Build GRCh38
rs1015412946 Chr.1: 22586688-22586688 on Build GRCh38
rs982574954 Chr.1: 22586422-22586422 on Build GRCh38
rs781181106 Chr.1: 22586639-22586639 on Build GRCh38
rs142995895 Chr.1: 22586857-22586857 on Build GRCh38
rs553960671 Chr.1: 22586754-22586754 on Build GRCh38
rs376694821 Chr.1: 22586418-22586418 on Build GRCh38
rs183497128 Chr.1: 22586421-22586421 on Build GRCh38
rs777586088 Chr.1: 22586438-22586438 on Build GRCh38
rs1009659403 Chr.1: 22586759-22586759 on Build GRCh38
rs60166039 Chr.1: 22586784-22586785 on Build GRCh38
rs562760537 Chr.1: 22586747-22586747 on Build GRCh38
rs201432613 Chr.1: 22586616-22586616 on Build GRCh38
rs953737275 Chr.1: 22586709-22586709 on Build GRCh38
rs1022769220 Chr.1: 22586768-22586768 on Build GRCh38
rs1041884941 Chr.1: 22586781-22586781 on Build GRCh38
rs747343434 Chr.1: 22586434-22586434 on Build GRCh38
rs66713995 Chr.1: 22586781-22586782 on Build GRCh38
rs754009354 Chr.1: 22586480-22586480 on Build GRCh38
rs866503176 Chr.1: 22586780-22586780 on Build GRCh38
rs190763552 Chr.1: 22586642-22586642 on Build GRCh38
rs770417352 Chr.1: 22586455-22586455 on Build GRCh38
rs557217944 Chr.1: 22586539-22586539 on Build GRCh38
rs638524 Chr.1: 22586558-22586558 on Build GRCh38
rs1004063831 Chr.1: 22586671-22586671 on Build GRCh38
rs767823050 Chr.1: 22586643-22586643 on Build GRCh38
rs779533729 Chr.1: 22586662-22586662 on Build GRCh38
rs947558247 Chr.1: 22586557-22586557 on Build GRCh38
rs752182268 Chr.1: 22586636-22586636 on Build GRCh38
rs765465256 Chr.1: 22586615-22586615 on Build GRCh38
rs756103809 Chr.1: 22586678-22586679 on Build GRCh38
rs759009996 Chr.1: 22586509-22586509 on Build GRCh38
rs892607047 Chr.1: 22586757-22586757 on Build GRCh38
rs12034618 Chr.1: 22586417-22586417 on Build GRCh38
rs989510785 Chr.1: 22586739-22586739 on Build GRCh38
rs762088600 Chr.1: 22586492-22586492 on Build GRCh38
rs1024160414 Chr.1: 22586816-22586816 on Build GRCh38
rs638058 Chr.1: 22586660-22586660 on Build GRCh38
rs748046581 Chr.1: 22586666-22586677 on Build GRCh38
rs750491008 Chr.1: 22586503-22586503 on Build GRCh38
rs780612087 Chr.1: 22586433-22586433 on Build GRCh38
rs548354661 Chr.1: 22586805-22586805 on Build GRCh38
rs999895083 Chr.1: 22586749-22586749 on Build GRCh38
rs532109530 Chr.1: 22586732-22586732 on Build GRCh38
rs200025773 Chr.1: 22586416-22586416 on Build GRCh38
rs776327630 Chr.1: 22586683-22586683 on Build GRCh38
rs1012928868 Chr.1: 22586812-22586812 on Build GRCh38
rs748953848 Chr.1: 22586542-22586542 on Build GRCh38
rs776470892 Chr.1: 22586599-22586599 on Build GRCh38
rs908469425 Chr.1: 22586462-22586462 on Build GRCh38
rs761987517 Chr.1: 22586600-22586600 on Build GRCh38
rs778649812 Chr.1: 22586656-22586656 on Build GRCh38

View Full Product Details