Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22597207-22597707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
UniGene
Hs.283613
Gene Chromosome Location
Chr.1: 22563508-22603594 on Build GRCh38
Transcripts
XM_011540969.1 XM_011540970.1 XM_011540972.1 XM_011540973.1 XM_011540975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6438712 c.1798C>T p.P600S Chr.1: 22597344-22597344 on Build GRCh38
679202 c.1891G>C p.E631Q Chr.1: 22597437-22597437 on Build GRCh38
4474030 c.1890C>T p.I630I Chr.1: 22597436-22597436 on Build GRCh38
3997324 c.1834G>C p.E612Q Chr.1: 22597380-22597380 on Build GRCh38
6845699 c.1948T>C p.C650R Chr.1: 22597693-22597693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202243952 Chr.1: 22597344-22597344 on Build GRCh38
rs977512009 Chr.1: 22597235-22597235 on Build GRCh38
rs745792239 Chr.1: 22597355-22597355 on Build GRCh38
rs199556802 Chr.1: 22597519-22597519 on Build GRCh38
rs1017180106 Chr.1: 22597577-22597577 on Build GRCh38
rs777064385 Chr.1: 22597486-22597486 on Build GRCh38
rs370891889 Chr.1: 22597507-22597507 on Build GRCh38
rs776818950 Chr.1: 22597311-22597311 on Build GRCh38
rs745679594 Chr.1: 22597287-22597287 on Build GRCh38
rs141532503 Chr.1: 22597369-22597369 on Build GRCh38
rs202086660 Chr.1: 22597249-22597249 on Build GRCh38
rs774864430 Chr.1: 22597457-22597457 on Build GRCh38
rs374070758 Chr.1: 22597641-22597641 on Build GRCh38
rs775850552 Chr.1: 22597302-22597302 on Build GRCh38
rs551128370 Chr.1: 22597567-22597567 on Build GRCh38
rs549187828 Chr.1: 22597500-22597500 on Build GRCh38
rs748375751 Chr.1: 22597429-22597429 on Build GRCh38
rs770886999 Chr.1: 22597264-22597264 on Build GRCh38
rs763379733 Chr.1: 22597339-22597339 on Build GRCh38
rs200419319 Chr.1: 22597250-22597250 on Build GRCh38
rs935850330 Chr.1: 22597683-22597683 on Build GRCh38
rs924662093 Chr.1: 22597254-22597254 on Build GRCh38
rs61767315 Chr.1: 22597631-22597631 on Build GRCh38
rs201178361 Chr.1: 22597244-22597244 on Build GRCh38
rs371894783 Chr.1: 22597395-22597395 on Build GRCh38
rs779999041 Chr.1: 22597412-22597412 on Build GRCh38
rs766237180 Chr.1: 22597498-22597498 on Build GRCh38
rs766646724 Chr.1: 22597342-22597342 on Build GRCh38
rs934020406 Chr.1: 22597402-22597402 on Build GRCh38
rs1056950438 Chr.1: 22597298-22597298 on Build GRCh38
rs373638442 Chr.1: 22597467-22597467 on Build GRCh38
rs771365576 Chr.1: 22597437-22597437 on Build GRCh38
rs367961894 Chr.1: 22597660-22597660 on Build GRCh38
rs757872048 Chr.1: 22597493-22597493 on Build GRCh38
rs371869168 Chr.1: 22597667-22597667 on Build GRCh38
rs762519630 Chr.1: 22597492-22597493 on Build GRCh38
rs751447420 Chr.1: 22597499-22597499 on Build GRCh38
rs144329757 Chr.1: 22597366-22597366 on Build GRCh38
rs753369755 Chr.1: 22597348-22597348 on Build GRCh38
rs68055833 Chr.1: 22597240-22597250 on Build GRCh38
rs139415593 Chr.1: 22597379-22597379 on Build GRCh38
rs189030587 Chr.1: 22597394-22597394 on Build GRCh38
rs888338745 Chr.1: 22597609-22597609 on Build GRCh38
rs753426993 Chr.1: 22597405-22597405 on Build GRCh38
rs531005316 Chr.1: 22597409-22597409 on Build GRCh38
rs542382430 Chr.1: 22597255-22597255 on Build GRCh38
rs774791599 Chr.1: 22597625-22597625 on Build GRCh38
rs754139784 Chr.1: 22597600-22597600 on Build GRCh38
rs761435838 Chr.1: 22597469-22597469 on Build GRCh38
rs756831723 Chr.1: 22597408-22597408 on Build GRCh38
rs751234036 Chr.1: 22597522-22597522 on Build GRCh38
rs771963113 Chr.1: 22597365-22597365 on Build GRCh38
rs572862634 Chr.1: 22597349-22597349 on Build GRCh38
rs866840934 Chr.1: 22597398-22597398 on Build GRCh38
rs752445102 Chr.1: 22597343-22597343 on Build GRCh38
rs200457972 Chr.1: 22597504-22597504 on Build GRCh38
rs758235140 Chr.1: 22597410-22597410 on Build GRCh38
rs778773288 Chr.1: 22597269-22597269 on Build GRCh38
rs368501633 Chr.1: 22597391-22597391 on Build GRCh38
rs781568823 Chr.1: 22597428-22597428 on Build GRCh38
rs777366214 Chr.1: 22597346-22597346 on Build GRCh38
rs948522180 Chr.1: 22597564-22597564 on Build GRCh38
rs764692137 Chr.1: 22597306-22597306 on Build GRCh38
rs750902137 Chr.1: 22597328-22597328 on Build GRCh38
rs866178136 Chr.1: 22597424-22597424 on Build GRCh38
rs776940252 Chr.1: 22597378-22597378 on Build GRCh38
rs770977752 Chr.1: 22597524-22597524 on Build GRCh38
rs369284774 Chr.1: 22597436-22597436 on Build GRCh38
rs763353844 Chr.1: 22597383-22597383 on Build GRCh38
rs761940802 Chr.1: 22597312-22597312 on Build GRCh38
rs11582356 Chr.1: 22597292-22597292 on Build GRCh38
rs754761927 Chr.1: 22597419-22597419 on Build GRCh38
rs958400334 Chr.1: 22597307-22597307 on Build GRCh38
rs539580007 Chr.1: 22597658-22597658 on Build GRCh38
rs117379243 Chr.1: 22597530-22597530 on Build GRCh38
rs775803053 Chr.1: 22597639-22597639 on Build GRCh38
rs999765 Chr.1: 22597380-22597380 on Build GRCh38
rs746799254 Chr.1: 22597417-22597417 on Build GRCh38
rs1015313398 Chr.1: 22597375-22597375 on Build GRCh38
rs1004838786 Chr.1: 22597549-22597549 on Build GRCh38
rs765845041 Chr.1: 22597513-22597532 on Build GRCh38
rs550696020 Chr.1: 22597415-22597415 on Build GRCh38
rs754698715 Chr.1: 22597655-22597656 on Build GRCh38
rs536702821 Chr.1: 22597470-22597470 on Build GRCh38
rs753191289 Chr.1: 22597665-22597665 on Build GRCh38
rs1010081987 Chr.1: 22597229-22597229 on Build GRCh38
rs569332823 Chr.1: 22597606-22597606 on Build GRCh38
rs376201963 Chr.1: 22597431-22597431 on Build GRCh38
rs772859945 Chr.1: 22597655-22597655 on Build GRCh38
rs781119439 Chr.1: 22597501-22597501 on Build GRCh38
rs567750244 Chr.1: 22597458-22597458 on Build GRCh38
rs769455811 Chr.1: 22597654-22597654 on Build GRCh38
rs766808717 Chr.1: 22597388-22597388 on Build GRCh38
rs768788331 Chr.1: 22597373-22597373 on Build GRCh38
rs533721836 Chr.1: 22597678-22597678 on Build GRCh38
rs749928938 Chr.1: 22597488-22597488 on Build GRCh38
rs760797198 Chr.1: 22597303-22597303 on Build GRCh38
rs765867803 Chr.1: 22597314-22597314 on Build GRCh38
rs201461269 Chr.1: 22597627-22597627 on Build GRCh38
rs374784294 Chr.1: 22597671-22597671 on Build GRCh38
rs374307788 Chr.1: 22597509-22597509 on Build GRCh38

View Full Product Details