Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22862901-22863400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB2
Entrez Gene ID
2048
Gene Name
EPH receptor B2
Gene Aliases
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5
UniGene
Hs.380705
Gene Chromosome Location
Chr.1: 22710770-22921500 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3 NR_036214.1

Gene Symbol
MIR4253
Entrez Gene ID
100422914
Gene Name
microRNA 4253
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 22863159-22863226 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3 NR_036214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735325 c.890G>A p.C297Y Chr.1: 22863133-22863133 on Build GRCh38
280833 c.853G>A p.A285T Chr.1: 22863078-22863078 on Build GRCh38
131752 c.952G>A p.D318N Chr.1: 22863177-22863177 on Build GRCh38
1205512 c.917G>A p.R306H Chr.1: 22863142-22863142 on Build GRCh38
212329 c.848A>G p.D283G Chr.1: 22863073-22863073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6679387 Chr.1: 22863010-22863010 on Build GRCh38
rs772406361 Chr.1: 22863066-22863066 on Build GRCh38
rs764555559 Chr.1: 22863104-22863104 on Build GRCh38
rs760756497 Chr.1: 22863228-22863228 on Build GRCh38
rs372620832 Chr.1: 22863044-22863044 on Build GRCh38
rs778958746 Chr.1: 22863272-22863272 on Build GRCh38
rs943753954 Chr.1: 22863203-22863203 on Build GRCh38
rs1003339055 Chr.1: 22863282-22863282 on Build GRCh38
rs770933694 Chr.1: 22863183-22863183 on Build GRCh38
rs188204221 Chr.1: 22863281-22863281 on Build GRCh38
rs142113032 Chr.1: 22863072-22863072 on Build GRCh38
rs755673470 Chr.1: 22863173-22863173 on Build GRCh38
rs755653550 Chr.1: 22863259-22863259 on Build GRCh38
rs754403229 Chr.1: 22863105-22863105 on Build GRCh38
rs773180070 Chr.1: 22863019-22863019 on Build GRCh38
rs765058138 Chr.1: 22863249-22863262 on Build GRCh38
rs768207785 Chr.1: 22862992-22862993 on Build GRCh38
rs958979997 Chr.1: 22863348-22863348 on Build GRCh38
rs778909458 Chr.1: 22863202-22863202 on Build GRCh38
rs72871539 Chr.1: 22863277-22863277 on Build GRCh38
rs1018573765 Chr.1: 22862927-22862927 on Build GRCh38
rs971304652 Chr.1: 22863006-22863006 on Build GRCh38
rs754191330 Chr.1: 22862987-22862987 on Build GRCh38
rs139990297 Chr.1: 22863360-22863360 on Build GRCh38
rs753387416 Chr.1: 22863056-22863056 on Build GRCh38
rs765604107 Chr.1: 22863130-22863130 on Build GRCh38
rs761593747 Chr.1: 22863027-22863028 on Build GRCh38
rs757592405 Chr.1: 22862995-22862995 on Build GRCh38
rs561796506 Chr.1: 22862942-22862942 on Build GRCh38
rs749130502 Chr.1: 22863025-22863025 on Build GRCh38
rs1033240161 Chr.1: 22862961-22862961 on Build GRCh38
rs772459312 Chr.1: 22863214-22863214 on Build GRCh38
rs768789658 Chr.1: 22863229-22863229 on Build GRCh38
rs150415344 Chr.1: 22863174-22863174 on Build GRCh38
rs183122628 Chr.1: 22863261-22863261 on Build GRCh38
rs111569109 Chr.1: 22863222-22863222 on Build GRCh38
rs147727992 Chr.1: 22863106-22863106 on Build GRCh38
rs1029184974 Chr.1: 22862972-22862972 on Build GRCh38
rs757326702 Chr.1: 22863266-22863266 on Build GRCh38
rs776403456 Chr.1: 22862995-22863014 on Build GRCh38
rs745650276 Chr.1: 22863210-22863210 on Build GRCh38
rs754570824 Chr.1: 22863011-22863011 on Build GRCh38
rs76487715 Chr.1: 22862976-22862976 on Build GRCh38
rs760775609 Chr.1: 22863092-22863092 on Build GRCh38
rs749240821 Chr.1: 22863178-22863178 on Build GRCh38
rs981468344 Chr.1: 22863012-22863012 on Build GRCh38
rs1035711185 Chr.1: 22863303-22863303 on Build GRCh38
rs12083517 Chr.1: 22863149-22863149 on Build GRCh38
rs529363271 Chr.1: 22862949-22862949 on Build GRCh38
rs763283802 Chr.1: 22863245-22863245 on Build GRCh38
rs902104256 Chr.1: 22862962-22862962 on Build GRCh38
rs75882465 Chr.1: 22863155-22863155 on Build GRCh38
rs750809200 Chr.1: 22863236-22863236 on Build GRCh38
rs931080937 Chr.1: 22862933-22862933 on Build GRCh38
rs765729152 Chr.1: 22863237-22863237 on Build GRCh38
rs780738757 Chr.1: 22863017-22863017 on Build GRCh38
rs990305495 Chr.1: 22862978-22862978 on Build GRCh38
rs752199354 Chr.1: 22863170-22863170 on Build GRCh38
rs981394239 Chr.1: 22863327-22863327 on Build GRCh38
rs1020413098 Chr.1: 22862983-22862983 on Build GRCh38
rs747689340 Chr.1: 22863018-22863018 on Build GRCh38
rs762369215 Chr.1: 22863234-22863234 on Build GRCh38
rs893384312 Chr.1: 22863258-22863258 on Build GRCh38
rs1046820691 Chr.1: 22862935-22862935 on Build GRCh38
rs567401714 Chr.1: 22863254-22863254 on Build GRCh38
rs776518179 Chr.1: 22863231-22863231 on Build GRCh38
rs940755901 Chr.1: 22863014-22863014 on Build GRCh38
rs889333439 Chr.1: 22863113-22863113 on Build GRCh38
rs752424394 Chr.1: 22863255-22863255 on Build GRCh38
rs35882952 Chr.1: 22863060-22863060 on Build GRCh38
rs1007935799 Chr.1: 22862941-22862941 on Build GRCh38
rs886852806 Chr.1: 22862940-22862940 on Build GRCh38
rs779211309 Chr.1: 22863000-22863000 on Build GRCh38
rs374426753 Chr.1: 22863242-22863242 on Build GRCh38
rs745703494 Chr.1: 22863276-22863276 on Build GRCh38
rs919822792 Chr.1: 22863077-22863077 on Build GRCh38
rs754693296 Chr.1: 22863145-22863145 on Build GRCh38
rs996803373 Chr.1: 22862971-22862971 on Build GRCh38
rs750776222 Chr.1: 22863142-22863142 on Build GRCh38

View Full Product Details