Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23194473-23194999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR1D
Entrez Gene ID
3352
Gene Name
5-hydroxytryptamine receptor 1D
Gene Aliases
5-HT1D, HT1DA, HTR1DA, HTRL, RDC4
UniGene
Hs.121482
Gene Chromosome Location
Chr.1: 23191895-23194729 on Build GRCh38
Transcripts
NM_000864.4

SNP Detail

SNP ID Chromosome Location
rs1006899621 Chr.1: 23194884-23194884 on Build GRCh38
rs780536282 Chr.1: 23194839-23194839 on Build GRCh38
rs1019661032 Chr.1: 23194890-23194890 on Build GRCh38
rs372820396 Chr.1: 23194841-23194841 on Build GRCh38
rs945286565 Chr.1: 23194669-23194669 on Build GRCh38
rs8192525 Chr.1: 23194815-23194815 on Build GRCh38
rs781413253 Chr.1: 23194578-23194578 on Build GRCh38
rs770262130 Chr.1: 23194731-23194731 on Build GRCh38
rs985459651 Chr.1: 23194588-23194588 on Build GRCh38
rs559638889 Chr.1: 23194595-23194595 on Build GRCh38
rs552109429 Chr.1: 23194568-23194568 on Build GRCh38
rs676643 Chr.1: 23194847-23194847 on Build GRCh38
rs986091953 Chr.1: 23194618-23194618 on Build GRCh38
rs937944687 Chr.1: 23194916-23194916 on Build GRCh38
rs556902780 Chr.1: 23194540-23194540 on Build GRCh38
rs550286344 Chr.1: 23194758-23194758 on Build GRCh38
rs112636810 Chr.1: 23194549-23194549 on Build GRCh38
rs1031160904 Chr.1: 23194925-23194925 on Build GRCh38
rs921046556 Chr.1: 23194628-23194628 on Build GRCh38
rs746187148 Chr.1: 23194715-23194715 on Build GRCh38
rs184182529 Chr.1: 23194602-23194602 on Build GRCh38
rs539748828 Chr.1: 23194881-23194881 on Build GRCh38
rs975524892 Chr.1: 23194835-23194835 on Build GRCh38
rs3753254 Chr.1: 23194658-23194658 on Build GRCh38
rs950934421 Chr.1: 23194558-23194558 on Build GRCh38
rs945226875 Chr.1: 23194868-23194868 on Build GRCh38
rs965502654 Chr.1: 23194648-23194648 on Build GRCh38
rs955076464 Chr.1: 23194840-23194840 on Build GRCh38
rs954101354 Chr.1: 23194571-23194571 on Build GRCh38
rs561594612 Chr.1: 23194533-23194533 on Build GRCh38
rs1044053868 Chr.1: 23194670-23194670 on Build GRCh38
rs896533561 Chr.1: 23194770-23194770 on Build GRCh38
rs748546260 Chr.1: 23194704-23194704 on Build GRCh38
rs568442904 Chr.1: 23194768-23194768 on Build GRCh38
rs1027995928 Chr.1: 23194836-23194836 on Build GRCh38
rs573774253 Chr.1: 23194696-23194696 on Build GRCh38
rs543135795 Chr.1: 23194515-23194515 on Build GRCh38
rs200435399 Chr.1: 23194701-23194701 on Build GRCh38
rs962713657 Chr.1: 23194601-23194601 on Build GRCh38
rs962395223 Chr.1: 23194833-23194833 on Build GRCh38
rs541080273 Chr.1: 23194971-23194971 on Build GRCh38
rs561867964 Chr.1: 23194955-23194955 on Build GRCh38
rs1006451894 Chr.1: 23194820-23194820 on Build GRCh38
rs8192526 Chr.1: 23194697-23194697 on Build GRCh38
rs1016956264 Chr.1: 23194827-23194827 on Build GRCh38
rs141946275 Chr.1: 23194900-23194900 on Build GRCh38
rs924143638 Chr.1: 23194664-23194664 on Build GRCh38
rs769991654 Chr.1: 23194724-23194724 on Build GRCh38
rs571866066 Chr.1: 23194596-23194596 on Build GRCh38
rs569885069 Chr.1: 23194647-23194647 on Build GRCh38
rs757460496 Chr.1: 23194526-23194526 on Build GRCh38
rs1046707248 Chr.1: 23194739-23194739 on Build GRCh38
rs573644291 Chr.1: 23194969-23194969 on Build GRCh38
rs187526429 Chr.1: 23194862-23194862 on Build GRCh38
rs957755163 Chr.1: 23194668-23194668 on Build GRCh38
rs191507594 Chr.1: 23194896-23194896 on Build GRCh38
rs1050878029 Chr.1: 23194655-23194655 on Build GRCh38
rs547073878 Chr.1: 23194562-23194562 on Build GRCh38
rs933811644 Chr.1: 23194634-23194634 on Build GRCh38
rs138508272 Chr.1: 23194772-23194772 on Build GRCh38
rs150656473 Chr.1: 23194922-23194922 on Build GRCh38
rs868441325 Chr.1: 23194814-23194814 on Build GRCh38

View Full Product Details