Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72351046-72351553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1937344 c.672+10C>T p.? Chr.15: 72351123-72351123 on Build GRCh38
6482499 c.672+1G>T p.? Chr.15: 72351132-72351132 on Build GRCh38
71052 c.650C>G p.T217S Chr.15: 72351155-72351155 on Build GRCh38
4920550 c.611A>T p.H204L Chr.15: 72351194-72351194 on Build GRCh38
5940520 c.625C>T p.P209S Chr.15: 72351180-72351180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910391278 Chr.15: 72351518-72351518 on Build GRCh38
rs543071358 Chr.15: 72351189-72351189 on Build GRCh38
rs763649208 Chr.15: 72351184-72351184 on Build GRCh38
rs117160567 Chr.15: 72351103-72351103 on Build GRCh38
rs767318868 Chr.15: 72351202-72351202 on Build GRCh38
rs887554856 Chr.15: 72351297-72351297 on Build GRCh38
rs914762492 Chr.15: 72351397-72351397 on Build GRCh38
rs1800429 Chr.15: 72351207-72351207 on Build GRCh38
rs954154784 Chr.15: 72351420-72351420 on Build GRCh38
rs948767589 Chr.15: 72351453-72351453 on Build GRCh38
rs1002712424 Chr.15: 72351198-72351198 on Build GRCh38
rs773862194 Chr.15: 72351092-72351092 on Build GRCh38
rs1031884101 Chr.15: 72351321-72351321 on Build GRCh38
rs932977864 Chr.15: 72351266-72351267 on Build GRCh38
rs914562942 Chr.15: 72351334-72351334 on Build GRCh38
rs757141411 Chr.15: 72351107-72351107 on Build GRCh38
rs749422284 Chr.15: 72351137-72351137 on Build GRCh38
rs747427536 Chr.15: 72351248-72351248 on Build GRCh38
rs964427838 Chr.15: 72351230-72351230 on Build GRCh38
rs758259959 Chr.15: 72351300-72351300 on Build GRCh38
rs768223779 Chr.15: 72351277-72351277 on Build GRCh38
rs576601675 Chr.15: 72351253-72351253 on Build GRCh38
rs1012652444 Chr.15: 72351301-72351301 on Build GRCh38
rs762482250 Chr.15: 72351102-72351102 on Build GRCh38
rs768751360 Chr.15: 72351141-72351141 on Build GRCh38
rs752356903 Chr.15: 72351100-72351100 on Build GRCh38
rs971662804 Chr.15: 72351078-72351078 on Build GRCh38
rs762581234 Chr.15: 72351178-72351178 on Build GRCh38
rs121907961 Chr.15: 72351176-72351176 on Build GRCh38
rs185429231 Chr.15: 72351235-72351235 on Build GRCh38
rs759062735 Chr.15: 72351093-72351093 on Build GRCh38
rs121907974 Chr.15: 72351173-72351173 on Build GRCh38
rs572232325 Chr.15: 72351130-72351130 on Build GRCh38
rs387906311 Chr.15: 72351132-72351132 on Build GRCh38
rs772581459 Chr.15: 72351148-72351148 on Build GRCh38
rs985479676 Chr.15: 72351425-72351425 on Build GRCh38
rs121907973 Chr.15: 72351215-72351215 on Build GRCh38
rs387906310 Chr.15: 72351231-72351231 on Build GRCh38
rs139948663 Chr.15: 72351163-72351163 on Build GRCh38
rs112614306 Chr.15: 72351223-72351223 on Build GRCh38
rs761297135 Chr.15: 72351155-72351155 on Build GRCh38
rs764863374 Chr.15: 72351094-72351094 on Build GRCh38
rs774842392 Chr.15: 72351169-72351169 on Build GRCh38
rs773807075 Chr.15: 72351154-72351154 on Build GRCh38
rs371212159 Chr.15: 72351278-72351278 on Build GRCh38
rs956489053 Chr.15: 72351329-72351329 on Build GRCh38
rs761536872 Chr.15: 72351200-72351200 on Build GRCh38
rs867369722 Chr.15: 72351274-72351274 on Build GRCh38
rs990642979 Chr.15: 72351121-72351121 on Build GRCh38
rs121907976 Chr.15: 72351194-72351194 on Build GRCh38
rs1057519461 Chr.15: 72351168-72351168 on Build GRCh38
rs748427632 Chr.15: 72351228-72351228 on Build GRCh38
rs971528702 Chr.15: 72351307-72351307 on Build GRCh38
rs771550714 Chr.15: 72351250-72351250 on Build GRCh38
rs990203696 Chr.15: 72351332-72351332 on Build GRCh38
rs756010672 Chr.15: 72351208-72351208 on Build GRCh38
rs778253689 Chr.15: 72351244-72351244 on Build GRCh38
rs770313582 Chr.15: 72351204-72351204 on Build GRCh38
rs773615096 Chr.15: 72351302-72351302 on Build GRCh38
rs778947516 Chr.15: 72351224-72351224 on Build GRCh38
rs753862880 Chr.15: 72351218-72351218 on Build GRCh38
rs780134593 Chr.15: 72351133-72351133 on Build GRCh38
rs367731228 Chr.15: 72351265-72351265 on Build GRCh38
rs551398130 Chr.15: 72351354-72351354 on Build GRCh38
rs781039189 Chr.15: 72351110-72351110 on Build GRCh38
rs189451248 Chr.15: 72351288-72351288 on Build GRCh38
rs944501929 Chr.15: 72351527-72351527 on Build GRCh38
rs532813213 Chr.15: 72351320-72351320 on Build GRCh38
rs751298761 Chr.15: 72351106-72351106 on Build GRCh38

View Full Product Details