Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74591070-74591570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
UniGene
Hs.647642
Gene Chromosome Location
Chr.15: 74541177-74598131 on Build GRCh38
Transcripts
NM_001307939.1 NM_006465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882710 c.1063C>T p.L355F Chr.15: 74591332-74591332 on Build GRCh38
4904737 c.1161G>A p.R387R Chr.15: 74591430-74591430 on Build GRCh38
1374541 c.940G>A p.E314K Chr.15: 74591209-74591209 on Build GRCh38
3782910 c.979C>T p.R327W Chr.15: 74591248-74591248 on Build GRCh38
5028002 c.884A>T p.Y295F Chr.15: 74591153-74591153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761179835 Chr.15: 74591102-74591102 on Build GRCh38
rs146340798 Chr.15: 74591274-74591274 on Build GRCh38
rs767023228 Chr.15: 74591104-74591104 on Build GRCh38
rs375626686 Chr.15: 74591146-74591146 on Build GRCh38
rs780287913 Chr.15: 74591433-74591433 on Build GRCh38
rs906162279 Chr.15: 74591175-74591175 on Build GRCh38
rs751710611 Chr.15: 74591400-74591400 on Build GRCh38
rs142326639 Chr.15: 74591244-74591244 on Build GRCh38
rs772723765 Chr.15: 74591319-74591319 on Build GRCh38
rs562060939 Chr.15: 74591299-74591300 on Build GRCh38
rs750462911 Chr.15: 74591534-74591534 on Build GRCh38
rs148734605 Chr.15: 74591184-74591184 on Build GRCh38
rs778669730 Chr.15: 74591174-74591174 on Build GRCh38
rs756306380 Chr.15: 74591543-74591543 on Build GRCh38
rs1053752911 Chr.15: 74591252-74591252 on Build GRCh38
rs776310617 Chr.15: 74591343-74591343 on Build GRCh38
rs770473860 Chr.15: 74591483-74591483 on Build GRCh38
rs765046727 Chr.15: 74591282-74591282 on Build GRCh38
rs370677559 Chr.15: 74591125-74591125 on Build GRCh38
rs758444015 Chr.15: 74591193-74591193 on Build GRCh38
rs760132413 Chr.15: 74591326-74591326 on Build GRCh38
rs751620803 Chr.15: 74591294-74591294 on Build GRCh38
rs916337272 Chr.15: 74591150-74591150 on Build GRCh38
rs904604091 Chr.15: 74591214-74591214 on Build GRCh38
rs779356206 Chr.15: 74591471-74591471 on Build GRCh38
rs766230840 Chr.15: 74591140-74591140 on Build GRCh38
rs760995922 Chr.15: 74591305-74591310 on Build GRCh38
rs1027859512 Chr.15: 74591333-74591333 on Build GRCh38
rs763968566 Chr.15: 74591524-74591524 on Build GRCh38
rs772776093 Chr.15: 74591251-74591251 on Build GRCh38
rs375732452 Chr.15: 74591351-74591351 on Build GRCh38
rs757566083 Chr.15: 74591295-74591295 on Build GRCh38
rs557153895 Chr.15: 74591336-74591336 on Build GRCh38
rs759163368 Chr.15: 74591350-74591350 on Build GRCh38
rs769141557 Chr.15: 74591318-74591318 on Build GRCh38
rs756355290 Chr.15: 74591419-74591419 on Build GRCh38
rs771270878 Chr.15: 74591217-74591217 on Build GRCh38
rs534871429 Chr.15: 74591219-74591219 on Build GRCh38
rs759290574 Chr.15: 74591278-74591278 on Build GRCh38
rs947780326 Chr.15: 74591155-74591155 on Build GRCh38
rs752622302 Chr.15: 74591283-74591283 on Build GRCh38
rs755465428 Chr.15: 74591448-74591448 on Build GRCh38
rs200028946 Chr.15: 74591385-74591385 on Build GRCh38
rs746101449 Chr.15: 74591547-74591547 on Build GRCh38
rs746175963 Chr.15: 74591226-74591226 on Build GRCh38
rs199931788 Chr.15: 74591331-74591331 on Build GRCh38
rs770445187 Chr.15: 74591236-74591236 on Build GRCh38
rs764032923 Chr.15: 74591397-74591397 on Build GRCh38
rs4886601 Chr.15: 74591510-74591510 on Build GRCh38
rs1041083511 Chr.15: 74591132-74591132 on Build GRCh38
rs201697711 Chr.15: 74591129-74591129 on Build GRCh38
rs772758926 Chr.15: 74591115-74591115 on Build GRCh38
rs911803549 Chr.15: 74591095-74591095 on Build GRCh38
rs760326719 Chr.15: 74591256-74591256 on Build GRCh38
rs539888569 Chr.15: 74591499-74591499 on Build GRCh38
rs186328752 Chr.15: 74591528-74591528 on Build GRCh38
rs749828698 Chr.15: 74591316-74591316 on Build GRCh38
rs775820241 Chr.15: 74591238-74591238 on Build GRCh38
rs1015112327 Chr.15: 74591305-74591305 on Build GRCh38
rs553291894 Chr.15: 74591302-74591302 on Build GRCh38
rs781515627 Chr.15: 74591301-74591301 on Build GRCh38
rs202245107 Chr.15: 74591443-74591443 on Build GRCh38
rs753566479 Chr.15: 74591145-74591145 on Build GRCh38
rs994507193 Chr.15: 74591317-74591317 on Build GRCh38
rs748626511 Chr.15: 74591475-74591475 on Build GRCh38
rs1046107127 Chr.15: 74591168-74591168 on Build GRCh38
rs541636797 Chr.15: 74591313-74591313 on Build GRCh38
rs770104893 Chr.15: 74591310-74591310 on Build GRCh38
rs371312027 Chr.15: 74591527-74591527 on Build GRCh38
rs545906143 Chr.15: 74591404-74591404 on Build GRCh38
rs769419249 Chr.15: 74591521-74591521 on Build GRCh38
rs965325542 Chr.15: 74591324-74591324 on Build GRCh38
rs764489563 Chr.15: 74591449-74591450 on Build GRCh38
rs765960523 Chr.15: 74591262-74591262 on Build GRCh38
rs777779519 Chr.15: 74591293-74591293 on Build GRCh38
rs563699167 Chr.15: 74591412-74591412 on Build GRCh38
rs986380329 Chr.15: 74591427-74591427 on Build GRCh38
rs376538273 Chr.15: 74591507-74591507 on Build GRCh38
rs200797671 Chr.15: 74591208-74591208 on Build GRCh38
rs777883936 Chr.15: 74591200-74591200 on Build GRCh38
rs745571368 Chr.15: 74591514-74591514 on Build GRCh38
rs754925309 Chr.15: 74591147-74591147 on Build GRCh38
rs916407496 Chr.15: 74591489-74591489 on Build GRCh38
rs775159484 Chr.15: 74591522-74591522 on Build GRCh38
rs758339906 Chr.15: 74591287-74591287 on Build GRCh38
rs775347667 Chr.15: 74591359-74591359 on Build GRCh38
rs139916482 Chr.15: 74591311-74591311 on Build GRCh38
rs372840399 Chr.15: 74591246-74591246 on Build GRCh38
rs762464589 Chr.15: 74591405-74591405 on Build GRCh38
rs200892963 Chr.15: 74591526-74591526 on Build GRCh38

View Full Product Details