Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74595779-74596274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
UniGene
Hs.647642
Gene Chromosome Location
Chr.15: 74541177-74598131 on Build GRCh38
Transcripts
NM_001307939.1 NM_006465.3

SNP Detail

SNP ID Chromosome Location
rs759734130 Chr.15: 74596169-74596169 on Build GRCh38
rs1016196621 Chr.15: 74595892-74595892 on Build GRCh38
rs916202150 Chr.15: 74595803-74595803 on Build GRCh38
rs74781061 Chr.15: 74595855-74595855 on Build GRCh38
rs1056851861 Chr.15: 74596091-74596091 on Build GRCh38
rs954263482 Chr.15: 74596180-74596180 on Build GRCh38
rs758595748 Chr.15: 74595812-74595812 on Build GRCh38
rs865876008 Chr.15: 74596137-74596137 on Build GRCh38
rs535459205 Chr.15: 74595952-74595952 on Build GRCh38
rs752755059 Chr.15: 74595809-74595809 on Build GRCh38
rs764249995 Chr.15: 74595816-74595816 on Build GRCh38
rs570715531 Chr.15: 74595909-74595909 on Build GRCh38
rs181342887 Chr.15: 74595904-74595904 on Build GRCh38
rs896464409 Chr.15: 74596102-74596102 on Build GRCh38
rs577101775 Chr.15: 74596240-74596240 on Build GRCh38
rs934387490 Chr.15: 74595950-74595950 on Build GRCh38
rs905782650 Chr.15: 74596079-74596079 on Build GRCh38
rs914402274 Chr.15: 74596032-74596032 on Build GRCh38
rs948382215 Chr.15: 74596120-74596120 on Build GRCh38
rs912342531 Chr.15: 74595923-74595923 on Build GRCh38
rs574924021 Chr.15: 74595930-74595930 on Build GRCh38
rs1002192401 Chr.15: 74596081-74596081 on Build GRCh38
rs534754882 Chr.15: 74595910-74595910 on Build GRCh38
rs576228268 Chr.15: 74596104-74596104 on Build GRCh38
rs35873352 Chr.15: 74596023-74596024 on Build GRCh38
rs1052848989 Chr.15: 74596030-74596030 on Build GRCh38
rs1044291274 Chr.15: 74596078-74596078 on Build GRCh38
rs912231737 Chr.15: 74596253-74596253 on Build GRCh38
rs553041523 Chr.15: 74595913-74595913 on Build GRCh38
rs3803462 Chr.15: 74596154-74596154 on Build GRCh38
rs557341273 Chr.15: 74596066-74596066 on Build GRCh38
rs963207616 Chr.15: 74596139-74596139 on Build GRCh38
rs765169379 Chr.15: 74595805-74595805 on Build GRCh38
rs868412271 Chr.15: 74596074-74596074 on Build GRCh38
rs543742795 Chr.15: 74596105-74596105 on Build GRCh38
rs1044344 Chr.15: 74596173-74596173 on Build GRCh38

View Full Product Details