Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78588177-78588718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965389 c.409G>A p.D137N Chr.15: 78588419-78588419 on Build GRCh38
7273638 c.322T>A p.L108I Chr.15: 78588332-78588332 on Build GRCh38
965388 c.356A>G p.K119R Chr.15: 78588366-78588366 on Build GRCh38
4425831 c.364C>T p.R122C Chr.15: 78588374-78588374 on Build GRCh38
6392833 c.380C>A p.S127Y Chr.15: 78588390-78588390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527893898 Chr.15: 78588443-78588443 on Build GRCh38
rs778473235 Chr.15: 78588364-78588364 on Build GRCh38
rs2229961 Chr.15: 78588410-78588410 on Build GRCh38
rs761851654 Chr.15: 78588215-78588216 on Build GRCh38
rs998335290 Chr.15: 78588612-78588612 on Build GRCh38
rs1029864122 Chr.15: 78588649-78588649 on Build GRCh38
rs1057377353 Chr.15: 78588428-78588428 on Build GRCh38
rs910242839 Chr.15: 78588325-78588325 on Build GRCh38
rs888482677 Chr.15: 78588546-78588546 on Build GRCh38
rs143659162 Chr.15: 78588375-78588375 on Build GRCh38
rs201138933 Chr.15: 78588448-78588448 on Build GRCh38
rs565382338 Chr.15: 78588241-78588242 on Build GRCh38
rs1015640931 Chr.15: 78588604-78588604 on Build GRCh38
rs954258446 Chr.15: 78588681-78588681 on Build GRCh38
rs139319291 Chr.15: 78588567-78588567 on Build GRCh38
rs932902057 Chr.15: 78588246-78588246 on Build GRCh38
rs769607456 Chr.15: 78588430-78588430 on Build GRCh38
rs936678735 Chr.15: 78588668-78588668 on Build GRCh38
rs949435276 Chr.15: 78588224-78588224 on Build GRCh38
rs199882720 Chr.15: 78588301-78588301 on Build GRCh38
rs56023835 Chr.15: 78588351-78588351 on Build GRCh38
rs552142003 Chr.15: 78588398-78588398 on Build GRCh38
rs201451815 Chr.15: 78588388-78588388 on Build GRCh38
rs766890816 Chr.15: 78588588-78588589 on Build GRCh38
rs774209829 Chr.15: 78588273-78588276 on Build GRCh38
rs995002486 Chr.15: 78588536-78588536 on Build GRCh38
rs1005974645 Chr.15: 78588591-78588591 on Build GRCh38
rs376394314 Chr.15: 78588354-78588354 on Build GRCh38
rs777337280 Chr.15: 78588392-78588392 on Build GRCh38
rs895726087 Chr.15: 78588480-78588480 on Build GRCh38
rs926623440 Chr.15: 78588272-78588272 on Build GRCh38
rs745811241 Chr.15: 78588313-78588317 on Build GRCh38
rs776748138 Chr.15: 78588389-78588392 on Build GRCh38
rs12898919 Chr.15: 78588235-78588235 on Build GRCh38
rs963036435 Chr.15: 78588489-78588489 on Build GRCh38
rs759579004 Chr.15: 78588211-78588218 on Build GRCh38
rs770803080 Chr.15: 78588409-78588409 on Build GRCh38
rs947116397 Chr.15: 78588347-78588347 on Build GRCh38
rs371640436 Chr.15: 78588349-78588349 on Build GRCh38
rs72650654 Chr.15: 78588217-78588218 on Build GRCh38
rs985747017 Chr.15: 78588692-78588692 on Build GRCh38
rs768655482 Chr.15: 78588388-78588389 on Build GRCh38
rs114945237 Chr.15: 78588545-78588545 on Build GRCh38
rs200616965 Chr.15: 78588424-78588424 on Build GRCh38
rs1007447626 Chr.15: 78588458-78588462 on Build GRCh38
rs902937153 Chr.15: 78588401-78588401 on Build GRCh38
rs754710874 Chr.15: 78588360-78588360 on Build GRCh38
rs755250024 Chr.15: 78588544-78588544 on Build GRCh38
rs147445612 Chr.15: 78588446-78588446 on Build GRCh38
rs752072464 Chr.15: 78588294-78588294 on Build GRCh38
rs981009607 Chr.15: 78588645-78588645 on Build GRCh38
rs1017706500 Chr.15: 78588483-78588483 on Build GRCh38
rs201192025 Chr.15: 78588308-78588308 on Build GRCh38
rs764085194 Chr.15: 78588475-78588475 on Build GRCh38
rs892335479 Chr.15: 78588381-78588381 on Build GRCh38
rs74913206 Chr.15: 78588374-78588374 on Build GRCh38
rs192556881 Chr.15: 78588647-78588647 on Build GRCh38
rs567766745 Chr.15: 78588520-78588520 on Build GRCh38
rs201778201 Chr.15: 78588324-78588324 on Build GRCh38
rs977328308 Chr.15: 78588213-78588213 on Build GRCh38
rs754054274 Chr.15: 78588502-78588502 on Build GRCh38
rs571599378 Chr.15: 78588609-78588609 on Build GRCh38
rs982928064 Chr.15: 78588557-78588558 on Build GRCh38

View Full Product Details