Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78593130-78593639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
UniGene
Hs.89605
Gene Chromosome Location
Chr.15: 78593052-78621295 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135727 c.1303T>C p.F435L Chr.15: 78593149-78593149 on Build GRCh38
6258205 c.1342G>T p.V448F Chr.15: 78593188-78593188 on Build GRCh38
2011172 c.1391G>A p.G464E Chr.15: 78593237-78593237 on Build GRCh38
2011171 c.1285C>T p.R429W Chr.15: 78593131-78593131 on Build GRCh38
5433056 c.1287G>C p.R429R Chr.15: 78593133-78593133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746886634 Chr.15: 78593203-78593203 on Build GRCh38
rs72650655 Chr.15: 78593564-78593566 on Build GRCh38
rs202044161 Chr.15: 78593252-78593252 on Build GRCh38
rs766695728 Chr.15: 78593236-78593236 on Build GRCh38
rs142884378 Chr.15: 78593530-78593530 on Build GRCh38
rs199796617 Chr.15: 78593259-78593259 on Build GRCh38
rs77773727 Chr.15: 78593171-78593171 on Build GRCh38
rs976095415 Chr.15: 78593291-78593291 on Build GRCh38
rs71528529 Chr.15: 78593564-78593567 on Build GRCh38
rs200875928 Chr.15: 78593537-78593537 on Build GRCh38
rs770782949 Chr.15: 78593224-78593224 on Build GRCh38
rs199932554 Chr.15: 78593382-78593382 on Build GRCh38
rs759148420 Chr.15: 78593297-78593297 on Build GRCh38
rs201437321 Chr.15: 78593506-78593506 on Build GRCh38
rs73463098 Chr.15: 78593570-78593570 on Build GRCh38
rs763265856 Chr.15: 78593194-78593197 on Build GRCh38
rs751038368 Chr.15: 78593237-78593237 on Build GRCh38
rs200339983 Chr.15: 78593487-78593487 on Build GRCh38
rs763412847 Chr.15: 78593156-78593156 on Build GRCh38
rs775537854 Chr.15: 78593251-78593251 on Build GRCh38
rs757859948 Chr.15: 78593189-78593189 on Build GRCh38
rs202136309 Chr.15: 78593157-78593157 on Build GRCh38
rs3830438 Chr.15: 78593277-78593278 on Build GRCh38
rs201714288 Chr.15: 78593327-78593327 on Build GRCh38
rs113264952 Chr.15: 78593388-78593388 on Build GRCh38
rs777707103 Chr.15: 78593199-78593199 on Build GRCh38
rs200501248 Chr.15: 78593559-78593559 on Build GRCh38
rs199559512 Chr.15: 78593172-78593172 on Build GRCh38
rs201184771 Chr.15: 78593350-78593350 on Build GRCh38
rs959165006 Chr.15: 78593198-78593198 on Build GRCh38
rs367546696 Chr.15: 78593158-78593158 on Build GRCh38
rs770027654 Chr.15: 78593243-78593243 on Build GRCh38
rs199881121 Chr.15: 78593335-78593335 on Build GRCh38
rs754643899 Chr.15: 78593165-78593165 on Build GRCh38
rs202175642 Chr.15: 78593385-78593385 on Build GRCh38
rs762405020 Chr.15: 78593277-78593277 on Build GRCh38
rs76450138 Chr.15: 78593405-78593405 on Build GRCh38
rs148376112 Chr.15: 78593170-78593170 on Build GRCh38
rs201177341 Chr.15: 78593229-78593229 on Build GRCh38
rs907484405 Chr.15: 78593434-78593434 on Build GRCh38
rs72534078 Chr.15: 78593276-78593277 on Build GRCh38
rs746129356 Chr.15: 78593234-78593234 on Build GRCh38
rs76071148 Chr.15: 78593232-78593232 on Build GRCh38
rs144182521 Chr.15: 78593273-78593274 on Build GRCh38
rs200921958 Chr.15: 78593383-78593383 on Build GRCh38
rs201239275 Chr.15: 78593456-78593456 on Build GRCh38
rs965993931 Chr.15: 78593278-78593278 on Build GRCh38
rs532896027 Chr.15: 78593183-78593183 on Build GRCh38
rs200919591 Chr.15: 78593273-78593273 on Build GRCh38
rs911264884 Chr.15: 78593424-78593424 on Build GRCh38
rs768051249 Chr.15: 78593284-78593284 on Build GRCh38
rs188919226 Chr.15: 78593575-78593575 on Build GRCh38
rs199612620 Chr.15: 78593545-78593545 on Build GRCh38
rs200384194 Chr.15: 78593174-78593174 on Build GRCh38
rs200454752 Chr.15: 78593246-78593246 on Build GRCh38
rs201776618 Chr.15: 78593359-78593359 on Build GRCh38
rs200202015 Chr.15: 78593450-78593450 on Build GRCh38
rs768799141 Chr.15: 78593261-78593261 on Build GRCh38
rs202018459 Chr.15: 78593447-78593447 on Build GRCh38
rs969050221 Chr.15: 78593610-78593611 on Build GRCh38
rs934245761 Chr.15: 78593414-78593414 on Build GRCh38
rs200561529 Chr.15: 78593411-78593411 on Build GRCh38
rs200357731 Chr.15: 78593164-78593164 on Build GRCh38
rs200133737 Chr.15: 78593609-78593609 on Build GRCh38
rs199714231 Chr.15: 78593477-78593477 on Build GRCh38
rs1023503238 Chr.15: 78593588-78593588 on Build GRCh38
rs372674046 Chr.15: 78593293-78593293 on Build GRCh38
rs541787390 Chr.15: 78593562-78593565 on Build GRCh38
rs201284306 Chr.15: 78593428-78593428 on Build GRCh38
rs200911665 Chr.15: 78593272-78593272 on Build GRCh38
rs946226625 Chr.15: 78593420-78593420 on Build GRCh38
rs202001164 Chr.15: 78593542-78593542 on Build GRCh38
rs773626999 Chr.15: 78593154-78593158 on Build GRCh38

View Full Product Details