Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80895791-80896303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEMIP
Entrez Gene ID
57214
Gene Name
cell migration inducing hyaluronan binding protein
Gene Aliases
CCSP1, HYBID, KIAA1199, TMEM2L
UniGene
Hs.459088
Gene Chromosome Location
Chr.15: 80779343-80951662 on Build GRCh38
Transcripts
NM_001293298.1 NM_001293304.1 NM_018689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7401465 c.1382C>T p.S461F Chr.15: 80896031-80896031 on Build GRCh38
4057256 c.1301A>C p.K434T Chr.15: 80895950-80895950 on Build GRCh38
6693775 c.1388delC p.N465fs*18 Chr.15: 80896037-80896037 on Build GRCh38
6588234 c.1233C>T p.L411L Chr.15: 80895882-80895882 on Build GRCh38
7373582 c.1231C>G p.L411V Chr.15: 80895880-80895880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762795463 Chr.15: 80896002-80896002 on Build GRCh38
rs752126339 Chr.15: 80896174-80896174 on Build GRCh38
rs749751432 Chr.15: 80895957-80895957 on Build GRCh38
rs752502897 Chr.15: 80896000-80896000 on Build GRCh38
rs144349326 Chr.15: 80895880-80895880 on Build GRCh38
rs369462089 Chr.15: 80896035-80896035 on Build GRCh38
rs758338201 Chr.15: 80895902-80895902 on Build GRCh38
rs752594779 Chr.15: 80895899-80895899 on Build GRCh38
rs77213153 Chr.15: 80895873-80895873 on Build GRCh38
rs1046322325 Chr.15: 80895980-80895980 on Build GRCh38
rs748715482 Chr.15: 80896063-80896063 on Build GRCh38
rs368277774 Chr.15: 80896036-80896036 on Build GRCh38
rs779906770 Chr.15: 80896049-80896049 on Build GRCh38
rs1010696910 Chr.15: 80896072-80896072 on Build GRCh38
rs114610043 Chr.15: 80896120-80896120 on Build GRCh38
rs113605876 Chr.15: 80896094-80896094 on Build GRCh38
rs772285746 Chr.15: 80896085-80896085 on Build GRCh38
rs200780187 Chr.15: 80896080-80896080 on Build GRCh38
rs377272717 Chr.15: 80896008-80896008 on Build GRCh38
rs767626693 Chr.15: 80895865-80895865 on Build GRCh38
rs1007931331 Chr.15: 80896142-80896142 on Build GRCh38
rs772245449 Chr.15: 80895992-80895992 on Build GRCh38
rs377685748 Chr.15: 80896081-80896081 on Build GRCh38
rs760921708 Chr.15: 80895996-80895996 on Build GRCh38
rs1040161725 Chr.15: 80895892-80895892 on Build GRCh38
rs1009874857 Chr.15: 80895962-80895962 on Build GRCh38
rs893409590 Chr.15: 80896075-80896076 on Build GRCh38
rs1027293607 Chr.15: 80896197-80896197 on Build GRCh38
rs764304374 Chr.15: 80896134-80896134 on Build GRCh38
rs771130058 Chr.15: 80896082-80896082 on Build GRCh38
rs866912635 Chr.15: 80896114-80896114 on Build GRCh38
rs766876498 Chr.15: 80896265-80896265 on Build GRCh38
rs142807800 Chr.15: 80896180-80896181 on Build GRCh38
rs750700998 Chr.15: 80895878-80895878 on Build GRCh38
rs761814989 Chr.15: 80896112-80896112 on Build GRCh38
rs1019359988 Chr.15: 80896153-80896153 on Build GRCh38
rs765043598 Chr.15: 80895888-80895888 on Build GRCh38
rs756830371 Chr.15: 80895925-80895925 on Build GRCh38
rs772009474 Chr.15: 80896067-80896067 on Build GRCh38
rs774304121 Chr.15: 80895852-80895852 on Build GRCh38
rs749935718 Chr.15: 80896267-80896267 on Build GRCh38
rs952674650 Chr.15: 80896233-80896233 on Build GRCh38
rs756671909 Chr.15: 80896014-80896014 on Build GRCh38
rs776906604 Chr.15: 80896092-80896092 on Build GRCh38
rs79026618 Chr.15: 80896062-80896062 on Build GRCh38
rs372964060 Chr.15: 80896100-80896100 on Build GRCh38
rs774987227 Chr.15: 80895986-80895986 on Build GRCh38
rs751020491 Chr.15: 80895923-80895923 on Build GRCh38
rs769269343 Chr.15: 80895974-80895974 on Build GRCh38
rs139948991 Chr.15: 80896006-80896006 on Build GRCh38
rs556306589 Chr.15: 80896143-80896143 on Build GRCh38
rs146608881 Chr.15: 80895938-80895938 on Build GRCh38
rs140156627 Chr.15: 80896146-80896146 on Build GRCh38
rs982988371 Chr.15: 80896254-80896254 on Build GRCh38
rs79390951 Chr.15: 80895943-80895943 on Build GRCh38
rs755749927 Chr.15: 80896273-80896273 on Build GRCh38
rs763585241 Chr.15: 80895828-80895828 on Build GRCh38
rs764946392 Chr.15: 80895999-80895999 on Build GRCh38
rs745555869 Chr.15: 80895936-80895936 on Build GRCh38
rs777207203 Chr.15: 80895921-80895921 on Build GRCh38
rs538920031 Chr.15: 80896240-80896240 on Build GRCh38
rs201833164 Chr.15: 80896059-80896059 on Build GRCh38
rs144229359 Chr.15: 80895851-80895851 on Build GRCh38
rs780684620 Chr.15: 80895928-80895928 on Build GRCh38
rs931122111 Chr.15: 80895825-80895825 on Build GRCh38
rs761770716 Chr.15: 80895988-80895988 on Build GRCh38
rs762009144 Chr.15: 80895859-80895859 on Build GRCh38
rs773262039 Chr.15: 80895993-80895993 on Build GRCh38

View Full Product Details