Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 16157584-16158089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC6
Entrez Gene ID
368
Gene Name
ATP binding cassette subfamily C member 6
Gene Aliases
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7
UniGene
Hs.442182
Gene Chromosome Location
Chr.16: 16149565-16223616 on Build GRCh38
Transcripts
NM_001171.5 XM_011522479.2 XM_011522480.1 XM_011522481.2 XM_017023212.1 XM_017023213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850264 c.3870C>G p.H1290Q Chr.16: 16157675-16157675 on Build GRCh38
6398140 c.3768G>T p.Q1256H Chr.16: 16157777-16157777 on Build GRCh38
5367878 c.3774_3775insC p.W1259fs*19 Chr.16: 16157770-16157771 on Build GRCh38
5790316 c.3882+3G>A p.? Chr.16: 16157660-16157660 on Build GRCh38
5384306 c.3850G>A p.G1284S Chr.16: 16157695-16157695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995125133 Chr.16: 16157725-16157725 on Build GRCh38
rs760560012 Chr.16: 16157815-16157815 on Build GRCh38
rs77913024 Chr.16: 16157709-16157709 on Build GRCh38
rs934129472 Chr.16: 16157842-16157842 on Build GRCh38
rs750893189 Chr.16: 16157699-16157699 on Build GRCh38
rs1021522024 Chr.16: 16158012-16158012 on Build GRCh38
rs63750625 Chr.16: 16157668-16157668 on Build GRCh38
rs376851894 Chr.16: 16157663-16157663 on Build GRCh38
rs74315128 Chr.16: 16157677-16157724 on Build GRCh38
rs776435670 Chr.16: 16157816-16157816 on Build GRCh38
rs760376992 Chr.16: 16157758-16157758 on Build GRCh38
rs61294695 Chr.16: 16157708-16157708 on Build GRCh38
rs58694313 Chr.16: 16157674-16157674 on Build GRCh38
rs759798432 Chr.16: 16157719-16157719 on Build GRCh38
rs72664235 Chr.16: 16157663-16157665 on Build GRCh38
rs200242428 Chr.16: 16157805-16157805 on Build GRCh38
rs72664221 Chr.16: 16157770-16157771 on Build GRCh38
rs63750209 Chr.16: 16157727-16157727 on Build GRCh38
rs111423295 Chr.16: 16157994-16157994 on Build GRCh38
rs72664234 Chr.16: 16157747-16157747 on Build GRCh38
rs964712550 Chr.16: 16157762-16157762 on Build GRCh38
rs1024138831 Chr.16: 16158018-16158018 on Build GRCh38
rs886342586 Chr.16: 16157991-16157991 on Build GRCh38
rs753457535 Chr.16: 16157689-16157689 on Build GRCh38
rs369871412 Chr.16: 16157673-16157673 on Build GRCh38
rs755354542 Chr.16: 16157845-16157845 on Build GRCh38
rs974849688 Chr.16: 16157772-16157772 on Build GRCh38
rs761870089 Chr.16: 16157632-16157632 on Build GRCh38
rs571019342 Chr.16: 16157996-16157996 on Build GRCh38
rs63750273 Chr.16: 16157810-16157810 on Build GRCh38
rs1026155579 Chr.16: 16157741-16157741 on Build GRCh38
rs902718185 Chr.16: 16158021-16158021 on Build GRCh38
rs56982924 Chr.16: 16157693-16157693 on Build GRCh38
rs763831330 Chr.16: 16157676-16157676 on Build GRCh38
rs375647381 Chr.16: 16157785-16157785 on Build GRCh38
rs756665345 Chr.16: 16158044-16158044 on Build GRCh38
rs370514701 Chr.16: 16157819-16157819 on Build GRCh38
rs937700322 Chr.16: 16157901-16157901 on Build GRCh38
rs749327665 Chr.16: 16157807-16157807 on Build GRCh38
rs559905923 Chr.16: 16158023-16158023 on Build GRCh38
rs762345158 Chr.16: 16157830-16157830 on Build GRCh38
rs772783837 Chr.16: 16157842-16157843 on Build GRCh38
rs576328904 Chr.16: 16157775-16157775 on Build GRCh38
rs767815589 Chr.16: 16157848-16157848 on Build GRCh38
rs72653749 Chr.16: 16157722-16157722 on Build GRCh38
rs149151337 Chr.16: 16157767-16157767 on Build GRCh38
rs754916743 Chr.16: 16157853-16157853 on Build GRCh38
rs2238471 Chr.16: 16158042-16158042 on Build GRCh38
rs896191742 Chr.16: 16157941-16157941 on Build GRCh38
rs200010958 Chr.16: 16157721-16157721 on Build GRCh38
rs2238472 Chr.16: 16157742-16157742 on Build GRCh38
rs753800428 Chr.16: 16157894-16157894 on Build GRCh38
rs184301360 Chr.16: 16157912-16157912 on Build GRCh38
rs1053328151 Chr.16: 16157605-16157605 on Build GRCh38
rs148326870 Chr.16: 16157794-16157794 on Build GRCh38
rs527851693 Chr.16: 16157874-16157874 on Build GRCh38
rs377115141 Chr.16: 16157833-16157833 on Build GRCh38
rs72664233 Chr.16: 16157770-16157770 on Build GRCh38
rs779603756 Chr.16: 16157695-16157695 on Build GRCh38
rs751507784 Chr.16: 16157850-16157851 on Build GRCh38
rs761440521 Chr.16: 16157773-16157773 on Build GRCh38
rs199694536 Chr.16: 16157806-16157806 on Build GRCh38
rs769820268 Chr.16: 16157723-16157723 on Build GRCh38
rs774245200 Chr.16: 16157658-16157658 on Build GRCh38
rs766376643 Chr.16: 16157846-16157849 on Build GRCh38
rs72664220 Chr.16: 16157775-16157776 on Build GRCh38
rs760304927 Chr.16: 16157675-16157675 on Build GRCh38
rs546064934 Chr.16: 16157759-16157759 on Build GRCh38
rs767330001 Chr.16: 16157857-16157864 on Build GRCh38
rs772108097 Chr.16: 16157808-16157808 on Build GRCh38
rs939902508 Chr.16: 16157863-16157863 on Build GRCh38
rs762571943 Chr.16: 16157842-16157842 on Build GRCh38
rs114175094 Chr.16: 16157750-16157750 on Build GRCh38
rs780710426 Chr.16: 16157621-16157621 on Build GRCh38
rs143212758 Chr.16: 16157790-16157790 on Build GRCh38
rs368379895 Chr.16: 16157743-16157743 on Build GRCh38
rs751910206 Chr.16: 16157682-16157682 on Build GRCh38
rs530115190 Chr.16: 16158031-16158031 on Build GRCh38
rs970168295 Chr.16: 16158021-16158023 on Build GRCh38
rs756399001 Chr.16: 16157857-16157857 on Build GRCh38
rs772835266 Chr.16: 16157823-16157823 on Build GRCh38
rs750897812 Chr.16: 16157776-16157776 on Build GRCh38
rs1038021017 Chr.16: 16157660-16157660 on Build GRCh38
rs752924031 Chr.16: 16157849-16157849 on Build GRCh38
rs764975772 Chr.16: 16157821-16157821 on Build GRCh38
rs766355600 Chr.16: 16157641-16157641 on Build GRCh38
rs577119089 Chr.16: 16157900-16157900 on Build GRCh38
rs548075587 Chr.16: 16157972-16157972 on Build GRCh38
rs1038326032 Chr.16: 16157872-16157872 on Build GRCh38
rs60232261 Chr.16: 16157615-16157615 on Build GRCh38
rs771770753 Chr.16: 16157749-16157749 on Build GRCh38
rs751967992 Chr.16: 16157835-16157835 on Build GRCh38
rs994778528 Chr.16: 16157903-16157903 on Build GRCh38
rs764868012 Chr.16: 16157788-16157788 on Build GRCh38
rs945151292 Chr.16: 16157655-16157655 on Build GRCh38
rs141731889 Chr.16: 16157692-16157692 on Build GRCh38
rs758402419 Chr.16: 16157934-16157936 on Build GRCh38

View Full Product Details