Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20431104-20431603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM5
Entrez Gene ID
54988
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
-
UniGene
Hs.659606
Gene Chromosome Location
Chr.16: 20409508-20441333 on Build GRCh38
Transcripts
NM_001324371.1 NM_001324372.1 NM_017888.2 XM_006721056.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
434800 c.1261G>A p.V421I Chr.16: 20431274-20431274 on Build GRCh38
4392393 c.1258G>A p.A420T Chr.16: 20431271-20431271 on Build GRCh38
249721 c.1227C>T p.N409N Chr.16: 20431240-20431240 on Build GRCh38
3506933 c.1293C>T p.F431F Chr.16: 20431306-20431306 on Build GRCh38
6522628 c.1255G>A p.V419I Chr.16: 20431268-20431268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947566297 Chr.16: 20431410-20431410 on Build GRCh38
rs763901919 Chr.16: 20431360-20431360 on Build GRCh38
rs142785089 Chr.16: 20431277-20431277 on Build GRCh38
rs35468793 Chr.16: 20431188-20431188 on Build GRCh38
rs111385695 Chr.16: 20431383-20431383 on Build GRCh38
rs751142396 Chr.16: 20431261-20431261 on Build GRCh38
rs749929942 Chr.16: 20431183-20431183 on Build GRCh38
rs765574822 Chr.16: 20431293-20431293 on Build GRCh38
rs201750481 Chr.16: 20431232-20431232 on Build GRCh38
rs772103503 Chr.16: 20431225-20431225 on Build GRCh38
rs112497504 Chr.16: 20431209-20431209 on Build GRCh38
rs1008764590 Chr.16: 20431474-20431474 on Build GRCh38
rs767028553 Chr.16: 20431351-20431351 on Build GRCh38
rs755882961 Chr.16: 20431326-20431326 on Build GRCh38
rs375068483 Chr.16: 20431342-20431342 on Build GRCh38
rs748037521 Chr.16: 20431206-20431206 on Build GRCh38
rs775744839 Chr.16: 20431281-20431281 on Build GRCh38
rs78006992 Chr.16: 20431240-20431240 on Build GRCh38
rs564107665 Chr.16: 20431144-20431144 on Build GRCh38
rs762136578 Chr.16: 20431241-20431241 on Build GRCh38
rs899857417 Chr.16: 20431484-20431484 on Build GRCh38
rs375674529 Chr.16: 20431221-20431221 on Build GRCh38
rs777435072 Chr.16: 20431329-20431329 on Build GRCh38
rs35178682 Chr.16: 20431483-20431483 on Build GRCh38
rs757161737 Chr.16: 20431364-20431364 on Build GRCh38
rs768981168 Chr.16: 20431287-20431287 on Build GRCh38
rs951818369 Chr.16: 20431556-20431556 on Build GRCh38
rs768927709 Chr.16: 20431334-20431334 on Build GRCh38
rs79364355 Chr.16: 20431286-20431286 on Build GRCh38
rs752352364 Chr.16: 20431323-20431323 on Build GRCh38
rs757908841 Chr.16: 20431260-20431260 on Build GRCh38
rs144464842 Chr.16: 20431278-20431278 on Build GRCh38
rs539819728 Chr.16: 20431579-20431579 on Build GRCh38
rs368996292 Chr.16: 20431353-20431353 on Build GRCh38
rs77351186 Chr.16: 20431291-20431291 on Build GRCh38
rs375332436 Chr.16: 20431274-20431274 on Build GRCh38
rs769255837 Chr.16: 20431372-20431372 on Build GRCh38
rs7195862 Chr.16: 20431561-20431561 on Build GRCh38
rs777216223 Chr.16: 20431352-20431356 on Build GRCh38
rs772224450 Chr.16: 20431182-20431185 on Build GRCh38
rs747014171 Chr.16: 20431260-20431261 on Build GRCh38
rs752328734 Chr.16: 20431352-20431352 on Build GRCh38
rs772124410 Chr.16: 20431280-20431280 on Build GRCh38
rs370049743 Chr.16: 20431161-20431161 on Build GRCh38
rs748330167 Chr.16: 20431337-20431337 on Build GRCh38
rs780355858 Chr.16: 20431253-20431253 on Build GRCh38
rs760982337 Chr.16: 20431233-20431233 on Build GRCh38
rs750446518 Chr.16: 20431370-20431370 on Build GRCh38
rs762235941 Chr.16: 20431292-20431292 on Build GRCh38
rs534667927 Chr.16: 20431439-20431439 on Build GRCh38
rs762480553 Chr.16: 20431359-20431359 on Build GRCh38
rs753556837 Chr.16: 20431361-20431361 on Build GRCh38
rs893329509 Chr.16: 20431197-20431197 on Build GRCh38
rs11297710 Chr.16: 20431567-20431567 on Build GRCh38
rs569949016 Chr.16: 20431312-20431312 on Build GRCh38
rs113795675 Chr.16: 20431341-20431341 on Build GRCh38
rs773707858 Chr.16: 20431295-20431295 on Build GRCh38
rs764517848 Chr.16: 20431236-20431236 on Build GRCh38
rs768797930 Chr.16: 20431276-20431276 on Build GRCh38
rs553345452 Chr.16: 20431442-20431442 on Build GRCh38
rs750955635 Chr.16: 20431245-20431245 on Build GRCh38
rs757889570 Chr.16: 20431186-20431186 on Build GRCh38
rs548352099 Chr.16: 20431301-20431301 on Build GRCh38
rs139795247 Chr.16: 20431315-20431315 on Build GRCh38
rs754529922 Chr.16: 20431313-20431313 on Build GRCh38
rs369403661 Chr.16: 20431273-20431273 on Build GRCh38
rs773476450 Chr.16: 20431340-20431340 on Build GRCh38
rs773070800 Chr.16: 20431216-20431216 on Build GRCh38
rs940374553 Chr.16: 20431144-20431145 on Build GRCh38
rs781460894 Chr.16: 20431336-20431336 on Build GRCh38
rs981621422 Chr.16: 20431577-20431577 on Build GRCh38
rs984503539 Chr.16: 20431311-20431311 on Build GRCh38
rs371510771 Chr.16: 20431333-20431333 on Build GRCh38
rs113319586 Chr.16: 20431392-20431392 on Build GRCh38
rs759040982 Chr.16: 20431348-20431348 on Build GRCh38
rs368984470 Chr.16: 20431330-20431330 on Build GRCh38
rs573108117 Chr.16: 20431562-20431562 on Build GRCh38
rs748603256 Chr.16: 20431408-20431408 on Build GRCh38
rs754656439 Chr.16: 20431271-20431271 on Build GRCh38
rs557185883 Chr.16: 20431519-20431519 on Build GRCh38
rs149191478 Chr.16: 20431558-20431558 on Build GRCh38
rs528029586 Chr.16: 20431146-20431146 on Build GRCh38
rs12923629 Chr.16: 20431533-20431533 on Build GRCh38
rs372936210 Chr.16: 20431369-20431369 on Build GRCh38
rs34985819 Chr.16: 20431331-20431331 on Build GRCh38
rs765831297 Chr.16: 20431306-20431306 on Build GRCh38
rs778242061 Chr.16: 20431177-20431177 on Build GRCh38
rs780969532 Chr.16: 20431202-20431202 on Build GRCh38
rs535990994 Chr.16: 20431510-20431510 on Build GRCh38
rs958378025 Chr.16: 20431580-20431580 on Build GRCh38
rs138834094 Chr.16: 20431272-20431272 on Build GRCh38
rs370692751 Chr.16: 20431166-20431166 on Build GRCh38
rs754707445 Chr.16: 20431189-20431189 on Build GRCh38
rs112259303 Chr.16: 20431138-20431138 on Build GRCh38
rs866278243 Chr.16: 20431296-20431296 on Build GRCh38
rs765746303 Chr.16: 20431244-20431244 on Build GRCh38
rs140218919 Chr.16: 20431336-20431337 on Build GRCh38

View Full Product Details