Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20777254-20777716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5732688 c.457C>A p.Q153K Chr.16: 20777399-20777399 on Build GRCh38
1134030 c.578C>T p.S193F Chr.16: 20777520-20777520 on Build GRCh38
434811 c.578C>T p.S193F Chr.16: 20777520-20777520 on Build GRCh38
6464254 c.436G>T p.V146F Chr.16: 20777378-20777378 on Build GRCh38
6875495 c.605G>A p.R202K Chr.16: 20777547-20777547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770276890 Chr.16: 20777609-20777609 on Build GRCh38
rs758393144 Chr.16: 20777601-20777602 on Build GRCh38
rs143213955 Chr.16: 20777546-20777546 on Build GRCh38
rs747284545 Chr.16: 20777577-20777577 on Build GRCh38
rs781100087 Chr.16: 20777453-20777453 on Build GRCh38
rs778986438 Chr.16: 20777366-20777366 on Build GRCh38
rs757258795 Chr.16: 20777467-20777467 on Build GRCh38
rs751297633 Chr.16: 20777447-20777447 on Build GRCh38
rs150984396 Chr.16: 20777560-20777560 on Build GRCh38
rs771106487 Chr.16: 20777352-20777352 on Build GRCh38
rs750175121 Chr.16: 20777508-20777508 on Build GRCh38
rs372704368 Chr.16: 20777505-20777505 on Build GRCh38
rs762382067 Chr.16: 20777515-20777515 on Build GRCh38
rs752449738 Chr.16: 20777372-20777373 on Build GRCh38
rs775951468 Chr.16: 20777383-20777383 on Build GRCh38
rs781577743 Chr.16: 20777585-20777585 on Build GRCh38
rs983176676 Chr.16: 20777500-20777500 on Build GRCh38
rs781120681 Chr.16: 20777340-20777340 on Build GRCh38
rs764853926 Chr.16: 20777427-20777427 on Build GRCh38
rs189780189 Chr.16: 20777565-20777565 on Build GRCh38
rs752542843 Chr.16: 20777547-20777547 on Build GRCh38
rs1043103252 Chr.16: 20777642-20777642 on Build GRCh38
rs778373703 Chr.16: 20777593-20777593 on Build GRCh38
rs746148438 Chr.16: 20777380-20777380 on Build GRCh38
rs370088701 Chr.16: 20777446-20777446 on Build GRCh38
rs984179274 Chr.16: 20777562-20777563 on Build GRCh38
rs772728883 Chr.16: 20777510-20777510 on Build GRCh38
rs371659980 Chr.16: 20777583-20777583 on Build GRCh38
rs777345053 Chr.16: 20777416-20777416 on Build GRCh38
rs946764078 Chr.16: 20777685-20777685 on Build GRCh38
rs748481816 Chr.16: 20777590-20777590 on Build GRCh38
rs182142229 Chr.16: 20777649-20777649 on Build GRCh38
rs749256255 Chr.16: 20777350-20777350 on Build GRCh38
rs769201801 Chr.16: 20777496-20777496 on Build GRCh38
rs754860763 Chr.16: 20777330-20777330 on Build GRCh38
rs761137788 Chr.16: 20777387-20777387 on Build GRCh38
rs145372426 Chr.16: 20777442-20777442 on Build GRCh38
rs147662918 Chr.16: 20777488-20777488 on Build GRCh38
rs773969718 Chr.16: 20777631-20777631 on Build GRCh38
rs5714 Chr.16: 20777530-20777530 on Build GRCh38
rs750454982 Chr.16: 20777563-20777563 on Build GRCh38
rs754246453 Chr.16: 20777595-20777597 on Build GRCh38
rs759310601 Chr.16: 20777542-20777542 on Build GRCh38
rs1000676491 Chr.16: 20777335-20777335 on Build GRCh38
rs750010645 Chr.16: 20777428-20777428 on Build GRCh38
rs765949959 Chr.16: 20777517-20777517 on Build GRCh38
rs13306612 Chr.16: 20777638-20777638 on Build GRCh38
rs778935281 Chr.16: 20777579-20777582 on Build GRCh38
rs780228731 Chr.16: 20777602-20777602 on Build GRCh38
rs142474793 Chr.16: 20777489-20777489 on Build GRCh38
rs761216925 Chr.16: 20777424-20777424 on Build GRCh38
rs772217945 Chr.16: 20777381-20777381 on Build GRCh38
rs779013336 Chr.16: 20777473-20777473 on Build GRCh38
rs755926697 Chr.16: 20777560-20777561 on Build GRCh38
rs745959106 Chr.16: 20777484-20777484 on Build GRCh38
rs372093711 Chr.16: 20777611-20777611 on Build GRCh38
rs780410233 Chr.16: 20777569-20777569 on Build GRCh38
rs769219892 Chr.16: 20777394-20777394 on Build GRCh38
rs776982840 Chr.16: 20777497-20777497 on Build GRCh38
rs916653279 Chr.16: 20777657-20777657 on Build GRCh38
rs751532131 Chr.16: 20777345-20777345 on Build GRCh38
rs549672711 Chr.16: 20777557-20777557 on Build GRCh38
rs748791925 Chr.16: 20777621-20777621 on Build GRCh38
rs545664454 Chr.16: 20777434-20777434 on Build GRCh38
rs754768789 Chr.16: 20777449-20777449 on Build GRCh38
rs757757477 Chr.16: 20777658-20777658 on Build GRCh38
rs74755779 Chr.16: 20777623-20777623 on Build GRCh38

View Full Product Details