Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67438331-67438826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0D1
Entrez Gene ID
9114
Gene Name
ATPase H+ transporting V0 subunit d1
Gene Aliases
ATP6D, ATP6DV, P39, VATX, VMA6, VPATPD
UniGene
Hs.106876
Gene Chromosome Location
Chr.16: 67438014-67481186 on Build GRCh38
Transcripts
NM_004691.4 XM_011523444.1 XM_011523445.1 XM_011523446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
703858 c.1015C>T p.R339C Chr.16: 67438569-67438569 on Build GRCh38
5054994 c.872A>C p.E291A Chr.16: 67438815-67438815 on Build GRCh38
3511032 c.1032C>T p.I344I Chr.16: 67438552-67438552 on Build GRCh38
6608070 c.1020C>T p.H340H Chr.16: 67438564-67438564 on Build GRCh38
4918902 c.1047T>A p.P349P Chr.16: 67438537-67438537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578126893 Chr.16: 67438470-67438470 on Build GRCh38
rs1036699897 Chr.16: 67438628-67438628 on Build GRCh38
rs878924881 Chr.16: 67438437-67438437 on Build GRCh38
rs1023638858 Chr.16: 67438459-67438460 on Build GRCh38
rs886239466 Chr.16: 67438424-67438424 on Build GRCh38
rs753620716 Chr.16: 67438563-67438563 on Build GRCh38
rs549838985 Chr.16: 67438777-67438777 on Build GRCh38
rs72650132 Chr.16: 67438399-67438399 on Build GRCh38
rs982353109 Chr.16: 67438462-67438463 on Build GRCh38
rs997438859 Chr.16: 67438411-67438411 on Build GRCh38
rs764219820 Chr.16: 67438630-67438630 on Build GRCh38
rs957162203 Chr.16: 67438702-67438702 on Build GRCh38
rs1019147198 Chr.16: 67438462-67438483 on Build GRCh38
rs796371986 Chr.16: 67438474-67438475 on Build GRCh38
rs369839747 Chr.16: 67438603-67438603 on Build GRCh38
rs780797657 Chr.16: 67438727-67438727 on Build GRCh38
rs765114517 Chr.16: 67438805-67438805 on Build GRCh38
rs772900902 Chr.16: 67438502-67438502 on Build GRCh38
rs551803640 Chr.16: 67438568-67438568 on Build GRCh38
rs140583108 Chr.16: 67438561-67438561 on Build GRCh38
rs796296060 Chr.16: 67438495-67438496 on Build GRCh38
rs1053099 Chr.16: 67438558-67438558 on Build GRCh38
rs759050226 Chr.16: 67438475-67438476 on Build GRCh38
rs997933140 Chr.16: 67438473-67438473 on Build GRCh38
rs1016332889 Chr.16: 67438451-67438454 on Build GRCh38
rs774521136 Chr.16: 67438767-67438767 on Build GRCh38
rs971643206 Chr.16: 67438527-67438527 on Build GRCh38
rs771325385 Chr.16: 67438518-67438518 on Build GRCh38
rs1038799686 Chr.16: 67438474-67438479 on Build GRCh38
rs980274520 Chr.16: 67438450-67438450 on Build GRCh38
rs752679223 Chr.16: 67438666-67438666 on Build GRCh38
rs761750442 Chr.16: 67438773-67438773 on Build GRCh38
rs201883077 Chr.16: 67438775-67438775 on Build GRCh38
rs376528391 Chr.16: 67438696-67438696 on Build GRCh38
rs937764068 Chr.16: 67438459-67438459 on Build GRCh38
rs777006819 Chr.16: 67438597-67438597 on Build GRCh38
rs758646608 Chr.16: 67438478-67438478 on Build GRCh38
rs762995882 Chr.16: 67438543-67438543 on Build GRCh38
rs537906663 Chr.16: 67438389-67438389 on Build GRCh38
rs759841989 Chr.16: 67438599-67438599 on Build GRCh38
rs778570597 Chr.16: 67438494-67438494 on Build GRCh38
rs41280900 Chr.16: 67438463-67438463 on Build GRCh38
rs1023732012 Chr.16: 67438576-67438576 on Build GRCh38
rs771582761 Chr.16: 67438582-67438582 on Build GRCh38
rs777066400 Chr.16: 67438519-67438519 on Build GRCh38
rs371591598 Chr.16: 67438587-67438587 on Build GRCh38
rs542032086 Chr.16: 67438408-67438408 on Build GRCh38
rs200429175 Chr.16: 67438493-67438496 on Build GRCh38
rs963443466 Chr.16: 67438452-67438461 on Build GRCh38
rs760987912 Chr.16: 67438506-67438506 on Build GRCh38
rs1037355921 Chr.16: 67438469-67438469 on Build GRCh38
rs372633318 Chr.16: 67438649-67438649 on Build GRCh38
rs775547778 Chr.16: 67438571-67438571 on Build GRCh38
rs760052943 Chr.16: 67438524-67438524 on Build GRCh38
rs756952941 Chr.16: 67438721-67438721 on Build GRCh38
rs767594277 Chr.16: 67438637-67438637 on Build GRCh38
rs377289330 Chr.16: 67438594-67438594 on Build GRCh38
rs775941945 Chr.16: 67438617-67438617 on Build GRCh38
rs909527419 Chr.16: 67438461-67438461 on Build GRCh38
rs574615864 Chr.16: 67438400-67438400 on Build GRCh38
rs779921900 Chr.16: 67438482-67438482 on Build GRCh38
rs768976563 Chr.16: 67438756-67438756 on Build GRCh38
rs947027035 Chr.16: 67438390-67438390 on Build GRCh38
rs746441844 Chr.16: 67438718-67438718 on Build GRCh38
rs956092828 Chr.16: 67438391-67438391 on Build GRCh38
rs777999629 Chr.16: 67438480-67438480 on Build GRCh38
rs939676803 Chr.16: 67438615-67438615 on Build GRCh38
rs570401854 Chr.16: 67438633-67438633 on Build GRCh38
rs56294631 Chr.16: 67438352-67438352 on Build GRCh38
rs531616220 Chr.16: 67438481-67438481 on Build GRCh38
rs987710350 Chr.16: 67438383-67438384 on Build GRCh38
rs773675588 Chr.16: 67438483-67438484 on Build GRCh38
rs764261151 Chr.16: 67438552-67438552 on Build GRCh38
rs970712579 Chr.16: 67438605-67438605 on Build GRCh38
rs751237326 Chr.16: 67438495-67438496 on Build GRCh38
rs926321748 Chr.16: 67438452-67438452 on Build GRCh38
rs746663639 Chr.16: 67438588-67438588 on Build GRCh38
rs1005125727 Chr.16: 67438451-67438452 on Build GRCh38
rs751799655 Chr.16: 67438560-67438560 on Build GRCh38
rs570363596 Chr.16: 67438368-67438368 on Build GRCh38
rs765307056 Chr.16: 67438655-67438655 on Build GRCh38
rs72650133 Chr.16: 67438475-67438475 on Build GRCh38
rs200216007 Chr.16: 67438569-67438569 on Build GRCh38
rs933921922 Chr.16: 67438647-67438647 on Build GRCh38
rs749813945 Chr.16: 67438754-67438754 on Build GRCh38
rs367983281 Chr.16: 67438490-67438490 on Build GRCh38
rs56123737 Chr.16: 67438412-67438412 on Build GRCh38
rs112420676 Chr.16: 67438790-67438790 on Build GRCh38
rs759472919 Chr.16: 67438789-67438789 on Build GRCh38
rs914343991 Chr.16: 67438388-67438388 on Build GRCh38
rs112722649 Chr.16: 67438526-67438526 on Build GRCh38
rs72650134 Chr.16: 67438496-67438496 on Build GRCh38
rs747711094 Chr.16: 67438575-67438575 on Build GRCh38
rs1048031882 Chr.16: 67438460-67438460 on Build GRCh38
rs752703060 Chr.16: 67438477-67438477 on Build GRCh38
rs1049660515 Chr.16: 67438699-67438699 on Build GRCh38
rs145078546 Chr.16: 67438474-67438477 on Build GRCh38
rs374496626 Chr.16: 67438639-67438639 on Build GRCh38
rs543440135 Chr.16: 67438483-67438483 on Build GRCh38
rs746269332 Chr.16: 67438729-67438729 on Build GRCh38
rs900198189 Chr.16: 67438409-67438409 on Build GRCh38
rs763007863 Chr.16: 67438629-67438629 on Build GRCh38
rs370081111 Chr.16: 67438474-67438475 on Build GRCh38
rs533634923 Chr.16: 67438562-67438562 on Build GRCh38
rs776033590 Chr.16: 67438744-67438744 on Build GRCh38
rs777359469 Chr.16: 67438701-67438701 on Build GRCh38
rs561831455 Chr.16: 67438486-67438486 on Build GRCh38
rs529386375 Chr.16: 67438495-67438495 on Build GRCh38
rs545786317 Chr.16: 67438471-67438471 on Build GRCh38
rs770136881 Chr.16: 67438739-67438739 on Build GRCh38
rs1010271486 Chr.16: 67438515-67438515 on Build GRCh38
rs778946445 Chr.16: 67438573-67438573 on Build GRCh38
rs62059264 Chr.16: 67438554-67438554 on Build GRCh38

View Full Product Details