Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68066233-68066741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370519 c.544G>A p.V182I Chr.16: 68066626-68066626 on Build GRCh38
148095 c.484-8C>T p.? Chr.16: 68066558-68066558 on Build GRCh38
5629536 c.426C>A p.S142R Chr.16: 68066325-68066325 on Build GRCh38
4141855 c.521G>A p.R174K Chr.16: 68066603-68066603 on Build GRCh38
5166335 c.483G>A p.S161S Chr.16: 68066382-68066382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560287191 Chr.16: 68066381-68066381 on Build GRCh38
rs779539040 Chr.16: 68066257-68066257 on Build GRCh38
rs780632542 Chr.16: 68066549-68066549 on Build GRCh38
rs371010915 Chr.16: 68066408-68066408 on Build GRCh38
rs370186983 Chr.16: 68066348-68066348 on Build GRCh38
rs781754215 Chr.16: 68066521-68066521 on Build GRCh38
rs746424094 Chr.16: 68066539-68066539 on Build GRCh38
rs777413248 Chr.16: 68066323-68066323 on Build GRCh38
rs2285911 Chr.16: 68066616-68066616 on Build GRCh38
rs758006703 Chr.16: 68066424-68066424 on Build GRCh38
rs527993932 Chr.16: 68066448-68066448 on Build GRCh38
rs750958768 Chr.16: 68066631-68066631 on Build GRCh38
rs759306836 Chr.16: 68066415-68066415 on Build GRCh38
rs753568808 Chr.16: 68066291-68066291 on Build GRCh38
rs59111022 Chr.16: 68066344-68066344 on Build GRCh38
rs932475681 Chr.16: 68066509-68066509 on Build GRCh38
rs762682039 Chr.16: 68066604-68066604 on Build GRCh38
rs765078349 Chr.16: 68066418-68066418 on Build GRCh38
rs757049539 Chr.16: 68066394-68066407 on Build GRCh38
rs2285912 Chr.16: 68066558-68066558 on Build GRCh38
rs760728678 Chr.16: 68066281-68066281 on Build GRCh38
rs199830338 Chr.16: 68066313-68066313 on Build GRCh38
rs974757607 Chr.16: 68066452-68066452 on Build GRCh38
rs371755202 Chr.16: 68066677-68066677 on Build GRCh38
rs1007119553 Chr.16: 68066296-68066296 on Build GRCh38
rs771516174 Chr.16: 68066594-68066594 on Build GRCh38
rs374450554 Chr.16: 68066423-68066423 on Build GRCh38
rs757836627 Chr.16: 68066322-68066322 on Build GRCh38
rs776538134 Chr.16: 68066592-68066592 on Build GRCh38
rs529784687 Chr.16: 68066634-68066634 on Build GRCh38
rs768865250 Chr.16: 68066557-68066557 on Build GRCh38
rs761809372 Chr.16: 68066382-68066382 on Build GRCh38
rs183964681 Chr.16: 68066483-68066483 on Build GRCh38
rs755328478 Chr.16: 68066667-68066667 on Build GRCh38
rs564941595 Chr.16: 68066455-68066455 on Build GRCh38
rs868256677 Chr.16: 68066589-68066589 on Build GRCh38
rs772095369 Chr.16: 68066562-68066562 on Build GRCh38
rs779163463 Chr.16: 68066671-68066671 on Build GRCh38
rs749971544 Chr.16: 68066658-68066658 on Build GRCh38
rs757724879 Chr.16: 68066342-68066342 on Build GRCh38
rs768614400 Chr.16: 68066369-68066369 on Build GRCh38
rs754787374 Chr.16: 68066297-68066297 on Build GRCh38
rs780720444 Chr.16: 68066639-68066639 on Build GRCh38
rs764283153 Chr.16: 68066268-68066268 on Build GRCh38
rs191637303 Chr.16: 68066266-68066266 on Build GRCh38
rs143329208 Chr.16: 68066593-68066593 on Build GRCh38
rs911882716 Chr.16: 68066422-68066422 on Build GRCh38
rs761361430 Chr.16: 68066622-68066622 on Build GRCh38
rs773009677 Chr.16: 68066278-68066278 on Build GRCh38
rs756751467 Chr.16: 68066519-68066519 on Build GRCh38
rs781470873 Chr.16: 68066661-68066661 on Build GRCh38
rs748379233 Chr.16: 68066676-68066676 on Build GRCh38
rs995039134 Chr.16: 68066715-68066715 on Build GRCh38
rs141092300 Chr.16: 68066327-68066327 on Build GRCh38
rs767190581 Chr.16: 68066623-68066623 on Build GRCh38
rs544740487 Chr.16: 68066307-68066307 on Build GRCh38
rs756532569 Chr.16: 68066544-68066544 on Build GRCh38
rs766227347 Chr.16: 68066290-68066290 on Build GRCh38
rs35174031 Chr.16: 68066620-68066620 on Build GRCh38
rs894069673 Chr.16: 68066288-68066288 on Build GRCh38
rs758705271 Chr.16: 68066476-68066476 on Build GRCh38
rs749578903 Chr.16: 68066359-68066359 on Build GRCh38
rs200147333 Chr.16: 68066403-68066403 on Build GRCh38
rs887215143 Chr.16: 68066718-68066718 on Build GRCh38
rs749619527 Chr.16: 68066553-68066553 on Build GRCh38
rs748217092 Chr.16: 68066560-68066560 on Build GRCh38
rs753166515 Chr.16: 68066368-68066368 on Build GRCh38
rs754180591 Chr.16: 68066646-68066646 on Build GRCh38
rs746197088 Chr.16: 68066345-68066345 on Build GRCh38
rs921102553 Chr.16: 68066463-68066463 on Build GRCh38
rs775211727 Chr.16: 68066596-68066596 on Build GRCh38
rs370618028 Chr.16: 68066694-68066694 on Build GRCh38
rs763667594 Chr.16: 68066518-68066518 on Build GRCh38
rs73604372 Chr.16: 68066449-68066449 on Build GRCh38

View Full Product Details