Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70356154-70356704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
UniGene
Hs.656037
Gene Chromosome Location
Chr.16: 70346829-70373383 on Build GRCh38
Transcripts
NM_001320522.1 NM_001320525.1 NM_001320526.1 NM_001320527.1 NM_018332.4 NR_135296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4516954 c.254_255CC>TT p.S85F Chr.16: 70356208-70356209 on Build GRCh38
6641589 c.242G>A p.R81Q Chr.16: 70356196-70356196 on Build GRCh38
7415039 c.256C>A p.P86T Chr.16: 70356210-70356210 on Build GRCh38
4481957 c.255C>T p.S85S Chr.16: 70356209-70356209 on Build GRCh38
3932406 c.261G>A p.L87L Chr.16: 70356215-70356215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972323812 Chr.16: 70356547-70356547 on Build GRCh38
rs984806680 Chr.16: 70356418-70356418 on Build GRCh38
rs746394824 Chr.16: 70356196-70356196 on Build GRCh38
rs913593411 Chr.16: 70356658-70356658 on Build GRCh38
rs1005859463 Chr.16: 70356658-70356659 on Build GRCh38
rs953370909 Chr.16: 70356637-70356637 on Build GRCh38
rs757576792 Chr.16: 70356192-70356192 on Build GRCh38
rs759409737 Chr.16: 70356382-70356382 on Build GRCh38
rs878904353 Chr.16: 70356423-70356423 on Build GRCh38
rs933309168 Chr.16: 70356362-70356363 on Build GRCh38
rs566146547 Chr.16: 70356449-70356449 on Build GRCh38
rs892064690 Chr.16: 70356502-70356502 on Build GRCh38
rs1034887766 Chr.16: 70356546-70356546 on Build GRCh38
rs747366110 Chr.16: 70356190-70356190 on Build GRCh38
rs576541771 Chr.16: 70356569-70356569 on Build GRCh38
rs907800248 Chr.16: 70356422-70356422 on Build GRCh38
rs1002901438 Chr.16: 70356647-70356647 on Build GRCh38
rs1047024814 Chr.16: 70356636-70356636 on Build GRCh38
rs971913831 Chr.16: 70356351-70356351 on Build GRCh38
rs758608117 Chr.16: 70356271-70356271 on Build GRCh38
rs529549411 Chr.16: 70356607-70356607 on Build GRCh38
rs748840873 Chr.16: 70356215-70356215 on Build GRCh38
rs986316546 Chr.16: 70356467-70356467 on Build GRCh38
rs925051787 Chr.16: 70356472-70356472 on Build GRCh38
rs558162418 Chr.16: 70356542-70356542 on Build GRCh38
rs778206737 Chr.16: 70356189-70356189 on Build GRCh38
rs750327028 Chr.16: 70356289-70356289 on Build GRCh38
rs1002048767 Chr.16: 70356387-70356387 on Build GRCh38
rs1024449609 Chr.16: 70356340-70356340 on Build GRCh38
rs981527985 Chr.16: 70356267-70356267 on Build GRCh38
rs756980710 Chr.16: 70356244-70356244 on Build GRCh38
rs113250197 Chr.16: 70356335-70356335 on Build GRCh38
rs935464628 Chr.16: 70356677-70356689 on Build GRCh38
rs931705828 Chr.16: 70356260-70356260 on Build GRCh38
rs548148267 Chr.16: 70356355-70356355 on Build GRCh38
rs894104422 Chr.16: 70356650-70356650 on Build GRCh38
rs533348608 Chr.16: 70356386-70356386 on Build GRCh38
rs769302675 Chr.16: 70356204-70356204 on Build GRCh38
rs945513262 Chr.16: 70356617-70356617 on Build GRCh38
rs766456844 Chr.16: 70356258-70356258 on Build GRCh38
rs1055153012 Chr.16: 70356307-70356307 on Build GRCh38
rs549401909 Chr.16: 70356648-70356648 on Build GRCh38
rs1010415417 Chr.16: 70356324-70356324 on Build GRCh38
rs550245297 Chr.16: 70356277-70356277 on Build GRCh38
rs1053951411 Chr.16: 70356408-70356408 on Build GRCh38
rs199754556 Chr.16: 70356272-70356272 on Build GRCh38
rs984584481 Chr.16: 70356290-70356290 on Build GRCh38
rs747092820 Chr.16: 70356556-70356556 on Build GRCh38
rs750463942 Chr.16: 70356642-70356642 on Build GRCh38
rs1025195469 Chr.16: 70356439-70356439 on Build GRCh38
rs887829804 Chr.16: 70356558-70356558 on Build GRCh38
rs539963316 Chr.16: 70356511-70356511 on Build GRCh38
rs560635654 Chr.16: 70356491-70356491 on Build GRCh38
rs138765022 Chr.16: 70356601-70356601 on Build GRCh38
rs952177756 Chr.16: 70356416-70356416 on Build GRCh38
rs892604433 Chr.16: 70356412-70356412 on Build GRCh38
rs570140381 Chr.16: 70356242-70356242 on Build GRCh38
rs1034425982 Chr.16: 70356401-70356401 on Build GRCh38
rs781676857 Chr.16: 70356194-70356194 on Build GRCh38
rs190949670 Chr.16: 70356372-70356372 on Build GRCh38
rs1015968155 Chr.16: 70356533-70356533 on Build GRCh38
rs986148793 Chr.16: 70356581-70356581 on Build GRCh38
rs1052072218 Chr.16: 70356501-70356501 on Build GRCh38
rs375778080 Chr.16: 70356243-70356243 on Build GRCh38
rs202151704 Chr.16: 70356249-70356249 on Build GRCh38
rs946256934 Chr.16: 70356503-70356503 on Build GRCh38
rs367756209 Chr.16: 70356609-70356609 on Build GRCh38
rs183995426 Chr.16: 70356437-70356437 on Build GRCh38
rs544486653 Chr.16: 70356595-70356595 on Build GRCh38
rs1045996039 Chr.16: 70356504-70356504 on Build GRCh38
rs780607979 Chr.16: 70356287-70356287 on Build GRCh38
rs923087123 Chr.16: 70356450-70356450 on Build GRCh38
rs751734875 Chr.16: 70356274-70356274 on Build GRCh38
rs892011819 Chr.16: 70356309-70356309 on Build GRCh38
rs540951555 Chr.16: 70356459-70356459 on Build GRCh38
rs768296316 Chr.16: 70356230-70356230 on Build GRCh38
rs372457359 Chr.16: 70356213-70356213 on Build GRCh38
rs958439151 Chr.16: 70356506-70356506 on Build GRCh38
rs931800596 Chr.16: 70356460-70356460 on Build GRCh38
rs774002056 Chr.16: 70356235-70356235 on Build GRCh38
rs764083084 Chr.16: 70356327-70356327 on Build GRCh38
rs936404263 Chr.16: 70356646-70356646 on Build GRCh38
rs746295754 Chr.16: 70356278-70356278 on Build GRCh38
rs1050887787 Chr.16: 70356363-70356363 on Build GRCh38
rs984469959 Chr.16: 70356519-70356519 on Build GRCh38
rs889190406 Chr.16: 70356368-70356368 on Build GRCh38
rs374463991 Chr.16: 70356372-70356373 on Build GRCh38
rs752819988 Chr.16: 70356268-70356268 on Build GRCh38
rs756551904 Chr.16: 70356281-70356281 on Build GRCh38
rs568633090 Chr.16: 70356334-70356334 on Build GRCh38
rs985941184 Chr.16: 70356638-70356638 on Build GRCh38

View Full Product Details