Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70364938-70365444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
UniGene
Hs.656037
Gene Chromosome Location
Chr.16: 70346829-70373383 on Build GRCh38
Transcripts
NM_001320522.1 NM_001320525.1 NM_001320526.1 NM_001320527.1 NM_018332.4 NR_135296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676769 c.597C>T p.G199G Chr.16: 70365124-70365124 on Build GRCh38
7415041 c.506C>A p.P169Q Chr.16: 70365033-70365033 on Build GRCh38
4882194 c.540G>A p.V180V Chr.16: 70365067-70365067 on Build GRCh38
7184351 c.544G>C p.E182Q Chr.16: 70365071-70365071 on Build GRCh38
1588514 c.592C>T p.R198* Chr.16: 70365119-70365119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779872534 Chr.16: 70365097-70365097 on Build GRCh38
rs187322384 Chr.16: 70365159-70365159 on Build GRCh38
rs929588484 Chr.16: 70365403-70365403 on Build GRCh38
rs912680010 Chr.16: 70365231-70365231 on Build GRCh38
rs373486221 Chr.16: 70365003-70365003 on Build GRCh38
rs755954098 Chr.16: 70364973-70364973 on Build GRCh38
rs927831950 Chr.16: 70365277-70365277 on Build GRCh38
rs759821766 Chr.16: 70365066-70365066 on Build GRCh38
rs571209680 Chr.16: 70365131-70365131 on Build GRCh38
rs537699457 Chr.16: 70365035-70365035 on Build GRCh38
rs776667114 Chr.16: 70365034-70365034 on Build GRCh38
rs1004245383 Chr.16: 70365417-70365417 on Build GRCh38
rs951343269 Chr.16: 70365414-70365414 on Build GRCh38
rs375119969 Chr.16: 70365182-70365182 on Build GRCh38
rs768522432 Chr.16: 70364996-70364996 on Build GRCh38
rs1043705997 Chr.16: 70365340-70365340 on Build GRCh38
rs750109222 Chr.16: 70365067-70365067 on Build GRCh38
rs924928504 Chr.16: 70365344-70365344 on Build GRCh38
rs879107549 Chr.16: 70365337-70365337 on Build GRCh38
rs978389676 Chr.16: 70365263-70365263 on Build GRCh38
rs879922723 Chr.16: 70365288-70365288 on Build GRCh38
rs749266310 Chr.16: 70364995-70364995 on Build GRCh38
rs199527052 Chr.16: 70365058-70365058 on Build GRCh38
rs1142621 Chr.16: 70365022-70365022 on Build GRCh38
rs756888308 Chr.16: 70365051-70365051 on Build GRCh38
rs747156360 Chr.16: 70365002-70365002 on Build GRCh38
rs951377551 Chr.16: 70365201-70365201 on Build GRCh38
rs530930838 Chr.16: 70365349-70365349 on Build GRCh38
rs78770095 Chr.16: 70365147-70365147 on Build GRCh38
rs139025377 Chr.16: 70365084-70365084 on Build GRCh38
rs749903226 Chr.16: 70365353-70365353 on Build GRCh38
rs1065164 Chr.16: 70365052-70365052 on Build GRCh38
rs371171153 Chr.16: 70365373-70365373 on Build GRCh38
rs927726375 Chr.16: 70364964-70364964 on Build GRCh38
rs746124056 Chr.16: 70365126-70365126 on Build GRCh38
rs916323288 Chr.16: 70365330-70365330 on Build GRCh38
rs201168154 Chr.16: 70365116-70365116 on Build GRCh38
rs754936890 Chr.16: 70365109-70365109 on Build GRCh38
rs368971017 Chr.16: 70365146-70365146 on Build GRCh38
rs78850640 Chr.16: 70365344-70365344 on Build GRCh38
rs939148908 Chr.16: 70365278-70365278 on Build GRCh38
rs966634429 Chr.16: 70365255-70365255 on Build GRCh38
rs536851920 Chr.16: 70365332-70365333 on Build GRCh38
rs1065165 Chr.16: 70365073-70365073 on Build GRCh38
rs754741041 Chr.16: 70365179-70365179 on Build GRCh38
rs779811698 Chr.16: 70364994-70364994 on Build GRCh38
rs541782346 Chr.16: 70365171-70365171 on Build GRCh38
rs1065163 Chr.16: 70365046-70365046 on Build GRCh38
rs1142622 Chr.16: 70365037-70365037 on Build GRCh38
rs1065166 Chr.16: 70365076-70365076 on Build GRCh38
rs995408543 Chr.16: 70365408-70365408 on Build GRCh38
rs941028528 Chr.16: 70365094-70365094 on Build GRCh38
rs1142624 Chr.16: 70365106-70365106 on Build GRCh38
rs763295747 Chr.16: 70365140-70365140 on Build GRCh38
rs533521616 Chr.16: 70365200-70365200 on Build GRCh38
rs947631037 Chr.16: 70365333-70365333 on Build GRCh38
rs776793650 Chr.16: 70365119-70365119 on Build GRCh38
rs749998772 Chr.16: 70365152-70365152 on Build GRCh38
rs759828933 Chr.16: 70365030-70365030 on Build GRCh38
rs545280419 Chr.16: 70365228-70365228 on Build GRCh38
rs965911445 Chr.16: 70365286-70365286 on Build GRCh38
rs192214557 Chr.16: 70365413-70365413 on Build GRCh38
rs918226968 Chr.16: 70365388-70365388 on Build GRCh38
rs111956811 Chr.16: 70365151-70365151 on Build GRCh38
rs778877553 Chr.16: 70365115-70365115 on Build GRCh38
rs776851368 Chr.16: 70365028-70365028 on Build GRCh38
rs990569906 Chr.16: 70365309-70365309 on Build GRCh38
rs74672881 Chr.16: 70365343-70365343 on Build GRCh38
rs935013724 Chr.16: 70365348-70365348 on Build GRCh38
rs563769161 Chr.16: 70365232-70365232 on Build GRCh38
rs769632901 Chr.16: 70364985-70364986 on Build GRCh38
rs905033757 Chr.16: 70365389-70365389 on Build GRCh38
rs749074446 Chr.16: 70365099-70365099 on Build GRCh38
rs773309555 Chr.16: 70364998-70364998 on Build GRCh38
rs200980673 Chr.16: 70365181-70365181 on Build GRCh38
rs764651371 Chr.16: 70365141-70365141 on Build GRCh38
rs145620463 Chr.16: 70365139-70365139 on Build GRCh38
rs560978330 Chr.16: 70365374-70365374 on Build GRCh38
rs751113198 Chr.16: 70365048-70365048 on Build GRCh38
rs567340676 Chr.16: 70365324-70365324 on Build GRCh38

View Full Product Details