Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70365959-70366459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
UniGene
Hs.656037
Gene Chromosome Location
Chr.16: 70346829-70373383 on Build GRCh38
Transcripts
NM_001320522.1 NM_001320525.1 NM_001320526.1 NM_001320527.1 NM_018332.4 NR_135296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379293 c.776G>A p.R259H Chr.16: 70366253-70366253 on Build GRCh38
4062339 c.685A>G p.K229E Chr.16: 70366162-70366162 on Build GRCh38
3272982 c.735C>T p.V245V Chr.16: 70366215-70366215 on Build GRCh38
3272973 c.681C>G p.L227L Chr.16: 70366161-70366161 on Build GRCh38
3272974 c.684C>G p.L228L Chr.16: 70366161-70366161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760158581 Chr.16: 70366045-70366045 on Build GRCh38
rs1000209904 Chr.16: 70366423-70366423 on Build GRCh38
rs775207 Chr.16: 70366029-70366029 on Build GRCh38
rs8053167 Chr.16: 70366430-70366430 on Build GRCh38
rs748586550 Chr.16: 70366125-70366125 on Build GRCh38
rs902583528 Chr.16: 70366168-70366168 on Build GRCh38
rs200890480 Chr.16: 70366295-70366295 on Build GRCh38
rs184118150 Chr.16: 70366365-70366365 on Build GRCh38
rs765789754 Chr.16: 70366402-70366402 on Build GRCh38
rs748459671 Chr.16: 70366226-70366226 on Build GRCh38
rs534563809 Chr.16: 70366366-70366366 on Build GRCh38
rs756163761 Chr.16: 70366104-70366104 on Build GRCh38
rs762676492 Chr.16: 70366298-70366298 on Build GRCh38
rs918326996 Chr.16: 70366006-70366006 on Build GRCh38
rs758849268 Chr.16: 70366255-70366255 on Build GRCh38
rs528424591 Chr.16: 70366073-70366073 on Build GRCh38
rs772742830 Chr.16: 70366015-70366015 on Build GRCh38
rs61741988 Chr.16: 70366100-70366100 on Build GRCh38
rs578179991 Chr.16: 70366414-70366414 on Build GRCh38
rs769150590 Chr.16: 70366011-70366011 on Build GRCh38
rs549148089 Chr.16: 70366334-70366334 on Build GRCh38
rs553026012 Chr.16: 70366409-70366409 on Build GRCh38
rs371184098 Chr.16: 70366164-70366164 on Build GRCh38
rs1023470599 Chr.16: 70366308-70366308 on Build GRCh38
rs1050154586 Chr.16: 70366400-70366400 on Build GRCh38
rs770052125 Chr.16: 70366000-70366000 on Build GRCh38
rs201189165 Chr.16: 70366098-70366098 on Build GRCh38
rs1025418046 Chr.16: 70366337-70366337 on Build GRCh38
rs201866549 Chr.16: 70366159-70366159 on Build GRCh38
rs775125839 Chr.16: 70366296-70366296 on Build GRCh38
rs946872428 Chr.16: 70366143-70366143 on Build GRCh38
rs879172598 Chr.16: 70366283-70366283 on Build GRCh38
rs774300984 Chr.16: 70366305-70366305 on Build GRCh38
rs867260263 Chr.16: 70366062-70366062 on Build GRCh38
rs986524016 Chr.16: 70366388-70366388 on Build GRCh38
rs537134667 Chr.16: 70366332-70366332 on Build GRCh38
rs963673130 Chr.16: 70366393-70366393 on Build GRCh38
rs569784904 Chr.16: 70366331-70366331 on Build GRCh38
rs767278230 Chr.16: 70366245-70366245 on Build GRCh38
rs758301632 Chr.16: 70366078-70366078 on Build GRCh38
rs143009306 Chr.16: 70366090-70366090 on Build GRCh38
rs779385922 Chr.16: 70366117-70366117 on Build GRCh38
rs774963593 Chr.16: 70366012-70366012 on Build GRCh38
rs376141982 Chr.16: 70366273-70366273 on Build GRCh38
rs575675204 Chr.16: 70366010-70366010 on Build GRCh38
rs1014405028 Chr.16: 70366436-70366436 on Build GRCh38
rs199645926 Chr.16: 70366137-70366137 on Build GRCh38
rs762446062 Chr.16: 70366391-70366391 on Build GRCh38
rs773603169 Chr.16: 70366134-70366134 on Build GRCh38
rs922170748 Chr.16: 70366426-70366426 on Build GRCh38
rs149332833 Chr.16: 70366206-70366206 on Build GRCh38
rs763815071 Chr.16: 70366301-70366301 on Build GRCh38
rs79528925 Chr.16: 70366004-70366004 on Build GRCh38
rs765996376 Chr.16: 70366060-70366060 on Build GRCh38
rs764910019 Chr.16: 70366151-70366151 on Build GRCh38
rs751462303 Chr.16: 70366166-70366166 on Build GRCh38
rs373817509 Chr.16: 70366289-70366289 on Build GRCh38
rs377661184 Chr.16: 70366128-70366128 on Build GRCh38
rs775262145 Chr.16: 70366158-70366158 on Build GRCh38
rs759334397 Chr.16: 70366063-70366063 on Build GRCh38
rs780259615 Chr.16: 70366105-70366105 on Build GRCh38
rs138660103 Chr.16: 70366189-70366189 on Build GRCh38
rs560941536 Chr.16: 70366068-70366068 on Build GRCh38
rs776976517 Chr.16: 70366390-70366390 on Build GRCh38
rs762384210 Chr.16: 70366014-70366014 on Build GRCh38
rs150154889 Chr.16: 70366138-70366138 on Build GRCh38
rs779347305 Chr.16: 70366223-70366223 on Build GRCh38
rs775781145 Chr.16: 70366336-70366336 on Build GRCh38
rs776297933 Chr.16: 70366140-70366140 on Build GRCh38
rs1028392514 Chr.16: 70365995-70365997 on Build GRCh38
rs781227576 Chr.16: 70365992-70365992 on Build GRCh38
rs754002692 Chr.16: 70366195-70366195 on Build GRCh38
rs745975379 Chr.16: 70365995-70365995 on Build GRCh38
rs769116860 Chr.16: 70366343-70366343 on Build GRCh38
rs576812276 Chr.16: 70366016-70366016 on Build GRCh38
rs769097524 Chr.16: 70366113-70366113 on Build GRCh38
rs771509381 Chr.16: 70366280-70366280 on Build GRCh38
rs749506982 Chr.16: 70366112-70366112 on Build GRCh38
rs912171770 Chr.16: 70366040-70366040 on Build GRCh38
rs367567797 Chr.16: 70366274-70366274 on Build GRCh38

View Full Product Details