Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70463136-70463639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUK
Entrez Gene ID
197258
Gene Name
fucokinase
Gene Aliases
1110046B12Rik
UniGene
Hs.7907
Gene Chromosome Location
Chr.16: 70454421-70480274 on Build GRCh38
Transcripts
NM_145059.2 XM_006721162.1 XM_011522928.1 XM_011522929.1 XM_011522930.2 XM_011522931.2 XM_017023012.1 XM_017023013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6320526 c.96G>T p.R32R Chr.16: 70463636-70463636 on Build GRCh38
4609982 c.43A>C p.T15P Chr.16: 70463233-70463233 on Build GRCh38
4973362 c.83A>T p.E28V Chr.16: 70463623-70463623 on Build GRCh38
5385286 c.80G>A p.R27K Chr.16: 70463270-70463270 on Build GRCh38
4062352 c.95G>A p.R32Q Chr.16: 70463635-70463635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755446321 Chr.16: 70463174-70463174 on Build GRCh38
rs368050018 Chr.16: 70463315-70463315 on Build GRCh38
rs1040737872 Chr.16: 70463194-70463194 on Build GRCh38
rs765675488 Chr.16: 70463179-70463179 on Build GRCh38
rs113870721 Chr.16: 70463280-70463280 on Build GRCh38
rs763503298 Chr.16: 70463244-70463244 on Build GRCh38
rs748622011 Chr.16: 70463304-70463304 on Build GRCh38
rs373796796 Chr.16: 70463486-70463486 on Build GRCh38
rs1027702708 Chr.16: 70463506-70463506 on Build GRCh38
rs781511019 Chr.16: 70463299-70463299 on Build GRCh38
rs756863195 Chr.16: 70463254-70463254 on Build GRCh38
rs183247209 Chr.16: 70463457-70463457 on Build GRCh38
rs8044110 Chr.16: 70463202-70463202 on Build GRCh38
rs772446889 Chr.16: 70463595-70463595 on Build GRCh38
rs17883115 Chr.16: 70463345-70463345 on Build GRCh38
rs1052978360 Chr.16: 70463357-70463357 on Build GRCh38
rs751100550 Chr.16: 70463250-70463250 on Build GRCh38
rs966346754 Chr.16: 70463580-70463580 on Build GRCh38
rs777994978 Chr.16: 70463294-70463294 on Build GRCh38
rs952200550 Chr.16: 70463530-70463530 on Build GRCh38
rs1001668460 Chr.16: 70463473-70463473 on Build GRCh38
rs1024338868 Chr.16: 70463433-70463433 on Build GRCh38
rs759844582 Chr.16: 70463322-70463322 on Build GRCh38
rs751019383 Chr.16: 70463595-70463595 on Build GRCh38
rs922331583 Chr.16: 70463602-70463603 on Build GRCh38
rs932449852 Chr.16: 70463286-70463286 on Build GRCh38
rs923680697 Chr.16: 70463460-70463460 on Build GRCh38
rs769711844 Chr.16: 70463201-70463201 on Build GRCh38
rs201790833 Chr.16: 70463199-70463199 on Build GRCh38
rs765694378 Chr.16: 70463234-70463234 on Build GRCh38
rs749067299 Chr.16: 70463381-70463381 on Build GRCh38
rs375614106 Chr.16: 70463188-70463188 on Build GRCh38
rs754213321 Chr.16: 70463169-70463169 on Build GRCh38
rs1025680440 Chr.16: 70463252-70463252 on Build GRCh38
rs771553911 Chr.16: 70463493-70463493 on Build GRCh38
rs1014570263 Chr.16: 70463378-70463378 on Build GRCh38
rs1015080648 Chr.16: 70463564-70463564 on Build GRCh38
rs771135505 Chr.16: 70463310-70463310 on Build GRCh38
rs531713325 Chr.16: 70463608-70463608 on Build GRCh38
rs776996217 Chr.16: 70463232-70463232 on Build GRCh38
rs989302479 Chr.16: 70463499-70463499 on Build GRCh38
rs976730405 Chr.16: 70463600-70463600 on Build GRCh38
rs760937384 Chr.16: 70463228-70463228 on Build GRCh38
rs916411274 Chr.16: 70463556-70463556 on Build GRCh38
rs564516849 Chr.16: 70463594-70463594 on Build GRCh38
rs17879135 Chr.16: 70463332-70463332 on Build GRCh38
rs775226283 Chr.16: 70463510-70463510 on Build GRCh38
rs201727216 Chr.16: 70463233-70463233 on Build GRCh38
rs780033403 Chr.16: 70463263-70463263 on Build GRCh38
rs560405578 Chr.16: 70463441-70463441 on Build GRCh38
rs748197913 Chr.16: 70463224-70463224 on Build GRCh38
rs375505442 Chr.16: 70463308-70463308 on Build GRCh38
rs977423312 Chr.16: 70463316-70463316 on Build GRCh38
rs771063449 Chr.16: 70463230-70463230 on Build GRCh38
rs777307290 Chr.16: 70463182-70463182 on Build GRCh38
rs768772315 Chr.16: 70463284-70463284 on Build GRCh38
rs767154340 Chr.16: 70463617-70463617 on Build GRCh38
rs891692279 Chr.16: 70463359-70463359 on Build GRCh38
rs763450292 Chr.16: 70463590-70463590 on Build GRCh38
rs780717198 Chr.16: 70463257-70463257 on Build GRCh38
rs757937839 Chr.16: 70463180-70463180 on Build GRCh38
rs1049940237 Chr.16: 70463307-70463307 on Build GRCh38
rs755890429 Chr.16: 70463261-70463261 on Build GRCh38
rs748231264 Chr.16: 70463589-70463589 on Build GRCh38
rs753175029 Chr.16: 70463239-70463239 on Build GRCh38
rs1005705961 Chr.16: 70463270-70463270 on Build GRCh38
rs772245753 Chr.16: 70463227-70463227 on Build GRCh38
rs750181274 Chr.16: 70463258-70463258 on Build GRCh38
rs192346978 Chr.16: 70463361-70463361 on Build GRCh38
rs750010776 Chr.16: 70463619-70463619 on Build GRCh38
rs947960803 Chr.16: 70463559-70463559 on Build GRCh38
rs17880218 Chr.16: 70463563-70463563 on Build GRCh38
rs373922468 Chr.16: 70463587-70463587 on Build GRCh38
rs770124529 Chr.16: 70463584-70463584 on Build GRCh38

View Full Product Details