Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70579770-70580282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_152456.2

SNP Detail

SNP ID Chromosome Location
rs894238966 Chr.16: 70579996-70579996 on Build GRCh38
rs933191539 Chr.16: 70579848-70579848 on Build GRCh38
rs982611209 Chr.16: 70580209-70580209 on Build GRCh38
rs34391559 Chr.16: 70579831-70579832 on Build GRCh38
rs16970335 Chr.16: 70580105-70580105 on Build GRCh38
rs999033767 Chr.16: 70579850-70579850 on Build GRCh38
rs934815495 Chr.16: 70580029-70580029 on Build GRCh38
rs1055927518 Chr.16: 70579985-70579985 on Build GRCh38
rs951251674 Chr.16: 70579908-70579908 on Build GRCh38
rs934024185 Chr.16: 70579981-70579981 on Build GRCh38
rs925632990 Chr.16: 70579819-70579819 on Build GRCh38
rs986609626 Chr.16: 70579883-70579883 on Build GRCh38
rs1039711229 Chr.16: 70579967-70579967 on Build GRCh38
rs961945412 Chr.16: 70579825-70579826 on Build GRCh38
rs990008796 Chr.16: 70580179-70580179 on Build GRCh38
rs957650774 Chr.16: 70579861-70579861 on Build GRCh38
rs893370672 Chr.16: 70580065-70580065 on Build GRCh38
rs1030237375 Chr.16: 70580046-70580046 on Build GRCh38
rs778897039 Chr.16: 70579874-70579874 on Build GRCh38
rs564707538 Chr.16: 70580255-70580255 on Build GRCh38
rs1016803460 Chr.16: 70580225-70580225 on Build GRCh38
rs961726380 Chr.16: 70580030-70580030 on Build GRCh38
rs148550908 Chr.16: 70579836-70579836 on Build GRCh38
rs1023280924 Chr.16: 70579885-70579885 on Build GRCh38
rs555819549 Chr.16: 70579939-70579939 on Build GRCh38
rs773839352 Chr.16: 70580078-70580078 on Build GRCh38
rs1002398239 Chr.16: 70579972-70579972 on Build GRCh38
rs868515381 Chr.16: 70580010-70580010 on Build GRCh38
rs892681789 Chr.16: 70579903-70579903 on Build GRCh38
rs909795087 Chr.16: 70579947-70579947 on Build GRCh38
rs572651396 Chr.16: 70580122-70580122 on Build GRCh38
rs879934877 Chr.16: 70579913-70579913 on Build GRCh38
rs745855131 Chr.16: 70579970-70579970 on Build GRCh38
rs371835053 Chr.16: 70580121-70580121 on Build GRCh38
rs941299972 Chr.16: 70579964-70579964 on Build GRCh38
rs903324765 Chr.16: 70579845-70579845 on Build GRCh38
rs951198531 Chr.16: 70580196-70580196 on Build GRCh38
rs903273388 Chr.16: 70580011-70580011 on Build GRCh38
rs142934786 Chr.16: 70580041-70580041 on Build GRCh38
rs962610004 Chr.16: 70580239-70580239 on Build GRCh38
rs772113174 Chr.16: 70580007-70580007 on Build GRCh38
rs367630446 Chr.16: 70580074-70580074 on Build GRCh38
rs899918489 Chr.16: 70579833-70579833 on Build GRCh38
rs1010525366 Chr.16: 70579862-70579862 on Build GRCh38
rs537584254 Chr.16: 70579886-70579886 on Build GRCh38
rs192615934 Chr.16: 70580140-70580140 on Build GRCh38
rs982239893 Chr.16: 70579895-70579895 on Build GRCh38
rs1033189545 Chr.16: 70579851-70579851 on Build GRCh38
rs936646515 Chr.16: 70579887-70579887 on Build GRCh38
rs1051576108 Chr.16: 70579982-70579982 on Build GRCh38
rs927755051 Chr.16: 70579902-70579902 on Build GRCh38
rs1039762065 Chr.16: 70579822-70579822 on Build GRCh38
rs1023186982 Chr.16: 70580102-70580102 on Build GRCh38
rs942472022 Chr.16: 70579830-70579830 on Build GRCh38
rs954529385 Chr.16: 70580049-70580049 on Build GRCh38
rs558378060 Chr.16: 70579835-70579835 on Build GRCh38
rs546471536 Chr.16: 70580240-70580240 on Build GRCh38
rs748638903 Chr.16: 70579818-70579818 on Build GRCh38
rs906686653 Chr.16: 70579963-70579963 on Build GRCh38

View Full Product Details