Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7732945-7733470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
UniGene
Hs.367649
Gene Chromosome Location
Chr.17: 7717354-7833744 on Build GRCh38
Transcripts
NM_001303270.1 NM_020877.3 XM_011523663.1 XM_011523664.2 XM_011523666.1 XM_011523667.2 XM_011523670.2 XM_017024219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6082191 c.517C>T p.R173W Chr.17: 7733204-7733204 on Build GRCh38
4412732 c.546G>C p.Q182H Chr.17: 7733233-7733233 on Build GRCh38
7146522 c.545A>T p.Q182L Chr.17: 7733232-7733232 on Build GRCh38
6465218 c.492G>A p.R164R Chr.17: 7733179-7733179 on Build GRCh38
6082192 c.517C>T p.R173W Chr.17: 7733204-7733204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898084478 Chr.17: 7733018-7733018 on Build GRCh38
rs528216274 Chr.17: 7733354-7733354 on Build GRCh38
rs948458218 Chr.17: 7733173-7733173 on Build GRCh38
rs761368818 Chr.17: 7733310-7733310 on Build GRCh38
rs758169548 Chr.17: 7733213-7733213 on Build GRCh38
rs935498866 Chr.17: 7732973-7732973 on Build GRCh38
rs377473397 Chr.17: 7733290-7733290 on Build GRCh38
rs906890046 Chr.17: 7732972-7732972 on Build GRCh38
rs772642172 Chr.17: 7733437-7733437 on Build GRCh38
rs1008155431 Chr.17: 7733247-7733247 on Build GRCh38
rs568168640 Chr.17: 7733413-7733413 on Build GRCh38
rs763767414 Chr.17: 7733202-7733202 on Build GRCh38
rs539491504 Chr.17: 7733095-7733095 on Build GRCh38
rs757874423 Chr.17: 7733356-7733356 on Build GRCh38
rs146302909 Chr.17: 7733092-7733092 on Build GRCh38
rs146222206 Chr.17: 7733184-7733184 on Build GRCh38
rs555700792 Chr.17: 7733189-7733189 on Build GRCh38
rs759660250 Chr.17: 7733172-7733172 on Build GRCh38
rs192385531 Chr.17: 7733000-7733000 on Build GRCh38
rs772380905 Chr.17: 7733285-7733285 on Build GRCh38
rs754927352 Chr.17: 7733339-7733339 on Build GRCh38
rs763148087 Chr.17: 7733090-7733090 on Build GRCh38
rs752596895 Chr.17: 7733347-7733347 on Build GRCh38
rs757499711 Chr.17: 7733253-7733253 on Build GRCh38
rs778840302 Chr.17: 7733038-7733038 on Build GRCh38
rs764934724 Chr.17: 7733212-7733212 on Build GRCh38
rs778856645 Chr.17: 7733156-7733156 on Build GRCh38
rs961155722 Chr.17: 7733392-7733392 on Build GRCh38
rs1035276711 Chr.17: 7733163-7733163 on Build GRCh38
rs760100076 Chr.17: 7733115-7733115 on Build GRCh38
rs896295274 Chr.17: 7733412-7733412 on Build GRCh38
rs760146349 Chr.17: 7733149-7733149 on Build GRCh38
rs777840214 Chr.17: 7733245-7733245 on Build GRCh38
rs1031670185 Chr.17: 7733308-7733308 on Build GRCh38
rs371004348 Chr.17: 7733331-7733331 on Build GRCh38
rs771839916 Chr.17: 7733060-7733060 on Build GRCh38
rs1034965983 Chr.17: 7733380-7733380 on Build GRCh38
rs200142891 Chr.17: 7733351-7733351 on Build GRCh38
rs771081074 Chr.17: 7733185-7733185 on Build GRCh38
rs141741337 Chr.17: 7733182-7733182 on Build GRCh38
rs754842665 Chr.17: 7733236-7733236 on Build GRCh38
rs117035657 Chr.17: 7733179-7733179 on Build GRCh38
rs1036699012 Chr.17: 7733198-7733198 on Build GRCh38
rs148065459 Chr.17: 7733153-7733153 on Build GRCh38
rs770086862 Chr.17: 7733081-7733081 on Build GRCh38
rs375467657 Chr.17: 7733073-7733073 on Build GRCh38
rs139184787 Chr.17: 7733292-7733292 on Build GRCh38
rs919506568 Chr.17: 7733203-7733203 on Build GRCh38
rs995375122 Chr.17: 7733030-7733030 on Build GRCh38
rs1038674483 Chr.17: 7733040-7733040 on Build GRCh38
rs767858354 Chr.17: 7733116-7733116 on Build GRCh38
rs370412978 Chr.17: 7733046-7733046 on Build GRCh38
rs563244058 Chr.17: 7733288-7733288 on Build GRCh38
rs979367070 Chr.17: 7733446-7733446 on Build GRCh38
rs200709517 Chr.17: 7733193-7733193 on Build GRCh38
rs539380687 Chr.17: 7733428-7733428 on Build GRCh38
rs557178381 Chr.17: 7733014-7733014 on Build GRCh38
rs752642968 Chr.17: 7733187-7733187 on Build GRCh38
rs144172284 Chr.17: 7733155-7733155 on Build GRCh38
rs764810326 Chr.17: 7733320-7733320 on Build GRCh38
rs763679293 Chr.17: 7733150-7733150 on Build GRCh38
rs991292564 Chr.17: 7733178-7733178 on Build GRCh38
rs751515646 Chr.17: 7733229-7733229 on Build GRCh38
rs749313903 Chr.17: 7733250-7733250 on Build GRCh38
rs369539181 Chr.17: 7733176-7733176 on Build GRCh38
rs930295855 Chr.17: 7733345-7733345 on Build GRCh38
rs781348453 Chr.17: 7733068-7733068 on Build GRCh38
rs540695293 Chr.17: 7733188-7733188 on Build GRCh38
rs752279384 Chr.17: 7733146-7733146 on Build GRCh38
rs761232886 Chr.17: 7733197-7733197 on Build GRCh38
rs776303589 Chr.17: 7733293-7733293 on Build GRCh38
rs546639375 Chr.17: 7733359-7733359 on Build GRCh38
rs761571154 Chr.17: 7733205-7733205 on Build GRCh38
rs201980957 Chr.17: 7733194-7733194 on Build GRCh38
rs150564814 Chr.17: 7733164-7733164 on Build GRCh38
rs1004819528 Chr.17: 7733362-7733362 on Build GRCh38
rs981279189 Chr.17: 7733364-7733364 on Build GRCh38
rs751310471 Chr.17: 7733338-7733338 on Build GRCh38
rs1051207537 Chr.17: 7732981-7732981 on Build GRCh38
rs750130473 Chr.17: 7733157-7733157 on Build GRCh38
rs200727244 Chr.17: 7733330-7733330 on Build GRCh38
rs779041413 Chr.17: 7733262-7733262 on Build GRCh38
rs900150234 Chr.17: 7733048-7733048 on Build GRCh38
rs1027860879 Chr.17: 7733034-7733034 on Build GRCh38
rs568748840 Chr.17: 7733031-7733031 on Build GRCh38
rs746145863 Chr.17: 7733358-7733358 on Build GRCh38
rs746084486 Chr.17: 7733264-7733264 on Build GRCh38
rs774628888 Chr.17: 7733107-7733107 on Build GRCh38
rs745857145 Chr.17: 7733068-7733072 on Build GRCh38
rs758383301 Chr.17: 7733159-7733159 on Build GRCh38
rs369799470 Chr.17: 7733327-7733327 on Build GRCh38
rs780299896 Chr.17: 7733367-7733367 on Build GRCh38
rs575235290 Chr.17: 7733088-7733088 on Build GRCh38
rs1024136472 Chr.17: 7733436-7733436 on Build GRCh38
rs767398369 Chr.17: 7733340-7733340 on Build GRCh38
rs939683862 Chr.17: 7732994-7732994 on Build GRCh38
rs375036619 Chr.17: 7733217-7733217 on Build GRCh38
rs1019006283 Chr.17: 7733052-7733052 on Build GRCh38
rs753334898 Chr.17: 7733204-7733204 on Build GRCh38
rs915777179 Chr.17: 7733168-7733168 on Build GRCh38

View Full Product Details