Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8002947-8003438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2D
Entrez Gene ID
3000
Gene Name
guanylate cyclase 2D, retinal
Gene Aliases
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC
UniGene
Hs.592109
Gene Chromosome Location
Chr.17: 8002670-8020340 on Build GRCh38
Transcripts
NM_000180.3 XM_011523816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740418 c.154G>T p.A52S Chr.17: 8003201-8003201 on Build GRCh38
5446725 c.164C>T p.T55M Chr.17: 8003211-8003211 on Build GRCh38
6438625 c.334G>A p.G112R Chr.17: 8003381-8003381 on Build GRCh38
6429813 c.92G>C p.R31P Chr.17: 8003139-8003139 on Build GRCh38
5343053 c.263C>T p.P88L Chr.17: 8003310-8003310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753766842 Chr.17: 8003227-8003227 on Build GRCh38
rs748156659 Chr.17: 8003151-8003151 on Build GRCh38
rs281865408 Chr.17: 8003049-8003049 on Build GRCh38
rs9905393 Chr.17: 8003101-8003101 on Build GRCh38
rs760016257 Chr.17: 8003195-8003195 on Build GRCh38
rs1017055646 Chr.17: 8002995-8002995 on Build GRCh38
rs879364904 Chr.17: 8002978-8002978 on Build GRCh38
rs1014118881 Chr.17: 8003187-8003187 on Build GRCh38
rs776974102 Chr.17: 8003161-8003161 on Build GRCh38
rs749800225 Chr.17: 8003191-8003191 on Build GRCh38
rs998657543 Chr.17: 8003043-8003043 on Build GRCh38
rs61749669 Chr.17: 8003361-8003361 on Build GRCh38
rs9905402 Chr.17: 8003108-8003108 on Build GRCh38
rs61749668 Chr.17: 8003354-8003354 on Build GRCh38
rs1008930073 Chr.17: 8003096-8003096 on Build GRCh38
rs749432012 Chr.17: 8003324-8003324 on Build GRCh38
rs774688311 Chr.17: 8003193-8003193 on Build GRCh38
rs529594203 Chr.17: 8003416-8003416 on Build GRCh38
rs752996693 Chr.17: 8003285-8003285 on Build GRCh38
rs937378694 Chr.17: 8003116-8003116 on Build GRCh38
rs758260297 Chr.17: 8003269-8003269 on Build GRCh38
rs1042929481 Chr.17: 8002977-8002977 on Build GRCh38
rs61749665 Chr.17: 8003201-8003201 on Build GRCh38
rs755150485 Chr.17: 8003145-8003145 on Build GRCh38
rs781514479 Chr.17: 8003148-8003148 on Build GRCh38
rs771426657 Chr.17: 8003335-8003335 on Build GRCh38
rs922225034 Chr.17: 8003005-8003005 on Build GRCh38
rs764072081 Chr.17: 8003058-8003058 on Build GRCh38
rs1013246655 Chr.17: 8002971-8002971 on Build GRCh38
rs1016696083 Chr.17: 8003215-8003215 on Build GRCh38
rs954746846 Chr.17: 8003090-8003090 on Build GRCh38
rs774934785 Chr.17: 8003382-8003382 on Build GRCh38
rs567463643 Chr.17: 8003386-8003386 on Build GRCh38
rs61749667 Chr.17: 8003336-8003336 on Build GRCh38
rs281865409 Chr.17: 8003050-8003050 on Build GRCh38
rs558294899 Chr.17: 8003279-8003279 on Build GRCh38
rs752146300 Chr.17: 8003035-8003035 on Build GRCh38
rs765331376 Chr.17: 8003254-8003254 on Build GRCh38
rs763697863 Chr.17: 8003225-8003225 on Build GRCh38
rs573367793 Chr.17: 8003318-8003318 on Build GRCh38
rs761828482 Chr.17: 8003234-8003234 on Build GRCh38
rs753855098 Chr.17: 8003083-8003083 on Build GRCh38
rs975454942 Chr.17: 8002998-8002998 on Build GRCh38
rs1005255417 Chr.17: 8002982-8002982 on Build GRCh38
rs952193754 Chr.17: 8003273-8003287 on Build GRCh38
rs757041611 Chr.17: 8003097-8003097 on Build GRCh38
rs201414567 Chr.17: 8003211-8003211 on Build GRCh38
rs868091683 Chr.17: 8003206-8003206 on Build GRCh38
rs917247032 Chr.17: 8003410-8003410 on Build GRCh38
rs987666168 Chr.17: 8003071-8003071 on Build GRCh38
rs755696177 Chr.17: 8003042-8003042 on Build GRCh38
rs987726980 Chr.17: 8003262-8003262 on Build GRCh38
rs557108466 Chr.17: 8003121-8003121 on Build GRCh38
rs540877830 Chr.17: 8003353-8003353 on Build GRCh38
rs1056763147 Chr.17: 8003224-8003224 on Build GRCh38
rs756121043 Chr.17: 8003311-8003311 on Build GRCh38
rs775228379 Chr.17: 8003203-8003203 on Build GRCh38
rs963754336 Chr.17: 8002996-8002996 on Build GRCh38
rs63749076 Chr.17: 8003146-8003147 on Build GRCh38
rs926004217 Chr.17: 8003109-8003109 on Build GRCh38
rs61749664 Chr.17: 8003168-8003168 on Build GRCh38
rs562248093 Chr.17: 8003390-8003390 on Build GRCh38
rs964109390 Chr.17: 8003221-8003221 on Build GRCh38
rs750153057 Chr.17: 8003265-8003265 on Build GRCh38
rs281865410 Chr.17: 8003273-8003286 on Build GRCh38
rs771232307 Chr.17: 8003192-8003192 on Build GRCh38
rs1017601312 Chr.17: 8003105-8003105 on Build GRCh38
rs772514611 Chr.17: 8003364-8003364 on Build GRCh38
rs552184470 Chr.17: 8003171-8003176 on Build GRCh38
rs979318508 Chr.17: 8003287-8003287 on Build GRCh38
rs920824717 Chr.17: 8003134-8003134 on Build GRCh38
rs61750198 Chr.17: 8003388-8003388 on Build GRCh38
rs61749663 Chr.17: 8003134-8003135 on Build GRCh38

View Full Product Details