Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9912292-9912803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS7
Entrez Gene ID
8522
Gene Name
growth arrest specific 7
Gene Aliases
MLL/GAS7
UniGene
Hs.462214
Gene Chromosome Location
Chr.17: 9910609-10198551 on Build GRCh38
Transcripts
NM_001130831.1 NM_003644.2 NM_201432.1 NM_201433.1 XM_005256831.3 XM_005256832.3 XM_005256833.3 XM_005256834.3 XM_005256835.3 XM_011524044.2 XM_017025239.1 XM_017025240.1 XM_017025241.1 XM_017025242.1

SNP Detail

SNP ID Chromosome Location
rs951010175 Chr.17: 9912738-9912738 on Build GRCh38
rs1024945888 Chr.17: 9912589-9912589 on Build GRCh38
rs982650002 Chr.17: 9912556-9912556 on Build GRCh38
rs543458639 Chr.17: 9912515-9912515 on Build GRCh38
rs1046509571 Chr.17: 9912780-9912780 on Build GRCh38
rs376434832 Chr.17: 9912351-9912351 on Build GRCh38
rs772751302 Chr.17: 9912591-9912591 on Build GRCh38
rs866312621 Chr.17: 9912464-9912464 on Build GRCh38
rs544991000 Chr.17: 9912615-9912615 on Build GRCh38
rs750520345 Chr.17: 9912349-9912349 on Build GRCh38
rs1051112783 Chr.17: 9912436-9912436 on Build GRCh38
rs745434889 Chr.17: 9912609-9912609 on Build GRCh38
rs946356503 Chr.17: 9912639-9912639 on Build GRCh38
rs866658397 Chr.17: 9912530-9912530 on Build GRCh38
rs773821282 Chr.17: 9912602-9912602 on Build GRCh38
rs73974331 Chr.17: 9912742-9912742 on Build GRCh38
rs913751236 Chr.17: 9912627-9912627 on Build GRCh38
rs549828607 Chr.17: 9912772-9912772 on Build GRCh38
rs762671102 Chr.17: 9912678-9912678 on Build GRCh38
rs2191109 Chr.17: 9912601-9912601 on Build GRCh38
rs143185466 Chr.17: 9912668-9912668 on Build GRCh38
rs998391589 Chr.17: 9912330-9912330 on Build GRCh38
rs1002244339 Chr.17: 9912595-9912595 on Build GRCh38
rs973343669 Chr.17: 9912734-9912734 on Build GRCh38
rs929583432 Chr.17: 9912546-9912546 on Build GRCh38
rs78370174 Chr.17: 9912685-9912685 on Build GRCh38
rs1026277020 Chr.17: 9912737-9912737 on Build GRCh38
rs976475820 Chr.17: 9912669-9912669 on Build GRCh38
rs748471747 Chr.17: 9912552-9912552 on Build GRCh38
rs16958968 Chr.17: 9912671-9912671 on Build GRCh38
rs988357537 Chr.17: 9912626-9912626 on Build GRCh38
rs774803910 Chr.17: 9912675-9912675 on Build GRCh38
rs554775802 Chr.17: 9912493-9912493 on Build GRCh38
rs779448088 Chr.17: 9912540-9912540 on Build GRCh38
rs1042082784 Chr.17: 9912652-9912652 on Build GRCh38
rs921049351 Chr.17: 9912670-9912670 on Build GRCh38
rs1029498132 Chr.17: 9912358-9912358 on Build GRCh38
rs780210263 Chr.17: 9912449-9912449 on Build GRCh38
rs572575120 Chr.17: 9912410-9912410 on Build GRCh38
rs536705853 Chr.17: 9912429-9912429 on Build GRCh38
rs963466506 Chr.17: 9912725-9912725 on Build GRCh38
rs75453384 Chr.17: 9912511-9912511 on Build GRCh38
rs954932126 Chr.17: 9912386-9912386 on Build GRCh38
rs982398742 Chr.17: 9912740-9912740 on Build GRCh38
rs16958974 Chr.17: 9912760-9912760 on Build GRCh38
rs556018103 Chr.17: 9912516-9912516 on Build GRCh38
rs1036502589 Chr.17: 9912701-9912701 on Build GRCh38
rs910667911 Chr.17: 9912459-9912459 on Build GRCh38

View Full Product Details