Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9913730-9914225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS7
Entrez Gene ID
8522
Gene Name
growth arrest specific 7
Gene Aliases
MLL/GAS7
UniGene
Hs.462214
Gene Chromosome Location
Chr.17: 9910609-10198551 on Build GRCh38
Transcripts
NM_001130831.1 NM_003644.2 NM_201432.1 NM_201433.1 XM_005256831.3 XM_005256832.3 XM_005256833.3 XM_005256834.3 XM_005256835.3 XM_011524044.2 XM_017025239.1 XM_017025240.1 XM_017025241.1 XM_017025242.1

SNP Detail

SNP ID Chromosome Location
rs935376420 Chr.17: 9913981-9913981 on Build GRCh38
rs991283949 Chr.17: 9914110-9914110 on Build GRCh38
rs966729357 Chr.17: 9913809-9913809 on Build GRCh38
rs559981420 Chr.17: 9913854-9913854 on Build GRCh38
rs1026717798 Chr.17: 9914052-9914052 on Build GRCh38
rs933998344 Chr.17: 9913851-9913851 on Build GRCh38
rs565016589 Chr.17: 9913805-9913805 on Build GRCh38
rs941246950 Chr.17: 9914109-9914109 on Build GRCh38
rs1026625873 Chr.17: 9913868-9913868 on Build GRCh38
rs918539644 Chr.17: 9914125-9914125 on Build GRCh38
rs1029688804 Chr.17: 9914076-9914076 on Build GRCh38
rs994038743 Chr.17: 9913970-9913970 on Build GRCh38
rs377302119 Chr.17: 9913891-9913891 on Build GRCh38
rs539092148 Chr.17: 9913835-9913835 on Build GRCh38
rs1001187174 Chr.17: 9913992-9913992 on Build GRCh38
rs944311541 Chr.17: 9914199-9914199 on Build GRCh38
rs1003918957 Chr.17: 9914072-9914072 on Build GRCh38
rs963772440 Chr.17: 9913834-9913834 on Build GRCh38
rs752965248 Chr.17: 9914107-9914107 on Build GRCh38
rs577432546 Chr.17: 9914004-9914004 on Build GRCh38
rs983091089 Chr.17: 9913888-9913888 on Build GRCh38
rs1052480929 Chr.17: 9914185-9914185 on Build GRCh38
rs554914251 Chr.17: 9914177-9914177 on Build GRCh38
rs73974333 Chr.17: 9913963-9913963 on Build GRCh38
rs1022381530 Chr.17: 9914048-9914048 on Build GRCh38
rs928590474 Chr.17: 9913931-9913931 on Build GRCh38
rs1008420145 Chr.17: 9913779-9913779 on Build GRCh38
rs568455649 Chr.17: 9914111-9914111 on Build GRCh38
rs1033925679 Chr.17: 9913755-9913755 on Build GRCh38
rs563492837 Chr.17: 9913964-9913964 on Build GRCh38
rs141903315 Chr.17: 9914034-9914034 on Build GRCh38
rs978264706 Chr.17: 9914065-9914065 on Build GRCh38
rs891416551 Chr.17: 9914003-9914003 on Build GRCh38
rs868611194 Chr.17: 9914184-9914184 on Build GRCh38
rs925415346 Chr.17: 9914140-9914140 on Build GRCh38
rs536506819 Chr.17: 9914123-9914123 on Build GRCh38
rs531018880 Chr.17: 9913984-9913984 on Build GRCh38
rs911221199 Chr.17: 9914108-9914108 on Build GRCh38
rs772812154 Chr.17: 9913864-9913864 on Build GRCh38
rs938509076 Chr.17: 9913939-9913939 on Build GRCh38
rs766190609 Chr.17: 9913971-9913971 on Build GRCh38
rs11555789 Chr.17: 9914056-9914056 on Build GRCh38
rs73974334 Chr.17: 9914020-9914020 on Build GRCh38
rs959912226 Chr.17: 9913759-9913759 on Build GRCh38
rs989637900 Chr.17: 9913798-9913798 on Build GRCh38
rs912767621 Chr.17: 9914189-9914189 on Build GRCh38
rs951299573 Chr.17: 9913884-9913884 on Build GRCh38
rs1038176970 Chr.17: 9913933-9913933 on Build GRCh38
rs1045612725 Chr.17: 9914165-9914165 on Build GRCh38
rs140006736 Chr.17: 9913824-9913824 on Build GRCh38
rs915421802 Chr.17: 9913802-9913802 on Build GRCh38
rs941289017 Chr.17: 9913923-9913923 on Build GRCh38
rs929780472 Chr.17: 9914127-9914127 on Build GRCh38
rs991357400 Chr.17: 9913946-9913946 on Build GRCh38
rs1011584355 Chr.17: 9914037-9914037 on Build GRCh38
rs530297227 Chr.17: 9913949-9913949 on Build GRCh38
rs978148978 Chr.17: 9913820-9913820 on Build GRCh38
rs777861800 Chr.17: 9913792-9913792 on Build GRCh38
rs955661688 Chr.17: 9914091-9914091 on Build GRCh38
rs570900907 Chr.17: 9914027-9914027 on Build GRCh38
rs562697164 Chr.17: 9914031-9914031 on Build GRCh38
rs903869915 Chr.17: 9913980-9913980 on Build GRCh38
rs911117249 Chr.17: 9913889-9913889 on Build GRCh38

View Full Product Details