Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32331575-32332076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
UniGene
Hs.655257
Gene Chromosome Location
Chr.17: 32328441-32350047 on Build GRCh38
Transcripts
NM_022344.3 XM_005258022.3 XM_017024940.1 XM_017024941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
86291 c.1035T>G p.Y345* Chr.17: 32331919-32331919 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752794931 Chr.17: 32331992-32331993 on Build GRCh38
rs768990260 Chr.17: 32331950-32331950 on Build GRCh38
rs775609529 Chr.17: 32331725-32331725 on Build GRCh38
rs764637468 Chr.17: 32332000-32332000 on Build GRCh38
rs546486052 Chr.17: 32331771-32331771 on Build GRCh38
rs1012875675 Chr.17: 32331879-32331879 on Build GRCh38
rs752266903 Chr.17: 32331845-32331845 on Build GRCh38
rs1019152 Chr.17: 32331987-32331987 on Build GRCh38
rs915407857 Chr.17: 32331854-32331854 on Build GRCh38
rs775130222 Chr.17: 32332014-32332014 on Build GRCh38
rs757776758 Chr.17: 32331873-32331873 on Build GRCh38
rs763381834 Chr.17: 32331719-32331720 on Build GRCh38
rs372489024 Chr.17: 32331941-32331941 on Build GRCh38
rs566913092 Chr.17: 32332038-32332038 on Build GRCh38
rs369641164 Chr.17: 32331810-32331810 on Build GRCh38
rs190347094 Chr.17: 32331695-32331695 on Build GRCh38
rs751928672 Chr.17: 32332024-32332024 on Build GRCh38
rs764841763 Chr.17: 32331843-32331843 on Build GRCh38
rs971291089 Chr.17: 32331929-32331929 on Build GRCh38
rs749459716 Chr.17: 32331946-32331946 on Build GRCh38
rs555785504 Chr.17: 32332048-32332048 on Build GRCh38
rs973743639 Chr.17: 32331639-32331639 on Build GRCh38
rs772924907 Chr.17: 32331955-32331955 on Build GRCh38
rs756118825 Chr.17: 32331901-32331901 on Build GRCh38
rs767971109 Chr.17: 32331848-32331849 on Build GRCh38
rs866298506 Chr.17: 32331894-32331894 on Build GRCh38
rs11557695 Chr.17: 32331635-32331635 on Build GRCh38
rs771743905 Chr.17: 32331760-32331760 on Build GRCh38
rs116000645 Chr.17: 32332051-32332051 on Build GRCh38
rs368821944 Chr.17: 32331856-32331856 on Build GRCh38
rs776093966 Chr.17: 32331718-32331719 on Build GRCh38
rs776517667 Chr.17: 32331998-32331998 on Build GRCh38
rs376413444 Chr.17: 32331747-32331747 on Build GRCh38
rs746827823 Chr.17: 32331983-32331983 on Build GRCh38
rs781771207 Chr.17: 32331874-32331874 on Build GRCh38
rs1025264261 Chr.17: 32331886-32331886 on Build GRCh38
rs774898851 Chr.17: 32331746-32331747 on Build GRCh38
rs370678601 Chr.17: 32332022-32332022 on Build GRCh38
rs751973701 Chr.17: 32331857-32331857 on Build GRCh38
rs771281387 Chr.17: 32331723-32331724 on Build GRCh38
rs758125219 Chr.17: 32331846-32331846 on Build GRCh38
rs761847652 Chr.17: 32331614-32331618 on Build GRCh38
rs1045239008 Chr.17: 32331898-32331898 on Build GRCh38
rs762443118 Chr.17: 32332019-32332019 on Build GRCh38
rs766308459 Chr.17: 32331776-32331776 on Build GRCh38
rs997396296 Chr.17: 32331646-32331646 on Build GRCh38
rs762304489 Chr.17: 32332025-32332025 on Build GRCh38
rs987130045 Chr.17: 32331764-32331764 on Build GRCh38
rs760015889 Chr.17: 32331757-32331758 on Build GRCh38
rs745905918 Chr.17: 32331878-32331878 on Build GRCh38
rs911554018 Chr.17: 32331785-32331785 on Build GRCh38
rs953281022 Chr.17: 32331700-32331700 on Build GRCh38
rs776070447 Chr.17: 32331799-32331799 on Build GRCh38
rs371772606 Chr.17: 32332044-32332044 on Build GRCh38
rs11557694 Chr.17: 32331641-32331641 on Build GRCh38

View Full Product Details