Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32334697-32335208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
UniGene
Hs.655257
Gene Chromosome Location
Chr.17: 32328441-32350047 on Build GRCh38
Transcripts
NM_022344.3 XM_005258022.3 XM_017024940.1 XM_017024941.1

SNP Detail

SNP ID Chromosome Location
rs746547031 Chr.17: 32334758-32334760 on Build GRCh38
rs541937909 Chr.17: 32334756-32334756 on Build GRCh38
rs763673 Chr.17: 32334903-32334903 on Build GRCh38
rs757482204 Chr.17: 32334788-32334788 on Build GRCh38
rs764124264 Chr.17: 32334759-32334759 on Build GRCh38
rs1000411272 Chr.17: 32334911-32334911 on Build GRCh38
rs746251152 Chr.17: 32334872-32334872 on Build GRCh38
rs1047698825 Chr.17: 32334955-32334955 on Build GRCh38
rs939901312 Chr.17: 32334989-32334989 on Build GRCh38
rs780965069 Chr.17: 32334850-32334851 on Build GRCh38
rs193057634 Chr.17: 32335142-32335142 on Build GRCh38
rs780093269 Chr.17: 32334757-32334758 on Build GRCh38
rs569939314 Chr.17: 32335116-32335116 on Build GRCh38
rs760400902 Chr.17: 32335060-32335070 on Build GRCh38
rs200381289 Chr.17: 32334825-32334825 on Build GRCh38
rs754907079 Chr.17: 32334800-32334800 on Build GRCh38
rs776925626 Chr.17: 32334731-32334731 on Build GRCh38
rs372303554 Chr.17: 32334830-32334830 on Build GRCh38
rs952631883 Chr.17: 32335080-32335080 on Build GRCh38
rs374835143 Chr.17: 32335165-32335165 on Build GRCh38
rs564377436 Chr.17: 32334919-32334919 on Build GRCh38
rs983758326 Chr.17: 32334724-32334724 on Build GRCh38
rs914688025 Chr.17: 32334837-32334837 on Build GRCh38
rs115007276 Chr.17: 32334866-32334866 on Build GRCh38
rs369049457 Chr.17: 32334880-32334880 on Build GRCh38
rs928765140 Chr.17: 32334918-32334918 on Build GRCh38
rs775496021 Chr.17: 32334740-32334740 on Build GRCh38
rs980589230 Chr.17: 32334873-32334873 on Build GRCh38
rs150075373 Chr.17: 32335012-32335012 on Build GRCh38
rs751787545 Chr.17: 32334784-32334784 on Build GRCh38
rs187999774 Chr.17: 32334970-32334970 on Build GRCh38
rs746667576 Chr.17: 32334843-32334843 on Build GRCh38
rs1014376825 Chr.17: 32335021-32335021 on Build GRCh38
rs777205617 Chr.17: 32334842-32334842 on Build GRCh38
rs774520638 Chr.17: 32334945-32334945 on Build GRCh38
rs770493008 Chr.17: 32334859-32334859 on Build GRCh38
rs764083782 Chr.17: 32335062-32335062 on Build GRCh38
rs753691500 Chr.17: 32334791-32334791 on Build GRCh38
rs1007919069 Chr.17: 32334729-32334729 on Build GRCh38
rs779016175 Chr.17: 32334821-32334821 on Build GRCh38
rs1040787844 Chr.17: 32334769-32334769 on Build GRCh38
rs894472179 Chr.17: 32335009-32335009 on Build GRCh38
rs759966846 Chr.17: 32334733-32334733 on Build GRCh38
rs1025863281 Chr.17: 32335076-32335076 on Build GRCh38
rs765745101 Chr.17: 32334737-32334737 on Build GRCh38
rs1024374950 Chr.17: 32335033-32335033 on Build GRCh38
rs767987268 Chr.17: 32334788-32334791 on Build GRCh38
rs554689154 Chr.17: 32335167-32335167 on Build GRCh38
rs976803922 Chr.17: 32335176-32335176 on Build GRCh38
rs537289521 Chr.17: 32335051-32335051 on Build GRCh38

View Full Product Details