Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35764698-35765205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf50
Entrez Gene ID
146853
Gene Name
chromosome 17 open reading frame 50
Gene Aliases
-
UniGene
Hs.127194
Gene Chromosome Location
Chr.17: 35760897-35765079 on Build GRCh38
Transcripts
NM_145272.3 NR_111988.1 XM_011525231.1 XM_017025064.1

Gene Symbol
MMP28
Entrez Gene ID
79148
Gene Name
matrix metallopeptidase 28
Gene Aliases
EPILYSIN, MM28, MMP-25, MMP-28, MMP25
UniGene
Hs.380710
Gene Chromosome Location
Chr.17: 35756249-35795707 on Build GRCh38
Transcripts
NM_145272.3 NR_111988.1 XM_011525231.1 XM_017025064.1

SNP Detail

SNP ID Chromosome Location
rs899438586 Chr.17: 35764974-35764974 on Build GRCh38
rs587642636 Chr.17: 35764899-35764899 on Build GRCh38
rs587652974 Chr.17: 35765174-35765174 on Build GRCh38
rs587656423 Chr.17: 35764742-35764742 on Build GRCh38
rs1010891691 Chr.17: 35765162-35765162 on Build GRCh38
rs938474603 Chr.17: 35764837-35764837 on Build GRCh38
rs1031064634 Chr.17: 35765042-35765042 on Build GRCh38
rs890640296 Chr.17: 35765001-35765001 on Build GRCh38
rs1022245564 Chr.17: 35765168-35765168 on Build GRCh38
rs374032267 Chr.17: 35765108-35765108 on Build GRCh38
rs782242573 Chr.17: 35764829-35764829 on Build GRCh38
rs587752891 Chr.17: 35764719-35764719 on Build GRCh38
rs587767145 Chr.17: 35764824-35764824 on Build GRCh38
rs587599722 Chr.17: 35764980-35764980 on Build GRCh38
rs17851934 Chr.17: 35764764-35764764 on Build GRCh38
rs113569545 Chr.17: 35764853-35764853 on Build GRCh38
rs1009453032 Chr.17: 35765030-35765030 on Build GRCh38
rs71381473 Chr.17: 35764852-35764852 on Build GRCh38
rs187126644 Chr.17: 35764913-35764913 on Build GRCh38
rs914515254 Chr.17: 35764728-35764728 on Build GRCh38
rs1051079004 Chr.17: 35764995-35764995 on Build GRCh38
rs1037351223 Chr.17: 35764960-35764960 on Build GRCh38
rs907595281 Chr.17: 35764893-35764893 on Build GRCh38
rs782569782 Chr.17: 35765121-35765121 on Build GRCh38
rs71381474 Chr.17: 35764942-35764942 on Build GRCh38
rs587723510 Chr.17: 35764718-35764718 on Build GRCh38
rs980045189 Chr.17: 35765181-35765181 on Build GRCh38
rs587705876 Chr.17: 35764823-35764823 on Build GRCh38
rs113176378 Chr.17: 35764828-35764828 on Build GRCh38
rs989418319 Chr.17: 35764717-35764717 on Build GRCh38
rs996432770 Chr.17: 35764975-35764975 on Build GRCh38
rs782662750 Chr.17: 35764785-35764785 on Build GRCh38
rs956770247 Chr.17: 35765150-35765150 on Build GRCh38

View Full Product Details