Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36554728-36555226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGNBP2
Entrez Gene ID
79893
Gene Name
gametogenetin binding protein 2
Gene Aliases
DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
UniGene
Hs.514116
Gene Chromosome Location
Chr.17: 36544888-36589848 on Build GRCh38
Transcripts
NM_024835.4 XM_005257689.2 XM_011525265.2 XM_011525266.1 XM_017025107.1

SNP Detail

SNP ID Chromosome Location
rs750733464 Chr.17: 36554918-36554918 on Build GRCh38
rs189665623 Chr.17: 36555080-36555080 on Build GRCh38
rs993024841 Chr.17: 36555064-36555064 on Build GRCh38
rs927407368 Chr.17: 36554983-36554983 on Build GRCh38
rs763575027 Chr.17: 36554911-36554911 on Build GRCh38
rs140552410 Chr.17: 36554855-36554855 on Build GRCh38
rs938672960 Chr.17: 36555005-36555005 on Build GRCh38
rs1044485137 Chr.17: 36554981-36554981 on Build GRCh38
rs920325855 Chr.17: 36555094-36555094 on Build GRCh38
rs371287129 Chr.17: 36554800-36554800 on Build GRCh38
rs908061897 Chr.17: 36554782-36554782 on Build GRCh38
rs779093465 Chr.17: 36554947-36554947 on Build GRCh38
rs576131318 Chr.17: 36555151-36555151 on Build GRCh38
rs1015154911 Chr.17: 36555068-36555068 on Build GRCh38
rs147217108 Chr.17: 36554852-36554852 on Build GRCh38
rs201262222 Chr.17: 36554806-36554806 on Build GRCh38
rs915090042 Chr.17: 36554953-36554953 on Build GRCh38
rs780521606 Chr.17: 36554927-36554927 on Build GRCh38
rs550800243 Chr.17: 36554938-36554938 on Build GRCh38
rs759267341 Chr.17: 36554856-36554856 on Build GRCh38
rs947917235 Chr.17: 36554970-36554970 on Build GRCh38
rs1005141428 Chr.17: 36555048-36555048 on Build GRCh38
rs199675142 Chr.17: 36554801-36554801 on Build GRCh38
rs764761789 Chr.17: 36554969-36554970 on Build GRCh38
rs892292095 Chr.17: 36554954-36554954 on Build GRCh38
rs754298440 Chr.17: 36554803-36554804 on Build GRCh38
rs963424429 Chr.17: 36554755-36554755 on Build GRCh38
rs989682122 Chr.17: 36554932-36554932 on Build GRCh38
rs60995670 Chr.17: 36554975-36554977 on Build GRCh38
rs781590045 Chr.17: 36554775-36554775 on Build GRCh38
rs985769952 Chr.17: 36555119-36555119 on Build GRCh38
rs756650514 Chr.17: 36554779-36554780 on Build GRCh38
rs746908543 Chr.17: 36554825-36554825 on Build GRCh38
rs202169466 Chr.17: 36554812-36554812 on Build GRCh38
rs770807947 Chr.17: 36554826-36554826 on Build GRCh38
rs547651645 Chr.17: 36555009-36555009 on Build GRCh38
rs775119796 Chr.17: 36554866-36554866 on Build GRCh38
rs879475508 Chr.17: 36555107-36555107 on Build GRCh38
rs764880764 Chr.17: 36554865-36554865 on Build GRCh38
rs570734410 Chr.17: 36555043-36555043 on Build GRCh38
rs534231132 Chr.17: 36554955-36554955 on Build GRCh38
rs757812813 Chr.17: 36554771-36554771 on Build GRCh38
rs1034329127 Chr.17: 36555022-36555022 on Build GRCh38
rs746239401 Chr.17: 36554778-36554778 on Build GRCh38
rs556465923 Chr.17: 36555124-36555124 on Build GRCh38
rs555342673 Chr.17: 36555181-36555181 on Build GRCh38
rs886669530 Chr.17: 36555013-36555013 on Build GRCh38
rs749495093 Chr.17: 36554940-36554940 on Build GRCh38
rs956625202 Chr.17: 36554917-36554917 on Build GRCh38
rs960250030 Chr.17: 36555028-36555028 on Build GRCh38
rs911574068 Chr.17: 36555134-36555134 on Build GRCh38
rs996772162 Chr.17: 36555176-36555176 on Build GRCh38
rs535199154 Chr.17: 36555177-36555177 on Build GRCh38
rs944625144 Chr.17: 36554919-36554919 on Build GRCh38
rs752048833 Chr.17: 36554770-36554770 on Build GRCh38
rs764566406 Chr.17: 36554789-36554792 on Build GRCh38
rs995932033 Chr.17: 36554796-36554796 on Build GRCh38
rs756429405 Chr.17: 36554920-36554920 on Build GRCh38
rs755288300 Chr.17: 36554944-36554944 on Build GRCh38
rs762652222 Chr.17: 36554888-36554888 on Build GRCh38
rs751111708 Chr.17: 36554915-36554915 on Build GRCh38
rs767921598 Chr.17: 36554916-36554916 on Build GRCh38
rs138223976 Chr.17: 36554972-36554975 on Build GRCh38
rs111254322 Chr.17: 36554858-36554858 on Build GRCh38
rs768839857 Chr.17: 36554809-36554809 on Build GRCh38

View Full Product Details