Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65538104-65538600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN2
Entrez Gene ID
8313
Gene Name
axin 2
Gene Aliases
AXIL, ODCRCS
UniGene
Hs.156527
Gene Chromosome Location
Chr.17: 65528563-65561622 on Build GRCh38
Transcripts
NM_004655.3 XM_011525319.2 XM_011525320.1 XM_011525321.2 XM_017025192.1 XM_017025193.1 XM_017025194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6975862 c.1100_1101CC>TT p.P367L Chr.17: 65538302-65538303 on Build GRCh38
4922321 c.1195C>T p.R399* Chr.17: 65538208-65538208 on Build GRCh38
6427420 c.1191G>T p.Q397H Chr.17: 65538212-65538212 on Build GRCh38
6081720 c.1102G>C p.A368P Chr.17: 65538301-65538301 on Build GRCh38
2796708 c.1092C>T p.P364P Chr.17: 65538311-65538311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022922128 Chr.17: 65538578-65538578 on Build GRCh38
rs775361721 Chr.17: 65538164-65538164 on Build GRCh38
rs776078682 Chr.17: 65538180-65538182 on Build GRCh38
rs929148280 Chr.17: 65538541-65538541 on Build GRCh38
rs941383163 Chr.17: 65538519-65538519 on Build GRCh38
rs561118074 Chr.17: 65538127-65538127 on Build GRCh38
rs545941909 Chr.17: 65538186-65538186 on Build GRCh38
rs776245264 Chr.17: 65538169-65538169 on Build GRCh38
rs914102463 Chr.17: 65538216-65538216 on Build GRCh38
rs775302620 Chr.17: 65538147-65538147 on Build GRCh38
rs778900546 Chr.17: 65538234-65538234 on Build GRCh38
rs769380091 Chr.17: 65538198-65538198 on Build GRCh38
rs999344211 Chr.17: 65538544-65538544 on Build GRCh38
rs761352798 Chr.17: 65538232-65538237 on Build GRCh38
rs568377447 Chr.17: 65538495-65538495 on Build GRCh38
rs898166595 Chr.17: 65538297-65538297 on Build GRCh38
rs4541111 Chr.17: 65538420-65538420 on Build GRCh38
rs117258157 Chr.17: 65538185-65538185 on Build GRCh38
rs906309952 Chr.17: 65538342-65538342 on Build GRCh38
rs1008151527 Chr.17: 65538516-65538516 on Build GRCh38
rs201104922 Chr.17: 65538385-65538385 on Build GRCh38
rs189213354 Chr.17: 65538124-65538124 on Build GRCh38
rs200899695 Chr.17: 65538222-65538222 on Build GRCh38
rs749323050 Chr.17: 65538190-65538190 on Build GRCh38
rs771831650 Chr.17: 65538162-65538162 on Build GRCh38
rs781583038 Chr.17: 65538133-65538133 on Build GRCh38
rs774207836 Chr.17: 65538152-65538160 on Build GRCh38
rs373945507 Chr.17: 65538153-65538153 on Build GRCh38
rs774405682 Chr.17: 65538140-65538140 on Build GRCh38
rs940164178 Chr.17: 65538421-65538421 on Build GRCh38
rs747959907 Chr.17: 65538196-65538196 on Build GRCh38
rs202063197 Chr.17: 65538165-65538165 on Build GRCh38
rs781106400 Chr.17: 65538326-65538326 on Build GRCh38
rs878854717 Chr.17: 65538258-65538258 on Build GRCh38
rs201451100 Chr.17: 65538338-65538338 on Build GRCh38
rs146249973 Chr.17: 65538324-65538324 on Build GRCh38
rs781122679 Chr.17: 65538239-65538239 on Build GRCh38
rs948958592 Chr.17: 65538209-65538209 on Build GRCh38
rs886053273 Chr.17: 65538241-65538241 on Build GRCh38
rs538414516 Chr.17: 65538135-65538135 on Build GRCh38
rs989632217 Chr.17: 65538205-65538205 on Build GRCh38
rs922684515 Chr.17: 65538224-65538224 on Build GRCh38
rs376505336 Chr.17: 65538353-65538353 on Build GRCh38
rs770964261 Chr.17: 65538132-65538133 on Build GRCh38
rs748698260 Chr.17: 65538314-65538314 on Build GRCh38
rs990634803 Chr.17: 65538577-65538577 on Build GRCh38
rs777937769 Chr.17: 65538129-65538130 on Build GRCh38
rs748426955 Chr.17: 65538164-65538164 on Build GRCh38
rs746967147 Chr.17: 65538193-65538193 on Build GRCh38
rs1032575034 Chr.17: 65538554-65538554 on Build GRCh38
rs755305827 Chr.17: 65538162-65538162 on Build GRCh38
rs919183538 Chr.17: 65538526-65538526 on Build GRCh38
rs749415145 Chr.17: 65538132-65538155 on Build GRCh38
rs985464151 Chr.17: 65538489-65538489 on Build GRCh38
rs551939302 Chr.17: 65538136-65538136 on Build GRCh38
rs760263280 Chr.17: 65538166-65538167 on Build GRCh38
rs8081536 Chr.17: 65538333-65538333 on Build GRCh38
rs771763530 Chr.17: 65538366-65538366 on Build GRCh38
rs754059595 Chr.17: 65538308-65538308 on Build GRCh38
rs116931989 Chr.17: 65538184-65538184 on Build GRCh38
rs772636882 Chr.17: 65538301-65538301 on Build GRCh38
rs376052287 Chr.17: 65538227-65538227 on Build GRCh38
rs373087123 Chr.17: 65538220-65538220 on Build GRCh38
rs909858027 Chr.17: 65538497-65538497 on Build GRCh38
rs756352400 Chr.17: 65538187-65538187 on Build GRCh38
rs145022269 Chr.17: 65538167-65538167 on Build GRCh38
rs949010391 Chr.17: 65538407-65538407 on Build GRCh38
rs762522074 Chr.17: 65538204-65538204 on Build GRCh38
rs574302210 Chr.17: 65538171-65538171 on Build GRCh38
rs778629171 Chr.17: 65538362-65538362 on Build GRCh38
rs764407281 Chr.17: 65538230-65538230 on Build GRCh38
rs533790501 Chr.17: 65538172-65538172 on Build GRCh38
rs35399989 Chr.17: 65538360-65538360 on Build GRCh38
rs750414544 Chr.17: 65538317-65538317 on Build GRCh38
rs770644676 Chr.17: 65538279-65538279 on Build GRCh38
rs8078753 Chr.17: 65538132-65538132 on Build GRCh38
rs375277654 Chr.17: 65538160-65538160 on Build GRCh38
rs765865516 Chr.17: 65538207-65538207 on Build GRCh38
rs957509903 Chr.17: 65538571-65538571 on Build GRCh38
rs763758748 Chr.17: 65538171-65538172 on Build GRCh38
rs765518896 Chr.17: 65538311-65538311 on Build GRCh38
rs761901627 Chr.17: 65538310-65538310 on Build GRCh38
rs764414708 Chr.17: 65538306-65538306 on Build GRCh38
rs730881403 Chr.17: 65538184-65538185 on Build GRCh38
rs760399682 Chr.17: 65538148-65538148 on Build GRCh38
rs765441834 Chr.17: 65538229-65538229 on Build GRCh38
rs753049092 Chr.17: 65538223-65538223 on Build GRCh38
rs149089442 Chr.17: 65538384-65538384 on Build GRCh38
rs1037206684 Chr.17: 65538431-65538431 on Build GRCh38
rs775294617 Chr.17: 65538166-65538166 on Build GRCh38
rs757216374 Chr.17: 65538436-65538436 on Build GRCh38
rs774887154 Chr.17: 65538213-65538213 on Build GRCh38
rs540249008 Chr.17: 65538356-65538356 on Build GRCh38
rs751897827 Chr.17: 65538157-65538157 on Build GRCh38
rs769237631 Chr.17: 65538390-65538390 on Build GRCh38
rs781562976 Chr.17: 65538163-65538163 on Build GRCh38
rs763735512 Chr.17: 65538149-65538149 on Build GRCh38
rs371063358 Chr.17: 65538254-65538254 on Build GRCh38
rs370461337 Chr.17: 65538192-65538192 on Build GRCh38
rs548155754 Chr.17: 65538451-65538451 on Build GRCh38
rs776118886 Chr.17: 65538371-65538371 on Build GRCh38
rs771929991 Chr.17: 65538144-65538144 on Build GRCh38
rs1021217900 Chr.17: 65538419-65538419 on Build GRCh38
rs761435975 Chr.17: 65538417-65538417 on Build GRCh38
rs902363610 Chr.17: 65538543-65538543 on Build GRCh38
rs755884266 Chr.17: 65538334-65538334 on Build GRCh38
rs776372607 Chr.17: 65538195-65538195 on Build GRCh38
rs549161295 Chr.17: 65538154-65538154 on Build GRCh38
rs774998353 Chr.17: 65538511-65538511 on Build GRCh38
rs75747149 Chr.17: 65538249-65538249 on Build GRCh38
rs761105129 Chr.17: 65538305-65538305 on Build GRCh38
rs745366441 Chr.17: 65538365-65538365 on Build GRCh38
rs759364903 Chr.17: 65538156-65538160 on Build GRCh38
rs139871607 Chr.17: 65538235-65538235 on Build GRCh38
rs114256284 Chr.17: 65538408-65538408 on Build GRCh38
rs370821074 Chr.17: 65538231-65538231 on Build GRCh38
rs530046280 Chr.17: 65538128-65538128 on Build GRCh38
rs889063871 Chr.17: 65538152-65538152 on Build GRCh38
rs768410635 Chr.17: 65538194-65538194 on Build GRCh38
rs748945736 Chr.17: 65538278-65538278 on Build GRCh38
rs957620918 Chr.17: 65538367-65538367 on Build GRCh38
rs759584945 Chr.17: 65538141-65538141 on Build GRCh38
rs898266856 Chr.17: 65538432-65538432 on Build GRCh38
rs772748458 Chr.17: 65538203-65538203 on Build GRCh38
rs375906660 Chr.17: 65538152-65538152 on Build GRCh38
rs759917468 Chr.17: 65538217-65538217 on Build GRCh38
rs1019510770 Chr.17: 65538534-65538534 on Build GRCh38
rs1003920705 Chr.17: 65538248-65538248 on Build GRCh38
rs759128363 Chr.17: 65538298-65538298 on Build GRCh38
rs368502813 Chr.17: 65538240-65538240 on Build GRCh38
rs780449193 Chr.17: 65538156-65538156 on Build GRCh38
rs766847390 Chr.17: 65538183-65538183 on Build GRCh38
rs772908702 Chr.17: 65538276-65538276 on Build GRCh38
rs1037542566 Chr.17: 65538521-65538521 on Build GRCh38
rs771628301 Chr.17: 65538452-65538452 on Build GRCh38
rs762386143 Chr.17: 65538175-65538175 on Build GRCh38
rs78028780 Chr.17: 65538437-65538437 on Build GRCh38
rs372767920 Chr.17: 65538232-65538232 on Build GRCh38
rs141697521 Chr.17: 65538302-65538302 on Build GRCh38

View Full Product Details