Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67055025-67055520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG1
Entrez Gene ID
786
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Gene Aliases
CACNLG
UniGene
Hs.147989
Gene Chromosome Location
Chr.17: 67044536-67056797 on Build GRCh38
Transcripts
NM_000727.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5430552 c.411G>A p.L137L Chr.17: 67055209-67055209 on Build GRCh38
3691740 c.373G>A p.V125I Chr.17: 67055171-67055171 on Build GRCh38
5475959 c.331G>A p.A111T Chr.17: 67055129-67055129 on Build GRCh38
3672539 c.417C>A p.P139P Chr.17: 67055215-67055215 on Build GRCh38
371341 c.418G>A p.A140T Chr.17: 67055216-67055216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755800128 Chr.17: 67055278-67055278 on Build GRCh38
rs758183813 Chr.17: 67055139-67055139 on Build GRCh38
rs763270909 Chr.17: 67055092-67055092 on Build GRCh38
rs370298645 Chr.17: 67055055-67055055 on Build GRCh38
rs113473709 Chr.17: 67055392-67055392 on Build GRCh38
rs758336354 Chr.17: 67055209-67055209 on Build GRCh38
rs775968110 Chr.17: 67055217-67055218 on Build GRCh38
rs921123953 Chr.17: 67055115-67055115 on Build GRCh38
rs746596639 Chr.17: 67055157-67055157 on Build GRCh38
rs539674475 Chr.17: 67055330-67055330 on Build GRCh38
rs757194613 Chr.17: 67055127-67055127 on Build GRCh38
rs1013688034 Chr.17: 67055160-67055160 on Build GRCh38
rs60144522 Chr.17: 67055287-67055287 on Build GRCh38
rs779836809 Chr.17: 67055153-67055153 on Build GRCh38
rs1019784823 Chr.17: 67055395-67055395 on Build GRCh38
rs752842571 Chr.17: 67055126-67055126 on Build GRCh38
rs1036627342 Chr.17: 67055150-67055150 on Build GRCh38
rs1019508032 Chr.17: 67055272-67055272 on Build GRCh38
rs138106706 Chr.17: 67055322-67055322 on Build GRCh38
rs776803189 Chr.17: 67055265-67055265 on Build GRCh38
rs112984376 Chr.17: 67055268-67055268 on Build GRCh38
rs1045321561 Chr.17: 67055367-67055367 on Build GRCh38
rs1009671139 Chr.17: 67055403-67055403 on Build GRCh38
rs747662157 Chr.17: 67055165-67055165 on Build GRCh38
rs1041413494 Chr.17: 67055412-67055412 on Build GRCh38
rs780632218 Chr.17: 67055164-67055164 on Build GRCh38
rs775606926 Chr.17: 67055196-67055196 on Build GRCh38
rs557319403 Chr.17: 67055181-67055181 on Build GRCh38
rs751379455 Chr.17: 67055215-67055215 on Build GRCh38
rs949635768 Chr.17: 67055358-67055358 on Build GRCh38
rs746272550 Chr.17: 67055253-67055253 on Build GRCh38
rs749694779 Chr.17: 67055233-67055233 on Build GRCh38
rs767896301 Chr.17: 67055188-67055188 on Build GRCh38
rs762829834 Chr.17: 67055267-67055267 on Build GRCh38
rs750267789 Chr.17: 67055203-67055203 on Build GRCh38
rs753434139 Chr.17: 67055286-67055286 on Build GRCh38
rs139399934 Chr.17: 67055237-67055237 on Build GRCh38
rs766706963 Chr.17: 67055099-67055099 on Build GRCh38
rs142384274 Chr.17: 67055186-67055186 on Build GRCh38
rs140836897 Chr.17: 67055179-67055179 on Build GRCh38
rs147258815 Chr.17: 67055171-67055171 on Build GRCh38
rs2363844 Chr.17: 67055064-67055064 on Build GRCh38
rs772416293 Chr.17: 67055255-67055255 on Build GRCh38
rs765124047 Chr.17: 67055128-67055128 on Build GRCh38
rs926780920 Chr.17: 67055375-67055375 on Build GRCh38
rs773605479 Chr.17: 67055090-67055090 on Build GRCh38
rs1013389049 Chr.17: 67055320-67055320 on Build GRCh38
rs374291670 Chr.17: 67055104-67055104 on Build GRCh38
rs896754789 Chr.17: 67055211-67055211 on Build GRCh38
rs775462666 Chr.17: 67055390-67055390 on Build GRCh38
rs143842742 Chr.17: 67055161-67055161 on Build GRCh38
rs375516186 Chr.17: 67055288-67055288 on Build GRCh38
rs779746872 Chr.17: 67055059-67055059 on Build GRCh38
rs1032782255 Chr.17: 67055350-67055350 on Build GRCh38
rs112631391 Chr.17: 67055236-67055236 on Build GRCh38
rs887724099 Chr.17: 67055144-67055144 on Build GRCh38
rs78733538 Chr.17: 67055391-67055391 on Build GRCh38
rs774863748 Chr.17: 67055185-67055185 on Build GRCh38
rs770225370 Chr.17: 67055083-67055083 on Build GRCh38
rs75667121 Chr.17: 67055485-67055485 on Build GRCh38
rs765206216 Chr.17: 67055198-67055198 on Build GRCh38
rs111690102 Chr.17: 67055274-67055274 on Build GRCh38
rs376425251 Chr.17: 67055073-67055073 on Build GRCh38
rs983830129 Chr.17: 67055300-67055300 on Build GRCh38
rs768096058 Chr.17: 67055071-67055071 on Build GRCh38
rs747693009 Chr.17: 67055220-67055220 on Build GRCh38
rs200335785 Chr.17: 67055122-67055122 on Build GRCh38
rs777111454 Chr.17: 67055283-67055283 on Build GRCh38
rs771721845 Chr.17: 67055053-67055053 on Build GRCh38
rs931600754 Chr.17: 67055116-67055116 on Build GRCh38
rs745696355 Chr.17: 67055052-67055052 on Build GRCh38
rs369953732 Chr.17: 67055074-67055074 on Build GRCh38
rs373779390 Chr.17: 67055273-67055273 on Build GRCh38
rs528510949 Chr.17: 67055426-67055426 on Build GRCh38
rs766121837 Chr.17: 67055210-67055210 on Build GRCh38
rs952537308 Chr.17: 67055476-67055476 on Build GRCh38
rs896340966 Chr.17: 67055301-67055301 on Build GRCh38
rs1000956594 Chr.17: 67055331-67055331 on Build GRCh38
rs944483952 Chr.17: 67055075-67055075 on Build GRCh38
rs746482808 Chr.17: 67055241-67055241 on Build GRCh38
rs1004798731 Chr.17: 67055146-67055146 on Build GRCh38
rs376840748 Chr.17: 67055129-67055129 on Build GRCh38
rs1036249185 Chr.17: 67055299-67055299 on Build GRCh38
rs570829074 Chr.17: 67055421-67055421 on Build GRCh38
rs975585681 Chr.17: 67055432-67055432 on Build GRCh38
rs772739042 Chr.17: 67055215-67055216 on Build GRCh38
rs936856868 Chr.17: 67055380-67055380 on Build GRCh38
rs151308706 Chr.17: 67055218-67055218 on Build GRCh38
rs997167244 Chr.17: 67055427-67055427 on Build GRCh38
rs746219371 Chr.17: 67055289-67055289 on Build GRCh38
rs916935298 Chr.17: 67055357-67055357 on Build GRCh38
rs770305526 Chr.17: 67055170-67055170 on Build GRCh38
rs575886291 Chr.17: 67055216-67055216 on Build GRCh38
rs558109810 Chr.17: 67055257-67055257 on Build GRCh38
rs201686961 Chr.17: 67055264-67055264 on Build GRCh38
rs72848810 Chr.17: 67055447-67055447 on Build GRCh38
rs111459753 Chr.17: 67055252-67055252 on Build GRCh38
rs755639468 Chr.17: 67055229-67055229 on Build GRCh38
rs767660766 Chr.17: 67055123-67055123 on Build GRCh38

View Full Product Details