Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67073901-67074412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELZ
Entrez Gene ID
9931
Gene Name
helicase with zinc finger
Gene Aliases
DHRC, DRHC, HUMORF5
UniGene
Hs.370140
Gene Chromosome Location
Chr.17: 67070438-67245989 on Build GRCh38
Transcripts
NM_014877.3 XM_005257888.4 XM_005257889.4 XM_006722214.3 XM_006722215.3 XM_006722216.3 XM_011525544.2 XM_017025477.1 XM_017025478.1

SNP Detail

SNP ID Chromosome Location
rs914212607 Chr.17: 67073994-67073994 on Build GRCh38
rs745805706 Chr.17: 67074235-67074235 on Build GRCh38
rs775411887 Chr.17: 67074335-67074335 on Build GRCh38
rs1024964741 Chr.17: 67073968-67073968 on Build GRCh38
rs1041742995 Chr.17: 67074191-67074191 on Build GRCh38
rs1050177476 Chr.17: 67074035-67074035 on Build GRCh38
rs989926455 Chr.17: 67074146-67074146 on Build GRCh38
rs397728193 Chr.17: 67074262-67074263 on Build GRCh38
rs777900490 Chr.17: 67074108-67074108 on Build GRCh38
rs147958301 Chr.17: 67074331-67074331 on Build GRCh38
rs377679661 Chr.17: 67073935-67073935 on Build GRCh38
rs778938298 Chr.17: 67074206-67074206 on Build GRCh38
rs926752476 Chr.17: 67074112-67074112 on Build GRCh38
rs1033917280 Chr.17: 67074378-67074378 on Build GRCh38
rs932269113 Chr.17: 67074305-67074305 on Build GRCh38
rs1033569019 Chr.17: 67074216-67074216 on Build GRCh38
rs1034031962 Chr.17: 67074055-67074055 on Build GRCh38
rs1021242764 Chr.17: 67074261-67074262 on Build GRCh38
rs997368523 Chr.17: 67074020-67074020 on Build GRCh38
rs922356596 Chr.17: 67074303-67074303 on Build GRCh38
rs989326932 Chr.17: 67074257-67074258 on Build GRCh38
rs540403107 Chr.17: 67074144-67074144 on Build GRCh38
rs142003678 Chr.17: 67073930-67073930 on Build GRCh38
rs545933305 Chr.17: 67073998-67073998 on Build GRCh38
rs933210535 Chr.17: 67074207-67074207 on Build GRCh38
rs922294244 Chr.17: 67074140-67074140 on Build GRCh38
rs976524040 Chr.17: 67074268-67074268 on Build GRCh38
rs749350854 Chr.17: 67074145-67074145 on Build GRCh38
rs530523228 Chr.17: 67074364-67074364 on Build GRCh38
rs34711760 Chr.17: 67074179-67074180 on Build GRCh38
rs1006235450 Chr.17: 67074057-67074057 on Build GRCh38
rs958198465 Chr.17: 67074252-67074252 on Build GRCh38
rs958455962 Chr.17: 67074132-67074132 on Build GRCh38
rs942170921 Chr.17: 67074174-67074174 on Build GRCh38
rs971021094 Chr.17: 67074357-67074357 on Build GRCh38
rs144887939 Chr.17: 67074034-67074034 on Build GRCh38
rs914306025 Chr.17: 67074148-67074148 on Build GRCh38
rs976409793 Chr.17: 67074120-67074120 on Build GRCh38
rs1025354654 Chr.17: 67074355-67074355 on Build GRCh38
rs570593823 Chr.17: 67074324-67074324 on Build GRCh38
rs74885885 Chr.17: 67074063-67074063 on Build GRCh38
rs537508678 Chr.17: 67074070-67074070 on Build GRCh38
rs901749940 Chr.17: 67074019-67074019 on Build GRCh38
rs1041657788 Chr.17: 67074001-67074001 on Build GRCh38
rs190063852 Chr.17: 67073997-67073997 on Build GRCh38
rs897900393 Chr.17: 67074189-67074189 on Build GRCh38
rs971353177 Chr.17: 67074090-67074090 on Build GRCh38
rs889218627 Chr.17: 67074038-67074038 on Build GRCh38
rs9902312 Chr.17: 67074188-67074188 on Build GRCh38
rs535763218 Chr.17: 67073950-67073950 on Build GRCh38
rs888979159 Chr.17: 67074283-67074283 on Build GRCh38
rs11390294 Chr.17: 67074251-67074252 on Build GRCh38
rs1025257584 Chr.17: 67074078-67074078 on Build GRCh38
rs202123754 Chr.17: 67074251-67074251 on Build GRCh38
rs1005994691 Chr.17: 67074297-67074297 on Build GRCh38
rs993905198 Chr.17: 67074072-67074072 on Build GRCh38
rs529507389 Chr.17: 67074244-67074244 on Build GRCh38
rs755471328 Chr.17: 67074248-67074249 on Build GRCh38
rs557039450 Chr.17: 67073965-67073965 on Build GRCh38
rs1024913834 Chr.17: 67074200-67074200 on Build GRCh38
rs1054140698 Chr.17: 67073981-67073981 on Build GRCh38
rs980678695 Chr.17: 67074031-67074031 on Build GRCh38

View Full Product Details