Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67075832-67076328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELZ
Entrez Gene ID
9931
Gene Name
helicase with zinc finger
Gene Aliases
DHRC, DRHC, HUMORF5
UniGene
Hs.370140
Gene Chromosome Location
Chr.17: 67070438-67245989 on Build GRCh38
Transcripts
NM_014877.3 XM_005257888.4 XM_005257889.4 XM_006722214.3 XM_006722215.3 XM_006722216.3 XM_011525544.2 XM_017025477.1 XM_017025478.1

SNP Detail

SNP ID Chromosome Location
rs182867496 Chr.17: 67076034-67076034 on Build GRCh38
rs978513658 Chr.17: 67075952-67075952 on Build GRCh38
rs149333451 Chr.17: 67076299-67076299 on Build GRCh38
rs1022116089 Chr.17: 67076216-67076216 on Build GRCh38
rs141959000 Chr.17: 67076215-67076215 on Build GRCh38
rs374660231 Chr.17: 67076043-67076043 on Build GRCh38
rs930382066 Chr.17: 67076083-67076083 on Build GRCh38
rs1022984924 Chr.17: 67076176-67076176 on Build GRCh38
rs990646055 Chr.17: 67075941-67075941 on Build GRCh38
rs768356827 Chr.17: 67075972-67075972 on Build GRCh38
rs1034919146 Chr.17: 67075876-67075876 on Build GRCh38
rs12948934 Chr.17: 67076129-67076129 on Build GRCh38
rs968186065 Chr.17: 67076180-67076181 on Build GRCh38
rs1046886086 Chr.17: 67076300-67076300 on Build GRCh38
rs919854775 Chr.17: 67076260-67076260 on Build GRCh38
rs1031484103 Chr.17: 67076254-67076254 on Build GRCh38
rs895031285 Chr.17: 67076175-67076175 on Build GRCh38
rs1046773456 Chr.17: 67076119-67076119 on Build GRCh38
rs776285925 Chr.17: 67075992-67075992 on Build GRCh38
rs898356515 Chr.17: 67076061-67076061 on Build GRCh38
rs533210240 Chr.17: 67076186-67076186 on Build GRCh38
rs1031058344 Chr.17: 67076229-67076229 on Build GRCh38
rs968526606 Chr.17: 67075894-67075894 on Build GRCh38
rs200301754 Chr.17: 67075859-67075859 on Build GRCh38
rs144853193 Chr.17: 67076257-67076257 on Build GRCh38
rs12949723 Chr.17: 67076138-67076138 on Build GRCh38
rs1047922805 Chr.17: 67076099-67076099 on Build GRCh38
rs187907355 Chr.17: 67076170-67076170 on Build GRCh38
rs138790322 Chr.17: 67076158-67076158 on Build GRCh38
rs748563036 Chr.17: 67076275-67076276 on Build GRCh38
rs191691559 Chr.17: 67075853-67075853 on Build GRCh38
rs987035316 Chr.17: 67076030-67076030 on Build GRCh38
rs886130127 Chr.17: 67076154-67076154 on Build GRCh38
rs898912188 Chr.17: 67076059-67076059 on Build GRCh38
rs942905282 Chr.17: 67076039-67076039 on Build GRCh38
rs978227494 Chr.17: 67076244-67076244 on Build GRCh38
rs12939278 Chr.17: 67076202-67076202 on Build GRCh38
rs928387643 Chr.17: 67076301-67076301 on Build GRCh38
rs933970725 Chr.17: 67075974-67075974 on Build GRCh38
rs964291244 Chr.17: 67076256-67076256 on Build GRCh38
rs986806683 Chr.17: 67075976-67075976 on Build GRCh38
rs138768651 Chr.17: 67076289-67076289 on Build GRCh38
rs1055506835 Chr.17: 67076156-67076156 on Build GRCh38
rs12938479 Chr.17: 67076146-67076146 on Build GRCh38
rs376548269 Chr.17: 67076048-67076048 on Build GRCh38
rs911470518 Chr.17: 67076031-67076031 on Build GRCh38
rs911121626 Chr.17: 67075986-67075986 on Build GRCh38
rs747757567 Chr.17: 67076235-67076235 on Build GRCh38
rs73340967 Chr.17: 67075938-67075938 on Build GRCh38
rs12941154 Chr.17: 67076270-67076270 on Build GRCh38
rs1022398848 Chr.17: 67075949-67075949 on Build GRCh38
rs369777814 Chr.17: 67075951-67075951 on Build GRCh38
rs550721096 Chr.17: 67076070-67076070 on Build GRCh38
rs1038592028 Chr.17: 67075992-67075997 on Build GRCh38

View Full Product Details