Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67992822-67993322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf58
Entrez Gene ID
284018
Gene Name
chromosome 17 open reading frame 58
Gene Aliases
-
UniGene
Hs.90790
Gene Chromosome Location
Chr.17: 67991099-67993649 on Build GRCh38
Transcripts
NM_181655.3 NM_181656.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1324880 c.41G>A p.R14Q Chr.17: 67993106-67993106 on Build GRCh38
1385442 c.208C>T p.R70* Chr.17: 67992939-67992939 on Build GRCh38
1324881 c.41G>A p.R14Q Chr.17: 67993106-67993106 on Build GRCh38
1385444 c.24delC p.D9fs*10 Chr.17: 67993123-67993123 on Build GRCh38
347068 c.77A>G p.N26S Chr.17: 67993070-67993070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782673283 Chr.17: 67993128-67993128 on Build GRCh38
rs782192091 Chr.17: 67992939-67992939 on Build GRCh38
rs782420116 Chr.17: 67993167-67993167 on Build GRCh38
rs782409920 Chr.17: 67993066-67993066 on Build GRCh38
rs782335135 Chr.17: 67993070-67993070 on Build GRCh38
rs112612674 Chr.17: 67992964-67992964 on Build GRCh38
rs540904686 Chr.17: 67993222-67993222 on Build GRCh38
rs374827455 Chr.17: 67993276-67993276 on Build GRCh38
rs782141144 Chr.17: 67992891-67992891 on Build GRCh38
rs376022307 Chr.17: 67993029-67993029 on Build GRCh38
rs782563524 Chr.17: 67992936-67992936 on Build GRCh38
rs782323911 Chr.17: 67993077-67993077 on Build GRCh38
rs557724927 Chr.17: 67993161-67993161 on Build GRCh38
rs782132500 Chr.17: 67993182-67993182 on Build GRCh38
rs781968319 Chr.17: 67993194-67993194 on Build GRCh38
rs781939369 Chr.17: 67992972-67992972 on Build GRCh38
rs532559674 Chr.17: 67992858-67992859 on Build GRCh38
rs370454631 Chr.17: 67993076-67993076 on Build GRCh38
rs782137706 Chr.17: 67992895-67992895 on Build GRCh38
rs782525838 Chr.17: 67992940-67992940 on Build GRCh38
rs781794923 Chr.17: 67993127-67993127 on Build GRCh38
rs190313223 Chr.17: 67992921-67992921 on Build GRCh38
rs373219606 Chr.17: 67993028-67993028 on Build GRCh38
rs528575640 Chr.17: 67992880-67992880 on Build GRCh38
rs782204107 Chr.17: 67993065-67993065 on Build GRCh38
rs192574029 Chr.17: 67993055-67993055 on Build GRCh38
rs201482119 Chr.17: 67992890-67992890 on Build GRCh38
rs782101326 Chr.17: 67992944-67992945 on Build GRCh38
rs782508814 Chr.17: 67992910-67992910 on Build GRCh38
rs557376202 Chr.17: 67993025-67993025 on Build GRCh38
rs568233827 Chr.17: 67992960-67992960 on Build GRCh38
rs781816917 Chr.17: 67992926-67992926 on Build GRCh38
rs782761075 Chr.17: 67993126-67993126 on Build GRCh38
rs782559467 Chr.17: 67992922-67992922 on Build GRCh38
rs782248344 Chr.17: 67993069-67993069 on Build GRCh38
rs781992440 Chr.17: 67993000-67993000 on Build GRCh38
rs782629024 Chr.17: 67992914-67992914 on Build GRCh38
rs782149344 Chr.17: 67992996-67992996 on Build GRCh38
rs782235077 Chr.17: 67992966-67992966 on Build GRCh38
rs368917203 Chr.17: 67993164-67993164 on Build GRCh38
rs782078172 Chr.17: 67993008-67993008 on Build GRCh38
rs781985963 Chr.17: 67992893-67992893 on Build GRCh38
rs782690381 Chr.17: 67992949-67992949 on Build GRCh38
rs782214443 Chr.17: 67993047-67993047 on Build GRCh38
rs782491175 Chr.17: 67993054-67993054 on Build GRCh38
rs782771037 Chr.17: 67992925-67992925 on Build GRCh38
rs79090657 Chr.17: 67992856-67992856 on Build GRCh38
rs782643332 Chr.17: 67993041-67993041 on Build GRCh38
rs782081818 Chr.17: 67993115-67993115 on Build GRCh38
rs782753773 Chr.17: 67993102-67993102 on Build GRCh38
rs782164024 Chr.17: 67993098-67993098 on Build GRCh38
rs782313253 Chr.17: 67992971-67992971 on Build GRCh38
rs782053503 Chr.17: 67993195-67993195 on Build GRCh38
rs28368756 Chr.17: 67992932-67992932 on Build GRCh38
rs777605198 Chr.17: 67992863-67992863 on Build GRCh38
rs782521219 Chr.17: 67993142-67993142 on Build GRCh38
rs781960314 Chr.17: 67993074-67993074 on Build GRCh38
rs782731023 Chr.17: 67992906-67992906 on Build GRCh38
rs782807885 Chr.17: 67993138-67993139 on Build GRCh38
rs781910509 Chr.17: 67993137-67993137 on Build GRCh38
rs782673638 Chr.17: 67993017-67993017 on Build GRCh38
rs782596661 Chr.17: 67992876-67992876 on Build GRCh38
rs539583221 Chr.17: 67993085-67993085 on Build GRCh38
rs782737968 Chr.17: 67993009-67993009 on Build GRCh38
rs781987574 Chr.17: 67993171-67993171 on Build GRCh38
rs111513651 Chr.17: 67993106-67993106 on Build GRCh38
rs782041315 Chr.17: 67993067-67993067 on Build GRCh38
rs201608189 Chr.17: 67992892-67992892 on Build GRCh38
rs374492199 Chr.17: 67993124-67993124 on Build GRCh38
rs782628571 Chr.17: 67993291-67993291 on Build GRCh38
rs782220822 Chr.17: 67993158-67993158 on Build GRCh38
rs201853045 Chr.17: 67993015-67993015 on Build GRCh38
rs116663321 Chr.17: 67993197-67993197 on Build GRCh38
rs782648173 Chr.17: 67993156-67993156 on Build GRCh38
rs552213460 Chr.17: 67992888-67992888 on Build GRCh38
rs781877517 Chr.17: 67993092-67993093 on Build GRCh38
rs782278541 Chr.17: 67993090-67993090 on Build GRCh38
rs782810899 Chr.17: 67992998-67992998 on Build GRCh38
rs775961103 Chr.17: 67993248-67993248 on Build GRCh38
rs200385109 Chr.17: 67992932-67992933 on Build GRCh38
rs782384030 Chr.17: 67993163-67993163 on Build GRCh38
rs782343169 Chr.17: 67993188-67993188 on Build GRCh38
rs782391206 Chr.17: 67992959-67992959 on Build GRCh38
rs377529814 Chr.17: 67992958-67992958 on Build GRCh38

View Full Product Details