Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68976043-68976536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA9
Entrez Gene ID
10350
Gene Name
ATP binding cassette subfamily A member 9
Gene Aliases
EST640918
UniGene
Hs.131686
Gene Chromosome Location
Chr.17: 68974632-69078978 on Build GRCh38
Transcripts
NM_080283.3 XM_017024011.1 XM_017024012.1 XM_017024013.1 XM_017024014.1 XM_017024015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736975 c.4768C>G p.L1590V Chr.17: 68976143-68976143 on Build GRCh38
3796003 c.4755C>A p.L1585L Chr.17: 68976156-68976156 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752217391 Chr.17: 68976165-68976165 on Build GRCh38
rs775185404 Chr.17: 68976223-68976223 on Build GRCh38
rs779274342 Chr.17: 68976141-68976142 on Build GRCh38
rs781095186 Chr.17: 68976211-68976211 on Build GRCh38
rs371641163 Chr.17: 68976214-68976214 on Build GRCh38
rs898630569 Chr.17: 68976382-68976382 on Build GRCh38
rs766195842 Chr.17: 68976134-68976134 on Build GRCh38
rs764557816 Chr.17: 68976177-68976177 on Build GRCh38
rs754389074 Chr.17: 68976181-68976181 on Build GRCh38
rs981726371 Chr.17: 68976473-68976473 on Build GRCh38
rs772350148 Chr.17: 68976090-68976090 on Build GRCh38
rs751285933 Chr.17: 68976270-68976270 on Build GRCh38
rs370117036 Chr.17: 68976158-68976158 on Build GRCh38
rs1006890857 Chr.17: 68976350-68976350 on Build GRCh38
rs750611025 Chr.17: 68976201-68976202 on Build GRCh38
rs191069233 Chr.17: 68976505-68976505 on Build GRCh38
rs781122458 Chr.17: 68976233-68976234 on Build GRCh38
rs774192122 Chr.17: 68976217-68976217 on Build GRCh38
rs760195501 Chr.17: 68976233-68976233 on Build GRCh38
rs780315045 Chr.17: 68976207-68976207 on Build GRCh38
rs967930018 Chr.17: 68976415-68976415 on Build GRCh38
rs141443255 Chr.17: 68976303-68976303 on Build GRCh38
rs2191212 Chr.17: 68976109-68976109 on Build GRCh38
rs557745133 Chr.17: 68976270-68976270 on Build GRCh38
rs375689201 Chr.17: 68976193-68976193 on Build GRCh38
rs757797303 Chr.17: 68976184-68976184 on Build GRCh38
rs770799041 Chr.17: 68976215-68976215 on Build GRCh38
rs886206614 Chr.17: 68976268-68976268 on Build GRCh38
rs745767343 Chr.17: 68976478-68976478 on Build GRCh38
rs562062545 Chr.17: 68976471-68976471 on Build GRCh38
rs540159609 Chr.17: 68976438-68976438 on Build GRCh38
rs747110393 Chr.17: 68976208-68976208 on Build GRCh38
rs200512913 Chr.17: 68976104-68976104 on Build GRCh38
rs371765033 Chr.17: 68976233-68976233 on Build GRCh38
rs1025910103 Chr.17: 68976121-68976121 on Build GRCh38
rs1022230415 Chr.17: 68976388-68976388 on Build GRCh38
rs934710374 Chr.17: 68976078-68976078 on Build GRCh38
rs377482274 Chr.17: 68976380-68976380 on Build GRCh38
rs113897544 Chr.17: 68976416-68976416 on Build GRCh38
rs991614116 Chr.17: 68976497-68976497 on Build GRCh38
rs990461869 Chr.17: 68976335-68976335 on Build GRCh38
rs1051036417 Chr.17: 68976335-68976336 on Build GRCh38
rs116438216 Chr.17: 68976487-68976487 on Build GRCh38
rs867374319 Chr.17: 68976141-68976141 on Build GRCh38
rs557555372 Chr.17: 68976218-68976218 on Build GRCh38
rs369941926 Chr.17: 68976102-68976102 on Build GRCh38
rs565967725 Chr.17: 68976077-68976077 on Build GRCh38
rs185630117 Chr.17: 68976479-68976479 on Build GRCh38
rs959337776 Chr.17: 68976319-68976319 on Build GRCh38
rs768808084 Chr.17: 68976210-68976210 on Build GRCh38
rs183165331 Chr.17: 68976205-68976205 on Build GRCh38
rs769683893 Chr.17: 68976112-68976112 on Build GRCh38
rs148428186 Chr.17: 68976138-68976138 on Build GRCh38
rs1018366739 Chr.17: 68976381-68976381 on Build GRCh38
rs750757041 Chr.17: 68976199-68976199 on Build GRCh38
rs760200514 Chr.17: 68976174-68976174 on Build GRCh38
rs369471537 Chr.17: 68976223-68976224 on Build GRCh38
rs562568260 Chr.17: 68976504-68976504 on Build GRCh38
rs962912079 Chr.17: 68976096-68976096 on Build GRCh38
rs201699458 Chr.17: 68976222-68976222 on Build GRCh38
rs1006962998 Chr.17: 68976076-68976076 on Build GRCh38
rs762832147 Chr.17: 68976127-68976127 on Build GRCh38
rs780093669 Chr.17: 68976224-68976224 on Build GRCh38

View Full Product Details