Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68984773-68985281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA9
Entrez Gene ID
10350
Gene Name
ATP binding cassette subfamily A member 9
Gene Aliases
EST640918
UniGene
Hs.131686
Gene Chromosome Location
Chr.17: 68974632-69078978 on Build GRCh38
Transcripts
NM_080283.3 XM_017024011.1 XM_017024012.1 XM_017024013.1 XM_017024014.1 XM_017024015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776061 c.4341G>T p.P1447P Chr.17: 68984923-68984923 on Build GRCh38
238633 c.4375A>C p.M1459L Chr.17: 68984889-68984889 on Build GRCh38
242731 c.4317G>T p.P1439P Chr.17: 68984947-68984947 on Build GRCh38
6337623 c.4305C>A p.I1435I Chr.17: 68984959-68984959 on Build GRCh38
6334317 c.4315C>A p.P1439T Chr.17: 68984949-68984949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779954514 Chr.17: 68985079-68985079 on Build GRCh38
rs777149156 Chr.17: 68984940-68984940 on Build GRCh38
rs1039157013 Chr.17: 68985220-68985220 on Build GRCh38
rs749973044 Chr.17: 68984876-68984876 on Build GRCh38
rs531633502 Chr.17: 68984981-68984981 on Build GRCh38
rs774078887 Chr.17: 68985100-68985100 on Build GRCh38
rs899312658 Chr.17: 68985221-68985221 on Build GRCh38
rs756524839 Chr.17: 68984947-68984947 on Build GRCh38
rs759020837 Chr.17: 68984884-68984900 on Build GRCh38
rs745549439 Chr.17: 68985180-68985180 on Build GRCh38
rs959185304 Chr.17: 68984863-68984863 on Build GRCh38
rs747057241 Chr.17: 68985149-68985149 on Build GRCh38
rs780194096 Chr.17: 68985141-68985141 on Build GRCh38
rs765874813 Chr.17: 68984923-68984923 on Build GRCh38
rs912110817 Chr.17: 68985186-68985186 on Build GRCh38
rs371951269 Chr.17: 68985009-68985009 on Build GRCh38
rs752091037 Chr.17: 68985113-68985113 on Build GRCh38
rs760650878 Chr.17: 68984839-68984839 on Build GRCh38
rs748545976 Chr.17: 68984899-68984899 on Build GRCh38
rs950641720 Chr.17: 68984850-68984850 on Build GRCh38
rs577137804 Chr.17: 68985224-68985224 on Build GRCh38
rs754240514 Chr.17: 68985052-68985052 on Build GRCh38
rs751113808 Chr.17: 68985020-68985020 on Build GRCh38
rs201261747 Chr.17: 68984977-68984977 on Build GRCh38
rs373607799 Chr.17: 68985162-68985162 on Build GRCh38
rs762340627 Chr.17: 68985031-68985033 on Build GRCh38
rs772210715 Chr.17: 68985074-68985074 on Build GRCh38
rs184929537 Chr.17: 68985041-68985041 on Build GRCh38
rs755506671 Chr.17: 68984928-68984928 on Build GRCh38
rs145682636 Chr.17: 68984885-68984885 on Build GRCh38
rs747542908 Chr.17: 68984890-68984890 on Build GRCh38
rs762707600 Chr.17: 68985088-68985088 on Build GRCh38
rs558789331 Chr.17: 68985169-68985169 on Build GRCh38
rs750094359 Chr.17: 68984920-68984920 on Build GRCh38
rs758480737 Chr.17: 68985140-68985140 on Build GRCh38
rs756513068 Chr.17: 68985048-68985048 on Build GRCh38
rs368711218 Chr.17: 68985049-68985049 on Build GRCh38
rs781705697 Chr.17: 68984933-68984933 on Build GRCh38
rs148120108 Chr.17: 68984917-68984917 on Build GRCh38
rs968853350 Chr.17: 68985123-68985123 on Build GRCh38
rs770758250 Chr.17: 68985257-68985257 on Build GRCh38
rs369324391 Chr.17: 68984982-68984982 on Build GRCh38
rs764223398 Chr.17: 68985208-68985208 on Build GRCh38
rs61732707 Chr.17: 68985093-68985093 on Build GRCh38
rs200241314 Chr.17: 68985086-68985086 on Build GRCh38
rs117967396 Chr.17: 68984891-68984891 on Build GRCh38
rs114366393 Chr.17: 68985116-68985116 on Build GRCh38
rs759068181 Chr.17: 68985026-68985026 on Build GRCh38
rs776357070 Chr.17: 68984914-68984914 on Build GRCh38
rs779117760 Chr.17: 68984857-68984857 on Build GRCh38
rs766950349 Chr.17: 68984926-68984927 on Build GRCh38
rs547391943 Chr.17: 68985056-68985056 on Build GRCh38
rs750843570 Chr.17: 68985052-68985052 on Build GRCh38
rs994930758 Chr.17: 68985008-68985008 on Build GRCh38
rs766888962 Chr.17: 68985028-68985028 on Build GRCh38
rs748514372 Chr.17: 68984944-68984944 on Build GRCh38
rs374600027 Chr.17: 68985058-68985058 on Build GRCh38
rs759070757 Chr.17: 68985106-68985106 on Build GRCh38
rs749472643 Chr.17: 68984901-68984901 on Build GRCh38
rs773952285 Chr.17: 68985179-68985179 on Build GRCh38
rs763802575 Chr.17: 68984843-68984843 on Build GRCh38
rs764691635 Chr.17: 68985117-68985117 on Build GRCh38
rs774507778 Chr.17: 68984904-68984904 on Build GRCh38
rs748118668 Chr.17: 68985084-68985084 on Build GRCh38
rs778243414 Chr.17: 68984966-68984966 on Build GRCh38
rs527659354 Chr.17: 68984987-68984987 on Build GRCh38
rs142934607 Chr.17: 68984921-68984921 on Build GRCh38
rs753609304 Chr.17: 68984853-68984853 on Build GRCh38
rs138791482 Chr.17: 68984819-68984819 on Build GRCh38
rs763017395 Chr.17: 68985172-68985172 on Build GRCh38
rs764937873 Chr.17: 68984869-68984869 on Build GRCh38
rs750686461 Chr.17: 68985139-68985139 on Build GRCh38
rs75215219 Chr.17: 68984878-68984878 on Build GRCh38
rs202172685 Chr.17: 68984854-68984854 on Build GRCh38
rs756761350 Chr.17: 68984848-68984849 on Build GRCh38
rs768516352 Chr.17: 68985001-68985001 on Build GRCh38
rs772727822 Chr.17: 68984824-68984824 on Build GRCh38
rs148927389 Chr.17: 68984907-68984907 on Build GRCh38
rs779245030 Chr.17: 68985057-68985057 on Build GRCh38
rs757672749 Chr.17: 68985120-68985120 on Build GRCh38
rs779191603 Chr.17: 68984986-68984986 on Build GRCh38
rs932208529 Chr.17: 68985223-68985223 on Build GRCh38
rs1051967678 Chr.17: 68985182-68985182 on Build GRCh38
rs769605774 Chr.17: 68985017-68985017 on Build GRCh38
rs779367534 Chr.17: 68985131-68985131 on Build GRCh38
rs913275332 Chr.17: 68985166-68985166 on Build GRCh38
rs181800371 Chr.17: 68985018-68985018 on Build GRCh38
rs28590334 Chr.17: 68985085-68985085 on Build GRCh38
rs761637364 Chr.17: 68984916-68984916 on Build GRCh38
rs371992345 Chr.17: 68984905-68984905 on Build GRCh38
rs376636691 Chr.17: 68985014-68985014 on Build GRCh38
rs201848433 Chr.17: 68985151-68985151 on Build GRCh38
rs765561566 Chr.17: 68985041-68985041 on Build GRCh38
rs781470283 Chr.17: 68984897-68984897 on Build GRCh38
rs34798261 Chr.17: 68984997-68984998 on Build GRCh38
rs751669932 Chr.17: 68985210-68985210 on Build GRCh38
rs1007640417 Chr.17: 68984924-68984924 on Build GRCh38
rs767059980 Chr.17: 68985112-68985112 on Build GRCh38
rs980032657 Chr.17: 68984842-68984842 on Build GRCh38

View Full Product Details