Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69148458-69149000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4439217 c.4618C>A p.Q1540K Chr.17: 69148841-69148841 on Build GRCh38
4439216 c.4619A>G p.Q1540R Chr.17: 69148840-69148840 on Build GRCh38
983559 c.4543G>T p.E1515* Chr.17: 69148916-69148916 on Build GRCh38
3521431 c.4541T>G p.L1514* Chr.17: 69148918-69148918 on Build GRCh38
1385544 c.4584A>C p.K1528N Chr.17: 69148875-69148875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768928197 Chr.17: 69148898-69148898 on Build GRCh38
rs780252647 Chr.17: 69148759-69148759 on Build GRCh38
rs755895861 Chr.17: 69148832-69148832 on Build GRCh38
rs958760667 Chr.17: 69148516-69148516 on Build GRCh38
rs747181599 Chr.17: 69148908-69148909 on Build GRCh38
rs910323000 Chr.17: 69148649-69148649 on Build GRCh38
rs988771561 Chr.17: 69148762-69148762 on Build GRCh38
rs1974576 Chr.17: 69148487-69148487 on Build GRCh38
rs781271701 Chr.17: 69148950-69148951 on Build GRCh38
rs370052354 Chr.17: 69148654-69148654 on Build GRCh38
rs372285641 Chr.17: 69148817-69148817 on Build GRCh38
rs780439909 Chr.17: 69148891-69148891 on Build GRCh38
rs753933098 Chr.17: 69148782-69148782 on Build GRCh38
rs758811477 Chr.17: 69148867-69148867 on Build GRCh38
rs187802376 Chr.17: 69148853-69148853 on Build GRCh38
rs869194608 Chr.17: 69148491-69148492 on Build GRCh38
rs867312556 Chr.17: 69148760-69148760 on Build GRCh38
rs768838179 Chr.17: 69148783-69148783 on Build GRCh38
rs544458844 Chr.17: 69148528-69148528 on Build GRCh38
rs761872076 Chr.17: 69148792-69148792 on Build GRCh38
rs571728264 Chr.17: 69148823-69148823 on Build GRCh38
rs756795824 Chr.17: 69148850-69148850 on Build GRCh38
rs796645386 Chr.17: 69148541-69148543 on Build GRCh38
rs1974578 Chr.17: 69148743-69148743 on Build GRCh38
rs1014638134 Chr.17: 69148667-69148667 on Build GRCh38
rs549998966 Chr.17: 69148797-69148797 on Build GRCh38
rs1037531244 Chr.17: 69148658-69148661 on Build GRCh38
rs955108861 Chr.17: 69148950-69148950 on Build GRCh38
rs1004543061 Chr.17: 69148660-69148660 on Build GRCh38
rs750363023 Chr.17: 69148608-69148608 on Build GRCh38
rs763914842 Chr.17: 69148969-69148969 on Build GRCh38
rs1027590203 Chr.17: 69148612-69148612 on Build GRCh38
rs777553626 Chr.17: 69148833-69148833 on Build GRCh38
rs776855686 Chr.17: 69148910-69148910 on Build GRCh38
rs960427326 Chr.17: 69148669-69148669 on Build GRCh38
rs913260008 Chr.17: 69148773-69148773 on Build GRCh38
rs747372773 Chr.17: 69148893-69148893 on Build GRCh38
rs767556211 Chr.17: 69148960-69148960 on Build GRCh38
rs769935609 Chr.17: 69148940-69148940 on Build GRCh38
rs375323266 Chr.17: 69148856-69148856 on Build GRCh38
rs931837317 Chr.17: 69148939-69148939 on Build GRCh38
rs768986047 Chr.17: 69148941-69148942 on Build GRCh38
rs991938681 Chr.17: 69148673-69148673 on Build GRCh38
rs527253106 Chr.17: 69148714-69148714 on Build GRCh38
rs560110351 Chr.17: 69148593-69148593 on Build GRCh38
rs369349661 Chr.17: 69148934-69148934 on Build GRCh38
rs183170494 Chr.17: 69148725-69148725 on Build GRCh38
rs547731193 Chr.17: 69148979-69148979 on Build GRCh38
rs887400520 Chr.17: 69148622-69148622 on Build GRCh38
rs924916009 Chr.17: 69148927-69148930 on Build GRCh38
rs561170697 Chr.17: 69148540-69148540 on Build GRCh38
rs143114731 Chr.17: 69148788-69148788 on Build GRCh38
rs991696436 Chr.17: 69148533-69148533 on Build GRCh38
rs958812201 Chr.17: 69148794-69148794 on Build GRCh38
rs756811172 Chr.17: 69148974-69148974 on Build GRCh38
rs977633171 Chr.17: 69148562-69148562 on Build GRCh38
rs34474344 Chr.17: 69148541-69148542 on Build GRCh38
rs980720243 Chr.17: 69148715-69148715 on Build GRCh38
rs746410212 Chr.17: 69148857-69148857 on Build GRCh38
rs576994177 Chr.17: 69148614-69148614 on Build GRCh38
rs924291747 Chr.17: 69148569-69148569 on Build GRCh38
rs759555190 Chr.17: 69148943-69148943 on Build GRCh38
rs1024451477 Chr.17: 69148677-69148677 on Build GRCh38
rs2233769 Chr.17: 69148541-69148541 on Build GRCh38
rs970267959 Chr.17: 69148711-69148711 on Build GRCh38
rs371513297 Chr.17: 69148965-69148965 on Build GRCh38
rs1974577 Chr.17: 69148543-69148543 on Build GRCh38
rs748198598 Chr.17: 69148962-69148962 on Build GRCh38
rs759339148 Chr.17: 69148808-69148808 on Build GRCh38
rs748273420 Chr.17: 69148931-69148931 on Build GRCh38
rs753483710 Chr.17: 69148842-69148842 on Build GRCh38
rs751543519 Chr.17: 69148805-69148805 on Build GRCh38

View Full Product Details