Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69154910-69155449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353417 c.3651G>T p.K1217N Chr.17: 69155062-69155062 on Build GRCh38
2975569 c.3596G>A p.S1199N Chr.17: 69155117-69155117 on Build GRCh38
4391401 c.3647G>C p.R1216T Chr.17: 69155066-69155066 on Build GRCh38
983565 c.3692A>C p.K1231T Chr.17: 69155021-69155021 on Build GRCh38
6002604 c.3694G>T p.G1232C Chr.17: 69155019-69155019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929200830 Chr.17: 69155406-69155406 on Build GRCh38
rs1052942425 Chr.17: 69155109-69155109 on Build GRCh38
rs541229332 Chr.17: 69155285-69155285 on Build GRCh38
rs781424475 Chr.17: 69155083-69155086 on Build GRCh38
rs566042247 Chr.17: 69155294-69155294 on Build GRCh38
rs565483430 Chr.17: 69155225-69155225 on Build GRCh38
rs951810148 Chr.17: 69154973-69154973 on Build GRCh38
rs869302689 Chr.17: 69155085-69155085 on Build GRCh38
rs146573339 Chr.17: 69155018-69155018 on Build GRCh38
rs778271547 Chr.17: 69155085-69155085 on Build GRCh38
rs1047097214 Chr.17: 69155417-69155417 on Build GRCh38
rs541888004 Chr.17: 69155273-69155273 on Build GRCh38
rs774580071 Chr.17: 69155120-69155120 on Build GRCh38
rs749623525 Chr.17: 69155114-69155114 on Build GRCh38
rs370449985 Chr.17: 69155052-69155052 on Build GRCh38
rs943320544 Chr.17: 69155019-69155019 on Build GRCh38
rs547832219 Chr.17: 69155255-69155255 on Build GRCh38
rs765126733 Chr.17: 69155016-69155016 on Build GRCh38
rs964932478 Chr.17: 69155299-69155299 on Build GRCh38
rs767539202 Chr.17: 69155011-69155011 on Build GRCh38
rs746050593 Chr.17: 69155134-69155134 on Build GRCh38
rs993505058 Chr.17: 69155009-69155009 on Build GRCh38
rs980104561 Chr.17: 69155038-69155038 on Build GRCh38
rs536751417 Chr.17: 69155315-69155315 on Build GRCh38
rs961844937 Chr.17: 69155415-69155415 on Build GRCh38
rs754216537 Chr.17: 69155019-69155020 on Build GRCh38
rs910401200 Chr.17: 69155288-69155288 on Build GRCh38
rs776583054 Chr.17: 69155012-69155012 on Build GRCh38
rs936109155 Chr.17: 69155067-69155067 on Build GRCh38
rs998509578 Chr.17: 69155248-69155248 on Build GRCh38
rs200537594 Chr.17: 69154991-69154991 on Build GRCh38
rs373780903 Chr.17: 69155311-69155314 on Build GRCh38
rs968816839 Chr.17: 69154951-69154951 on Build GRCh38
rs149485519 Chr.17: 69155309-69155312 on Build GRCh38
rs968991993 Chr.17: 69155245-69155245 on Build GRCh38
rs1035878341 Chr.17: 69155166-69155166 on Build GRCh38
rs1040698623 Chr.17: 69155163-69155163 on Build GRCh38
rs759625765 Chr.17: 69155010-69155010 on Build GRCh38
rs771057238 Chr.17: 69154993-69154993 on Build GRCh38
rs1031266142 Chr.17: 69155252-69155252 on Build GRCh38
rs766130666 Chr.17: 69155037-69155037 on Build GRCh38
rs755309963 Chr.17: 69155054-69155055 on Build GRCh38
rs188308482 Chr.17: 69155254-69155254 on Build GRCh38
rs887509237 Chr.17: 69155264-69155264 on Build GRCh38
rs768599826 Chr.17: 69155006-69155006 on Build GRCh38
rs1048798869 Chr.17: 69155215-69155215 on Build GRCh38
rs761868194 Chr.17: 69155182-69155182 on Build GRCh38
rs199584550 Chr.17: 69155188-69155188 on Build GRCh38
rs1022316751 Chr.17: 69155226-69155226 on Build GRCh38
rs751195093 Chr.17: 69155040-69155040 on Build GRCh38
rs779948271 Chr.17: 69154989-69154989 on Build GRCh38
rs748551218 Chr.17: 69154971-69154971 on Build GRCh38
rs75435520 Chr.17: 69155153-69155153 on Build GRCh38
rs561157424 Chr.17: 69155116-69155116 on Build GRCh38
rs756568298 Chr.17: 69155079-69155079 on Build GRCh38
rs765080845 Chr.17: 69155187-69155187 on Build GRCh38
rs372530243 Chr.17: 69155013-69155013 on Build GRCh38
rs780806475 Chr.17: 69155065-69155065 on Build GRCh38
rs1002918503 Chr.17: 69155139-69155139 on Build GRCh38
rs72853603 Chr.17: 69155066-69155066 on Build GRCh38
rs1005717470 Chr.17: 69154986-69154986 on Build GRCh38
rs751881385 Chr.17: 69155027-69155028 on Build GRCh38
rs1013071736 Chr.17: 69155211-69155211 on Build GRCh38
rs548652665 Chr.17: 69155401-69155401 on Build GRCh38
rs770237859 Chr.17: 69154978-69154978 on Build GRCh38
rs758135672 Chr.17: 69155020-69155020 on Build GRCh38

View Full Product Details