Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74862374-74862866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684285 c.1475T>C p.L492P Chr.17: 74862836-74862836 on Build GRCh38
3521928 c.1489C>T p.H497Y Chr.17: 74862822-74862822 on Build GRCh38
3672570 c.1464T>G p.P488P Chr.17: 74862847-74862847 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746212359 Chr.17: 74862439-74862439 on Build GRCh38
rs962805963 Chr.17: 74862681-74862681 on Build GRCh38
rs564572063 Chr.17: 74862801-74862801 on Build GRCh38
rs541132854 Chr.17: 74862565-74862565 on Build GRCh38
rs895297808 Chr.17: 74862714-74862714 on Build GRCh38
rs34607406 Chr.17: 74862587-74862587 on Build GRCh38
rs546460456 Chr.17: 74862748-74862748 on Build GRCh38
rs184434227 Chr.17: 74862699-74862699 on Build GRCh38
rs957925580 Chr.17: 74862463-74862463 on Build GRCh38
rs866764736 Chr.17: 74862651-74862651 on Build GRCh38
rs1022454818 Chr.17: 74862645-74862645 on Build GRCh38
rs1004001338 Chr.17: 74862514-74862514 on Build GRCh38
rs376002306 Chr.17: 74862816-74862816 on Build GRCh38
rs955080990 Chr.17: 74862653-74862654 on Build GRCh38
rs572245124 Chr.17: 74862516-74862516 on Build GRCh38
rs756866241 Chr.17: 74862802-74862802 on Build GRCh38
rs989369555 Chr.17: 74862481-74862481 on Build GRCh38
rs779966131 Chr.17: 74862813-74862813 on Build GRCh38
rs1011019409 Chr.17: 74862601-74862601 on Build GRCh38
rs559997810 Chr.17: 74862757-74862757 on Build GRCh38
rs528650611 Chr.17: 74862784-74862784 on Build GRCh38
rs1045085002 Chr.17: 74862488-74862488 on Build GRCh38
rs753252635 Chr.17: 74862791-74862791 on Build GRCh38
rs973893521 Chr.17: 74862708-74862708 on Build GRCh38
rs773424540 Chr.17: 74862777-74862777 on Build GRCh38
rs192928186 Chr.17: 74862498-74862498 on Build GRCh38
rs982916107 Chr.17: 74862669-74862669 on Build GRCh38
rs768341015 Chr.17: 74862613-74862613 on Build GRCh38
rs1006478424 Chr.17: 74862406-74862406 on Build GRCh38
rs143458938 Chr.17: 74862834-74862834 on Build GRCh38
rs748385798 Chr.17: 74862800-74862801 on Build GRCh38
rs781398098 Chr.17: 74862781-74862782 on Build GRCh38
rs933408197 Chr.17: 74862628-74862628 on Build GRCh38
rs538853708 Chr.17: 74862422-74862422 on Build GRCh38
rs1016415039 Chr.17: 74862408-74862408 on Build GRCh38
rs534647083 Chr.17: 74862806-74862806 on Build GRCh38
rs368374398 Chr.17: 74862788-74862788 on Build GRCh38
rs543439770 Chr.17: 74862677-74862677 on Build GRCh38
rs1024749397 Chr.17: 74862430-74862430 on Build GRCh38
rs570747783 Chr.17: 74862646-74862646 on Build GRCh38
rs778010849 Chr.17: 74862817-74862817 on Build GRCh38
rs761138033 Chr.17: 74862800-74862800 on Build GRCh38
rs35033646 Chr.17: 74862678-74862678 on Build GRCh38
rs1012455715 Chr.17: 74862718-74862718 on Build GRCh38
rs1031088099 Chr.17: 74862452-74862452 on Build GRCh38
rs911044769 Chr.17: 74862497-74862497 on Build GRCh38
rs375440525 Chr.17: 74862780-74862780 on Build GRCh38
rs946843655 Chr.17: 74862790-74862790 on Build GRCh38
rs948650944 Chr.17: 74862478-74862479 on Build GRCh38
rs906243867 Chr.17: 74862739-74862739 on Build GRCh38
rs768032757 Chr.17: 74862789-74862789 on Build GRCh38
rs558945542 Chr.17: 74862410-74862410 on Build GRCh38
rs948578098 Chr.17: 74862747-74862747 on Build GRCh38
rs569829644 Chr.17: 74862470-74862471 on Build GRCh38
rs976577363 Chr.17: 74862810-74862810 on Build GRCh38
rs773444973 Chr.17: 74862444-74862444 on Build GRCh38
rs768871114 Chr.17: 74862755-74862755 on Build GRCh38
rs892602238 Chr.17: 74862590-74862590 on Build GRCh38
rs970796154 Chr.17: 74862434-74862434 on Build GRCh38
rs865791521 Chr.17: 74862752-74862752 on Build GRCh38
rs749319199 Chr.17: 74862831-74862831 on Build GRCh38
rs771085930 Chr.17: 74862833-74862833 on Build GRCh38
rs1037986896 Chr.17: 74862683-74862683 on Build GRCh38
rs975232572 Chr.17: 74862612-74862612 on Build GRCh38
rs941222363 Chr.17: 74862435-74862435 on Build GRCh38
rs1025136017 Chr.17: 74862719-74862719 on Build GRCh38
rs1012396026 Chr.17: 74862424-74862424 on Build GRCh38
rs780022072 Chr.17: 74862655-74862655 on Build GRCh38
rs369079771 Chr.17: 74862479-74862480 on Build GRCh38
rs770037026 Chr.17: 74862766-74862766 on Build GRCh38
rs758322347 Chr.17: 74862808-74862808 on Build GRCh38
rs747012697 Chr.17: 74862814-74862814 on Build GRCh38
rs34015823 Chr.17: 74862469-74862469 on Build GRCh38
rs747210043 Chr.17: 74862819-74862819 on Build GRCh38
rs981362714 Chr.17: 74862751-74862751 on Build GRCh38
rs184862339 Chr.17: 74862415-74862415 on Build GRCh38

View Full Product Details