Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74866040-74866537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6386890 c.371A>G p.Q124R Chr.17: 74866135-74866135 on Build GRCh38
6390835 c.353A>C p.N118T Chr.17: 74866153-74866153 on Build GRCh38
6398526 c.334G>A p.A112T Chr.17: 74866172-74866172 on Build GRCh38
6401395 c.382G>T p.A128S Chr.17: 74866457-74866457 on Build GRCh38
6314385 c.242C>A p.A81D Chr.17: 74866465-74866465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986280345 Chr.17: 74866346-74866346 on Build GRCh38
rs752675360 Chr.17: 74866175-74866175 on Build GRCh38
rs200936845 Chr.17: 74866223-74866223 on Build GRCh38
rs753518416 Chr.17: 74866153-74866153 on Build GRCh38
rs753084863 Chr.17: 74866130-74866130 on Build GRCh38
rs761558804 Chr.17: 74866409-74866409 on Build GRCh38
rs918940252 Chr.17: 74866402-74866402 on Build GRCh38
rs690514 Chr.17: 74866471-74866471 on Build GRCh38
rs374082956 Chr.17: 74866187-74866187 on Build GRCh38
rs776221918 Chr.17: 74866398-74866398 on Build GRCh38
rs760056199 Chr.17: 74866204-74866204 on Build GRCh38
rs374317439 Chr.17: 74866458-74866458 on Build GRCh38
rs527632348 Chr.17: 74866301-74866301 on Build GRCh38
rs376312654 Chr.17: 74866143-74866143 on Build GRCh38
rs371202127 Chr.17: 74866239-74866239 on Build GRCh38
rs769402154 Chr.17: 74866490-74866490 on Build GRCh38
rs953853087 Chr.17: 74866302-74866302 on Build GRCh38
rs368300344 Chr.17: 74866385-74866385 on Build GRCh38
rs997615675 Chr.17: 74866096-74866096 on Build GRCh38
rs748413291 Chr.17: 74866113-74866113 on Build GRCh38
rs975722496 Chr.17: 74866389-74866389 on Build GRCh38
rs774882335 Chr.17: 74866125-74866125 on Build GRCh38
rs369049042 Chr.17: 74866480-74866480 on Build GRCh38
rs779256067 Chr.17: 74866249-74866249 on Build GRCh38
rs756192203 Chr.17: 74866236-74866236 on Build GRCh38
rs963933595 Chr.17: 74866310-74866310 on Build GRCh38
rs377030891 Chr.17: 74866093-74866093 on Build GRCh38
rs375878174 Chr.17: 74866341-74866341 on Build GRCh38
rs760380314 Chr.17: 74866145-74866145 on Build GRCh38
rs373706276 Chr.17: 74866081-74866081 on Build GRCh38
rs201386849 Chr.17: 74866119-74866119 on Build GRCh38
rs776491969 Chr.17: 74866211-74866211 on Build GRCh38
rs772657420 Chr.17: 74866265-74866265 on Build GRCh38
rs146885267 Chr.17: 74866479-74866479 on Build GRCh38
rs372414429 Chr.17: 74866262-74866262 on Build GRCh38
rs984749438 Chr.17: 74866457-74866457 on Build GRCh38
rs376477275 Chr.17: 74866435-74866435 on Build GRCh38
rs757157557 Chr.17: 74866162-74866162 on Build GRCh38
rs550083102 Chr.17: 74866464-74866464 on Build GRCh38
rs772395253 Chr.17: 74866194-74866194 on Build GRCh38
rs770691402 Chr.17: 74866514-74866514 on Build GRCh38
rs542442019 Chr.17: 74866182-74866182 on Build GRCh38
rs766174200 Chr.17: 74866226-74866226 on Build GRCh38
rs1024259725 Chr.17: 74866308-74866308 on Build GRCh38
rs183154538 Chr.17: 74866515-74866515 on Build GRCh38
rs759100178 Chr.17: 74866083-74866083 on Build GRCh38
rs764175967 Chr.17: 74866456-74866456 on Build GRCh38
rs777828005 Chr.17: 74866238-74866238 on Build GRCh38
rs374337519 Chr.17: 74866098-74866098 on Build GRCh38
rs760036190 Chr.17: 74866127-74866127 on Build GRCh38
rs965793415 Chr.17: 74866386-74866386 on Build GRCh38
rs769528827 Chr.17: 74866414-74866414 on Build GRCh38
rs773160243 Chr.17: 74866420-74866420 on Build GRCh38
rs372200621 Chr.17: 74866092-74866092 on Build GRCh38
rs557781696 Chr.17: 74866110-74866110 on Build GRCh38
rs894527866 Chr.17: 74866484-74866484 on Build GRCh38
rs372205255 Chr.17: 74866429-74866429 on Build GRCh38
rs375196494 Chr.17: 74866123-74866123 on Build GRCh38
rs753620788 Chr.17: 74866094-74866094 on Build GRCh38
rs7223627 Chr.17: 74866491-74866491 on Build GRCh38
rs551451161 Chr.17: 74866485-74866485 on Build GRCh38
rs766909104 Chr.17: 74866091-74866091 on Build GRCh38
rs143778881 Chr.17: 74866179-74866179 on Build GRCh38
rs1055925083 Chr.17: 74866270-74866270 on Build GRCh38
rs781596335 Chr.17: 74866110-74866111 on Build GRCh38
rs757503961 Chr.17: 74866460-74866460 on Build GRCh38
rs368862112 Chr.17: 74866247-74866247 on Build GRCh38
rs923683362 Chr.17: 74866426-74866426 on Build GRCh38
rs200778123 Chr.17: 74866208-74866208 on Build GRCh38
rs751496458 Chr.17: 74866235-74866235 on Build GRCh38
rs909458140 Chr.17: 74866117-74866117 on Build GRCh38
rs553757022 Chr.17: 74866122-74866122 on Build GRCh38
rs370741969 Chr.17: 74866174-74866174 on Build GRCh38
rs368931937 Chr.17: 74866222-74866222 on Build GRCh38
rs138454707 Chr.17: 74866219-74866219 on Build GRCh38
rs748969330 Chr.17: 74866509-74866509 on Build GRCh38
rs767115077 Chr.17: 74866130-74866130 on Build GRCh38
rs762974345 Chr.17: 74866225-74866225 on Build GRCh38
rs564944791 Chr.17: 74866292-74866292 on Build GRCh38
rs750442269 Chr.17: 74866170-74866170 on Build GRCh38
rs910191001 Chr.17: 74866213-74866213 on Build GRCh38
rs914970965 Chr.17: 74866307-74866325 on Build GRCh38
rs547705555 Chr.17: 74866332-74866332 on Build GRCh38
rs766169513 Chr.17: 74866431-74866431 on Build GRCh38
rs761542421 Chr.17: 74866216-74866216 on Build GRCh38
rs775097448 Chr.17: 74866199-74866199 on Build GRCh38
rs537929036 Chr.17: 74866472-74866472 on Build GRCh38
rs910335934 Chr.17: 74866347-74866347 on Build GRCh38
rs772598174 Chr.17: 74866315-74866315 on Build GRCh38
rs757282111 Chr.17: 74866106-74866106 on Build GRCh38
rs765117011 Chr.17: 74866167-74866167 on Build GRCh38
rs756333157 Chr.17: 74866228-74866229 on Build GRCh38
rs369416041 Chr.17: 74866266-74866266 on Build GRCh38
rs746389904 Chr.17: 74866191-74866191 on Build GRCh38
rs571288519 Chr.17: 74866486-74866486 on Build GRCh38
rs991100246 Chr.17: 74866326-74866326 on Build GRCh38
rs747546591 Chr.17: 74866291-74866291 on Build GRCh38
rs756377654 Chr.17: 74866178-74866178 on Build GRCh38
rs201911602 Chr.17: 74866152-74866152 on Build GRCh38
rs199988623 Chr.17: 74866176-74866176 on Build GRCh38
rs999074147 Chr.17: 74866324-74866324 on Build GRCh38
rs149831556 Chr.17: 74866114-74866114 on Build GRCh38
rs370799304 Chr.17: 74866245-74866245 on Build GRCh38
rs781115825 Chr.17: 74866111-74866111 on Build GRCh38

View Full Product Details